Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 64400554-64400953 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIP4
Entrez Gene ID
9325
Gene Name
thyroid hormone receptor interactor 4
Gene Aliases
ASC-1, ASC1, HsT17391, SMABF1, ZC2HC5
UniGene
Hs.500340
Gene Chromosome Location
Chr.15: 64387804-64455303 on Build GRCh38
Transcripts
NM_001321924.1 NM_016213.4 NR_135855.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
963943 c.697G>T p.G233* Chr.15: 64400821-64400821 on Build GRCh38
6554997 c.668G>T p.S223I Chr.15: 64400792-64400792 on Build GRCh38
5649658 c.647T>C p.L216S Chr.15: 64400771-64400771 on Build GRCh38
6808906 c.643A>G p.I215V Chr.15: 64400767-64400767 on Build GRCh38
299568 c.653G>A p.R218H Chr.15: 64400777-64400777 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs372298448 Chr.15: 64400742-64400742 on Build GRCh38
rs985208996 Chr.15: 64400909-64400909 on Build GRCh38
rs1000295815 Chr.15: 64400923-64400923 on Build GRCh38
rs774095998 Chr.15: 64400789-64400789 on Build GRCh38
rs781617873 Chr.15: 64400788-64400789 on Build GRCh38
rs1045879991 Chr.15: 64400691-64400691 on Build GRCh38
rs950198198 Chr.15: 64400649-64400649 on Build GRCh38
rs545850539 Chr.15: 64400692-64400692 on Build GRCh38
rs763450897 Chr.15: 64400856-64400856 on Build GRCh38
rs755206322 Chr.15: 64400721-64400726 on Build GRCh38
rs767041423 Chr.15: 64400738-64400738 on Build GRCh38
rs182507164 Chr.15: 64400868-64400868 on Build GRCh38
rs929979393 Chr.15: 64400727-64400727 on Build GRCh38
rs368048455 Chr.15: 64400877-64400877 on Build GRCh38
rs760256538 Chr.15: 64400728-64400728 on Build GRCh38
rs749130311 Chr.15: 64400779-64400779 on Build GRCh38
rs77997699 Chr.15: 64400849-64400849 on Build GRCh38
rs1039729585 Chr.15: 64400853-64400853 on Build GRCh38
rs150491308 Chr.15: 64400851-64400851 on Build GRCh38
rs751922652 Chr.15: 64400864-64400864 on Build GRCh38
rs377491660 Chr.15: 64400724-64400724 on Build GRCh38
rs373011414 Chr.15: 64400872-64400872 on Build GRCh38
rs766298673 Chr.15: 64400698-64400702 on Build GRCh38
rs377304404 Chr.15: 64400802-64400802 on Build GRCh38
rs373910178 Chr.15: 64400782-64400782 on Build GRCh38
rs751724846 Chr.15: 64400718-64400719 on Build GRCh38
rs935689690 Chr.15: 64400604-64400604 on Build GRCh38
rs764363211 Chr.15: 64400768-64400768 on Build GRCh38
rs768066258 Chr.15: 64400869-64400869 on Build GRCh38
rs190677062 Chr.15: 64400632-64400632 on Build GRCh38
rs774253650 Chr.15: 64400700-64400700 on Build GRCh38
rs112913281 Chr.15: 64400732-64400732 on Build GRCh38
rs755471393 Chr.15: 64400750-64400750 on Build GRCh38
rs1042041262 Chr.15: 64400875-64400875 on Build GRCh38
rs748194484 Chr.15: 64400765-64400765 on Build GRCh38
rs897076408 Chr.15: 64400749-64400749 on Build GRCh38
rs377226534 Chr.15: 64400861-64400861 on Build GRCh38
rs747689856 Chr.15: 64400822-64400822 on Build GRCh38
rs759090288 Chr.15: 64400701-64400701 on Build GRCh38
rs989064416 Chr.15: 64400622-64400622 on Build GRCh38
rs903088553 Chr.15: 64400904-64400904 on Build GRCh38
rs763708447 Chr.15: 64400848-64400848 on Build GRCh38
rs867666933 Chr.15: 64400787-64400787 on Build GRCh38
rs771989879 Chr.15: 64400805-64400805 on Build GRCh38
rs937435828 Chr.15: 64400704-64400704 on Build GRCh38
rs781152404 Chr.15: 64400762-64400762 on Build GRCh38
rs766970697 Chr.15: 64400717-64400717 on Build GRCh38
rs777959222 Chr.15: 64400777-64400777 on Build GRCh38
rs529158551 Chr.15: 64400884-64400884 on Build GRCh38
rs982772315 Chr.15: 64400686-64400686 on Build GRCh38
rs374201058 Chr.15: 64400734-64400734 on Build GRCh38
rs146954502 Chr.15: 64400784-64400784 on Build GRCh38
rs756197581 Chr.15: 64400873-64400873 on Build GRCh38
rs954053331 Chr.15: 64400889-64400889 on Build GRCh38
rs913500572 Chr.15: 64400626-64400626 on Build GRCh38
rs756359639 Chr.15: 64400776-64400776 on Build GRCh38
rs373712602 Chr.15: 64400718-64400718 on Build GRCh38
rs774864951 Chr.15: 64400816-64400816 on Build GRCh38
rs916740327 Chr.15: 64400625-64400625 on Build GRCh38
rs760224952 Chr.15: 64400835-64400835 on Build GRCh38

View Full Product Details