Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 89210609-89211105 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RLBP1
Entrez Gene ID
6017
Gene Name
retinaldehyde binding protein 1
Gene Aliases
CRALBP
UniGene
Hs.1933
Gene Chromosome Location
Chr.15: 89209867-89223179 on Build GRCh38
Transcripts
NM_000326.4 XM_011521870.2 XM_017022460.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5646658 c.705C>G p.F235L Chr.15: 89210789-89210789 on Build GRCh38
5697817 c.690C>A p.S230S Chr.15: 89210804-89210804 on Build GRCh38
2015183 c.711C>T p.A237A Chr.15: 89210783-89210783 on Build GRCh38
4597444 c.733T>C p.W245R Chr.15: 89210761-89210761 on Build GRCh38
4917005 c.714C>A p.I238I Chr.15: 89210780-89210780 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746386544 Chr.15: 89210669-89210669 on Build GRCh38
rs867173110 Chr.15: 89211035-89211035 on Build GRCh38
rs1010601961 Chr.15: 89210689-89210689 on Build GRCh38
rs370900681 Chr.15: 89210648-89210648 on Build GRCh38
rs762020753 Chr.15: 89210732-89210732 on Build GRCh38
rs76369173 Chr.15: 89211083-89211083 on Build GRCh38
rs530126740 Chr.15: 89210666-89210666 on Build GRCh38
rs893287831 Chr.15: 89211008-89211008 on Build GRCh38
rs577938532 Chr.15: 89210978-89210978 on Build GRCh38
rs367775127 Chr.15: 89210825-89210825 on Build GRCh38
rs773171161 Chr.15: 89210832-89210832 on Build GRCh38
rs774546810 Chr.15: 89210715-89210715 on Build GRCh38
rs557342006 Chr.15: 89210654-89210654 on Build GRCh38
rs765136210 Chr.15: 89210854-89210854 on Build GRCh38
rs144986240 Chr.15: 89211080-89211080 on Build GRCh38
rs201148370 Chr.15: 89210683-89210683 on Build GRCh38
rs373610746 Chr.15: 89210649-89210649 on Build GRCh38
rs980907450 Chr.15: 89210917-89210917 on Build GRCh38
rs566750493 Chr.15: 89210886-89210886 on Build GRCh38
rs28933990 Chr.15: 89210794-89210794 on Build GRCh38
rs761622139 Chr.15: 89210691-89210691 on Build GRCh38
rs929270614 Chr.15: 89210999-89210999 on Build GRCh38
rs917632510 Chr.15: 89210902-89210902 on Build GRCh38
rs72762628 Chr.15: 89210985-89210985 on Build GRCh38
rs769052745 Chr.15: 89210705-89210705 on Build GRCh38
rs767522649 Chr.15: 89210739-89210739 on Build GRCh38
rs890513377 Chr.15: 89211011-89211011 on Build GRCh38
rs746571986 Chr.15: 89210663-89210663 on Build GRCh38
rs923543047 Chr.15: 89210953-89210953 on Build GRCh38
rs1032409102 Chr.15: 89210761-89210761 on Build GRCh38
rs759669185 Chr.15: 89210747-89210747 on Build GRCh38
rs758050207 Chr.15: 89210650-89210650 on Build GRCh38
rs150318794 Chr.15: 89210793-89210793 on Build GRCh38
rs1021991545 Chr.15: 89210691-89210691 on Build GRCh38
rs794727900 Chr.15: 89210750-89210750 on Build GRCh38
rs1040598739 Chr.15: 89210652-89210652 on Build GRCh38
rs769502221 Chr.15: 89210824-89210824 on Build GRCh38
rs1037742501 Chr.15: 89210977-89210977 on Build GRCh38
rs751242555 Chr.15: 89210765-89210765 on Build GRCh38
rs949538609 Chr.15: 89210908-89210908 on Build GRCh38
rs969052414 Chr.15: 89210693-89210693 on Build GRCh38
rs984816764 Chr.15: 89210851-89210851 on Build GRCh38
rs777441605 Chr.15: 89210758-89210758 on Build GRCh38
rs943636493 Chr.15: 89210645-89210645 on Build GRCh38
rs942262533 Chr.15: 89210924-89210924 on Build GRCh38
rs1008036194 Chr.15: 89211023-89211023 on Build GRCh38
rs746316188 Chr.15: 89210792-89210792 on Build GRCh38
rs202155238 Chr.15: 89210682-89210682 on Build GRCh38
rs781696668 Chr.15: 89210668-89210668 on Build GRCh38
rs786205494 Chr.15: 89210721-89210721 on Build GRCh38
rs926913264 Chr.15: 89210921-89210921 on Build GRCh38
rs943348166 Chr.15: 89210922-89210922 on Build GRCh38
rs759514093 Chr.15: 89210833-89210833 on Build GRCh38
rs140131617 Chr.15: 89210748-89210748 on Build GRCh38
rs756740450 Chr.15: 89210769-89210769 on Build GRCh38
rs780578200 Chr.15: 89210778-89210778 on Build GRCh38
rs151141842 Chr.15: 89210741-89210741 on Build GRCh38
rs554944860 Chr.15: 89210813-89210813 on Build GRCh38
rs181863443 Chr.15: 89210684-89210684 on Build GRCh38
rs575472152 Chr.15: 89211066-89211066 on Build GRCh38
rs1053264529 Chr.15: 89211004-89211004 on Build GRCh38
rs535051712 Chr.15: 89210937-89210937 on Build GRCh38
rs371507954 Chr.15: 89210690-89210690 on Build GRCh38
rs1051807311 Chr.15: 89211007-89211007 on Build GRCh38
rs542235805 Chr.15: 89210971-89210971 on Build GRCh38
rs146462570 Chr.15: 89210795-89210795 on Build GRCh38
rs905409877 Chr.15: 89210636-89210636 on Build GRCh38
rs776264789 Chr.15: 89210666-89210667 on Build GRCh38
rs993982002 Chr.15: 89210678-89210678 on Build GRCh38
rs761966746 Chr.15: 89210817-89210817 on Build GRCh38
rs557384258 Chr.15: 89211039-89211039 on Build GRCh38

View Full Product Details