Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2056109-2056603 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSC2
Entrez Gene ID
7249
Gene Name
tuberous sclerosis 2
Gene Aliases
LAM, PPP1R160, TSC4
UniGene
Hs.90303
Gene Chromosome Location
Chr.16: 2047804-2088720 on Build GRCh38
Transcripts
NM_000548.4 NM_001077183.2 NM_001114382.2 NM_001318827.1 NM_001318829.1 NM_001318831.1 NM_001318832.1 XM_005255529.4 XM_005255531.4 XM_011522636.2 XM_011522637.2 XM_011522638.2 XM_011522639.2 XM_011522640.2 XM_017023615.1 XM_017023616.1 XM_017023617.1 XM_017023618.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7177626 c.600-4G>T p.? Chr.16: 2056192-2056192 on Build GRCh38
165201 c.629C>T p.A210V Chr.16: 2056225-2056225 on Build GRCh38
7349605 c.139_*del p.? Chr.16: 2050400-2070620 on Build GRCh38
3728816 c.600-5T>C p.? Chr.16: 2056191-2056191 on Build GRCh38
7282968 c.637delG p.V213fs*3 Chr.16: 2056233-2056233 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs573404958 Chr.16: 2056406-2056406 on Build GRCh38
rs539332756 Chr.16: 2056335-2056335 on Build GRCh38
rs1011954024 Chr.16: 2056303-2056303 on Build GRCh38
rs377733757 Chr.16: 2056153-2056153 on Build GRCh38
rs756588732 Chr.16: 2056253-2056253 on Build GRCh38
rs985614043 Chr.16: 2056526-2056526 on Build GRCh38
rs772932343 Chr.16: 2056550-2056550 on Build GRCh38
rs762659998 Chr.16: 2056229-2056229 on Build GRCh38
rs569908571 Chr.16: 2056304-2056304 on Build GRCh38
rs369708731 Chr.16: 2056181-2056181 on Build GRCh38
rs115009851 Chr.16: 2056473-2056473 on Build GRCh38
rs45517118 Chr.16: 2056242-2056242 on Build GRCh38
rs770911792 Chr.16: 2056272-2056272 on Build GRCh38
rs1011677082 Chr.16: 2056485-2056485 on Build GRCh38
rs374674912 Chr.16: 2056139-2056139 on Build GRCh38
rs45517116 Chr.16: 2056183-2056183 on Build GRCh38
rs755164905 Chr.16: 2056188-2056188 on Build GRCh38
rs567756494 Chr.16: 2056226-2056226 on Build GRCh38
rs1011471170 Chr.16: 2056389-2056389 on Build GRCh38
rs758038522 Chr.16: 2056169-2056169 on Build GRCh38
rs368386966 Chr.16: 2056219-2056219 on Build GRCh38
rs776448117 Chr.16: 2056155-2056155 on Build GRCh38
rs147196739 Chr.16: 2056224-2056224 on Build GRCh38
rs899893080 Chr.16: 2056477-2056477 on Build GRCh38
rs139929314 Chr.16: 2056215-2056215 on Build GRCh38
rs1022033890 Chr.16: 2056495-2056495 on Build GRCh38
rs929959820 Chr.16: 2056567-2056567 on Build GRCh38
rs371474410 Chr.16: 2056170-2056170 on Build GRCh38
rs1018210102 Chr.16: 2056145-2056145 on Build GRCh38
rs1018371812 Chr.16: 2056534-2056534 on Build GRCh38
rs776293094 Chr.16: 2056223-2056223 on Build GRCh38
rs878854120 Chr.16: 2056228-2056228 on Build GRCh38
rs978032015 Chr.16: 2056313-2056313 on Build GRCh38
rs371896390 Chr.16: 2056255-2056255 on Build GRCh38
rs918628156 Chr.16: 2056559-2056559 on Build GRCh38
rs868655249 Chr.16: 2056378-2056378 on Build GRCh38
rs45460197 Chr.16: 2056361-2056361 on Build GRCh38
rs751005367 Chr.16: 2056172-2056172 on Build GRCh38
rs756600331 Chr.16: 2056173-2056173 on Build GRCh38
rs562520936 Chr.16: 2056453-2056453 on Build GRCh38
rs534873651 Chr.16: 2056384-2056384 on Build GRCh38
rs372007135 Chr.16: 2056250-2056250 on Build GRCh38
rs1013400296 Chr.16: 2056577-2056577 on Build GRCh38
rs752391078 Chr.16: 2056168-2056168 on Build GRCh38
rs967996074 Chr.16: 2056179-2056179 on Build GRCh38
rs745769641 Chr.16: 2056293-2056293 on Build GRCh38
rs367881085 Chr.16: 2056269-2056269 on Build GRCh38
rs776686789 Chr.16: 2056283-2056283 on Build GRCh38
rs577202733 Chr.16: 2056164-2056164 on Build GRCh38
rs1011228500 Chr.16: 2056176-2056176 on Build GRCh38
rs779227767 Chr.16: 2056267-2056267 on Build GRCh38
rs45506197 Chr.16: 2056199-2056199 on Build GRCh38
rs572627212 Chr.16: 2056362-2056362 on Build GRCh38
rs773192790 Chr.16: 2056149-2056149 on Build GRCh38
rs201617383 Chr.16: 2056271-2056271 on Build GRCh38
rs111700202 Chr.16: 2056520-2056520 on Build GRCh38
rs764925296 Chr.16: 2056225-2056225 on Build GRCh38
rs553491113 Chr.16: 2056391-2056391 on Build GRCh38
rs527525473 Chr.16: 2056540-2056540 on Build GRCh38
rs755431618 Chr.16: 2056261-2056261 on Build GRCh38
rs941595244 Chr.16: 2056258-2056258 on Build GRCh38
rs899922912 Chr.16: 2056297-2056297 on Build GRCh38
rs772137982 Chr.16: 2056148-2056148 on Build GRCh38
rs45498496 Chr.16: 2056214-2056214 on Build GRCh38
rs45517117 Chr.16: 2056195-2056195 on Build GRCh38
rs761112481 Chr.16: 2056136-2056137 on Build GRCh38
rs761818728 Chr.16: 2056142-2056142 on Build GRCh38
rs779176291 Chr.16: 2056191-2056191 on Build GRCh38
rs947846643 Chr.16: 2056399-2056399 on Build GRCh38
rs45468592 Chr.16: 2056194-2056194 on Build GRCh38
rs773867705 Chr.16: 2056380-2056380 on Build GRCh38
rs1030822016 Chr.16: 2056326-2056326 on Build GRCh38
rs374410454 Chr.16: 2056218-2056218 on Build GRCh38
rs376590558 Chr.16: 2056308-2056308 on Build GRCh38
rs77281500 Chr.16: 2056470-2056470 on Build GRCh38
rs397515057 Chr.16: 2056160-2056160 on Build GRCh38
rs137854205 Chr.16: 2056229-2056229 on Build GRCh38
rs199683502 Chr.16: 2056178-2056178 on Build GRCh38
rs749498694 Chr.16: 2056133-2056133 on Build GRCh38
rs145549047 Chr.16: 2056533-2056533 on Build GRCh38
rs45488893 Chr.16: 2056245-2056245 on Build GRCh38
rs768869777 Chr.16: 2056138-2056138 on Build GRCh38
rs763726222 Chr.16: 2056238-2056238 on Build GRCh38
rs976201466 Chr.16: 2056192-2056192 on Build GRCh38
rs572754464 Chr.16: 2056434-2056435 on Build GRCh38
rs371356151 Chr.16: 2056270-2056270 on Build GRCh38
rs1023796947 Chr.16: 2056474-2056474 on Build GRCh38
rs1022599845 Chr.16: 2056175-2056175 on Build GRCh38
rs1030958826 Chr.16: 2056549-2056549 on Build GRCh38
rs187353366 Chr.16: 2056446-2056446 on Build GRCh38
rs1018346033 Chr.16: 2056275-2056275 on Build GRCh38
rs771610974 Chr.16: 2056418-2056418 on Build GRCh38
rs891284962 Chr.16: 2056471-2056471 on Build GRCh38

View Full Product Details