Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2091603-2092134 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC105371049
Entrez Gene ID
105371049
Gene Name
uncharacterized LOC105371049
Gene Aliases
-
UniGene
Hs.710709
Gene Chromosome Location
Chr.16: 2091436-2095017 on Build GRCh38
Transcripts
NR_135175.1 NM_000296.3 NM_001009944.2 XM_005255370.3 XM_011522528.2 XM_011522529.2 XM_011522530.2 XM_011522532.2 XM_011522534.2 XM_011522535.2 XM_011522537.2 XM_017023285.1

Gene Symbol
PKD1
Entrez Gene ID
5310
Gene Name
polycystin 1, transient receptor potential channel interacting
Gene Aliases
PBP, Pc-1, TRPP1
UniGene
Hs.75813
Gene Chromosome Location
Chr.16: 2088708-2135898 on Build GRCh38
Transcripts
NR_135175.1 NM_000296.3 NM_001009944.2 XM_005255370.3 XM_011522528.2 XM_011522529.2 XM_011522530.2 XM_011522532.2 XM_011522534.2 XM_011522535.2 XM_011522537.2 XM_017023285.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5758646 c.11493C>G p.R3831R Chr.16: 2091825-2091825 on Build GRCh38
3741834 c.11410G>A p.G3804R Chr.16: 2092048-2092048 on Build GRCh38
6896387 c.11458G>A p.V3820M Chr.16: 2091860-2091860 on Build GRCh38
5600215 c.11375C>T p.S3792L Chr.16: 2092083-2092083 on Build GRCh38
3277689 c.11504G>A p.R3835H Chr.16: 2091814-2091814 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756308252 Chr.16: 2092054-2092054 on Build GRCh38
rs927594751 Chr.16: 2091707-2091708 on Build GRCh38
rs373922889 Chr.16: 2091911-2091911 on Build GRCh38
rs1020513361 Chr.16: 2091801-2091801 on Build GRCh38
rs759323548 Chr.16: 2091980-2091980 on Build GRCh38
rs971553671 Chr.16: 2091690-2091690 on Build GRCh38
rs532307379 Chr.16: 2091770-2091770 on Build GRCh38
rs543197516 Chr.16: 2092031-2092031 on Build GRCh38
rs760723063 Chr.16: 2092075-2092076 on Build GRCh38
rs142455071 Chr.16: 2092107-2092107 on Build GRCh38
rs756149323 Chr.16: 2091955-2091955 on Build GRCh38
rs961699986 Chr.16: 2091661-2091661 on Build GRCh38
rs759434553 Chr.16: 2091745-2091745 on Build GRCh38
rs767032565 Chr.16: 2091858-2091858 on Build GRCh38
rs772916096 Chr.16: 2091937-2091937 on Build GRCh38
rs543715117 Chr.16: 2091656-2091656 on Build GRCh38
rs968259191 Chr.16: 2092077-2092077 on Build GRCh38
rs766462565 Chr.16: 2091743-2091743 on Build GRCh38
rs199571695 Chr.16: 2091761-2091761 on Build GRCh38
rs780113152 Chr.16: 2092058-2092058 on Build GRCh38
rs773002078 Chr.16: 2091944-2091960 on Build GRCh38
rs769371299 Chr.16: 2092093-2092093 on Build GRCh38
rs143436918 Chr.16: 2092088-2092088 on Build GRCh38
rs904985369 Chr.16: 2091968-2091968 on Build GRCh38
rs761870700 Chr.16: 2091635-2091635 on Build GRCh38
rs577599260 Chr.16: 2091905-2091905 on Build GRCh38
rs773672893 Chr.16: 2092097-2092097 on Build GRCh38
rs748007143 Chr.16: 2092005-2092005 on Build GRCh38
rs765732726 Chr.16: 2091870-2091870 on Build GRCh38
rs1001917670 Chr.16: 2091969-2091969 on Build GRCh38
rs553377243 Chr.16: 2091913-2091913 on Build GRCh38
rs769902705 Chr.16: 2091918-2091918 on Build GRCh38
rs893313827 Chr.16: 2091978-2091978 on Build GRCh38
rs746396351 Chr.16: 2091625-2091625 on Build GRCh38
rs562214956 Chr.16: 2091986-2091986 on Build GRCh38
rs770815472 Chr.16: 2092014-2092027 on Build GRCh38
rs774643617 Chr.16: 2091634-2091634 on Build GRCh38
rs745427767 Chr.16: 2092089-2092089 on Build GRCh38
rs752010352 Chr.16: 2091948-2091948 on Build GRCh38
rs35327793 Chr.16: 2091679-2091679 on Build GRCh38
rs778795540 Chr.16: 2092003-2092003 on Build GRCh38
rs747172136 Chr.16: 2091821-2091821 on Build GRCh38
rs146650596 Chr.16: 2091796-2091796 on Build GRCh38
rs577656201 Chr.16: 2091671-2091671 on Build GRCh38
rs199476098 Chr.16: 2091861-2091861 on Build GRCh38
rs1050362263 Chr.16: 2091651-2091651 on Build GRCh38
rs753824647 Chr.16: 2091744-2091744 on Build GRCh38
rs749246687 Chr.16: 2091954-2091954 on Build GRCh38
rs748504204 Chr.16: 2091815-2091815 on Build GRCh38
rs760310609 Chr.16: 2092016-2092016 on Build GRCh38
rs765112219 Chr.16: 2091784-2091784 on Build GRCh38
rs772756642 Chr.16: 2092012-2092012 on Build GRCh38
rs548274621 Chr.16: 2091837-2091837 on Build GRCh38
rs987743414 Chr.16: 2091709-2091709 on Build GRCh38
rs201509188 Chr.16: 2092082-2092082 on Build GRCh38
rs113369380 Chr.16: 2091795-2091795 on Build GRCh38
rs573205764 Chr.16: 2091933-2091933 on Build GRCh38
rs758832889 Chr.16: 2091819-2091819 on Build GRCh38
rs768886583 Chr.16: 2091633-2091633 on Build GRCh38
rs762777473 Chr.16: 2091789-2091789 on Build GRCh38
rs775002585 Chr.16: 2092094-2092094 on Build GRCh38
rs986637119 Chr.16: 2092030-2092030 on Build GRCh38
rs770667157 Chr.16: 2092083-2092083 on Build GRCh38
rs2854584 Chr.16: 2091892-2091892 on Build GRCh38
rs199952780 Chr.16: 2091983-2091983 on Build GRCh38
rs1036702689 Chr.16: 2091917-2091917 on Build GRCh38
rs1005925235 Chr.16: 2091653-2091653 on Build GRCh38
rs541920017 Chr.16: 2091957-2091957 on Build GRCh38
rs775196410 Chr.16: 2091786-2091786 on Build GRCh38
rs371394349 Chr.16: 2092062-2092062 on Build GRCh38
rs943538758 Chr.16: 2091711-2091711 on Build GRCh38
rs200120839 Chr.16: 2092111-2092111 on Build GRCh38
rs917172938 Chr.16: 2091996-2091996 on Build GRCh38
rs1057518959 Chr.16: 2091794-2091794 on Build GRCh38
rs201204878 Chr.16: 2091775-2091776 on Build GRCh38
rs528557867 Chr.16: 2091715-2091715 on Build GRCh38
rs758307192 Chr.16: 2092114-2092114 on Build GRCh38
rs912121307 Chr.16: 2091710-2091710 on Build GRCh38
rs201371972 Chr.16: 2091882-2091882 on Build GRCh38
rs371331679 Chr.16: 2091629-2091629 on Build GRCh38
rs763514263 Chr.16: 2091942-2091942 on Build GRCh38
rs781325135 Chr.16: 2091950-2091950 on Build GRCh38
rs772228852 Chr.16: 2091644-2091644 on Build GRCh38
rs546212886 Chr.16: 2091684-2091684 on Build GRCh38
rs749953469 Chr.16: 2091860-2091860 on Build GRCh38
rs142125098 Chr.16: 2091833-2091833 on Build GRCh38
rs199476096 Chr.16: 2091806-2091806 on Build GRCh38
rs980229060 Chr.16: 2091703-2091703 on Build GRCh38
rs1046045524 Chr.16: 2091951-2091951 on Build GRCh38
rs979431713 Chr.16: 2092078-2092078 on Build GRCh38
rs897735779 Chr.16: 2091920-2091920 on Build GRCh38
rs542332982 Chr.16: 2091717-2091717 on Build GRCh38
rs559823238 Chr.16: 2091693-2091693 on Build GRCh38
rs760818311 Chr.16: 2091739-2091739 on Build GRCh38
rs754048939 Chr.16: 2092109-2092109 on Build GRCh38
rs370455159 Chr.16: 2091841-2091841 on Build GRCh38
rs991680191 Chr.16: 2091625-2091636 on Build GRCh38
rs1032052352 Chr.16: 2091977-2091977 on Build GRCh38
rs1056351922 Chr.16: 2091961-2091961 on Build GRCh38
rs149331148 Chr.16: 2092070-2092070 on Build GRCh38
rs201660914 Chr.16: 2091764-2091764 on Build GRCh38
rs772236459 Chr.16: 2091766-2091766 on Build GRCh38
rs752399775 Chr.16: 2091750-2091750 on Build GRCh38
rs768468097 Chr.16: 2091924-2091924 on Build GRCh38
rs138629093 Chr.16: 2092068-2092068 on Build GRCh38
rs762337976 Chr.16: 2092095-2092095 on Build GRCh38
rs750639082 Chr.16: 2092044-2092044 on Build GRCh38
rs368634612 Chr.16: 2092020-2092020 on Build GRCh38
rs755622553 Chr.16: 2091867-2091867 on Build GRCh38
rs950357849 Chr.16: 2091703-2091703 on Build GRCh38
rs751107765 Chr.16: 2091754-2091754 on Build GRCh38
rs770333312 Chr.16: 2091627-2091627 on Build GRCh38
rs757941153 Chr.16: 2091623-2091623 on Build GRCh38
rs114241458 Chr.16: 2092064-2092064 on Build GRCh38
rs534949676 Chr.16: 2091855-2091855 on Build GRCh38
rs557687541 Chr.16: 2091665-2091665 on Build GRCh38
rs544060015 Chr.16: 2092035-2092036 on Build GRCh38
rs780593395 Chr.16: 2091755-2091755 on Build GRCh38
rs754396996 Chr.16: 2091805-2091805 on Build GRCh38
rs367775096 Chr.16: 2091912-2091912 on Build GRCh38
rs768625783 Chr.16: 2091997-2091997 on Build GRCh38
rs751197845 Chr.16: 2091791-2091791 on Build GRCh38
rs990201940 Chr.16: 2091723-2091723 on Build GRCh38
rs777080940 Chr.16: 2091624-2091624 on Build GRCh38
rs761577582 Chr.16: 2091929-2091929 on Build GRCh38
rs764884125 Chr.16: 2091746-2091746 on Build GRCh38
rs374554930 Chr.16: 2091949-2091949 on Build GRCh38
rs183078730 Chr.16: 2092029-2092029 on Build GRCh38
rs773379794 Chr.16: 2091734-2091734 on Build GRCh38
rs771139859 Chr.16: 2091826-2091826 on Build GRCh38
rs779457101 Chr.16: 2091810-2091810 on Build GRCh38
rs151157369 Chr.16: 2091845-2091845 on Build GRCh38
rs780505754 Chr.16: 2091628-2091628 on Build GRCh38
rs769987725 Chr.16: 2092039-2092039 on Build GRCh38
rs949081660 Chr.16: 2091938-2091938 on Build GRCh38
rs771889701 Chr.16: 2092008-2092008 on Build GRCh38
rs560886704 Chr.16: 2091728-2091728 on Build GRCh38
rs36221082 Chr.16: 2091820-2091820 on Build GRCh38
rs770380793 Chr.16: 2092017-2092017 on Build GRCh38
rs773539182 Chr.16: 2091767-2091767 on Build GRCh38
rs764576916 Chr.16: 2091945-2091945 on Build GRCh38
rs763815841 Chr.16: 2091843-2091843 on Build GRCh38
rs777380909 Chr.16: 2091753-2091753 on Build GRCh38
rs1012723566 Chr.16: 2091646-2091646 on Build GRCh38
rs969094441 Chr.16: 2091774-2091774 on Build GRCh38
rs757938764 Chr.16: 2091751-2091751 on Build GRCh38
rs557630487 Chr.16: 2091904-2091904 on Build GRCh38
rs762280267 Chr.16: 2092035-2092035 on Build GRCh38
rs974900344 Chr.16: 2091720-2091720 on Build GRCh38
rs919217705 Chr.16: 2092063-2092063 on Build GRCh38
rs764653613 Chr.16: 2091987-2091987 on Build GRCh38
rs377486837 Chr.16: 2091668-2091668 on Build GRCh38
rs761411408 Chr.16: 2091851-2091851 on Build GRCh38
rs755394153 Chr.16: 2091809-2091809 on Build GRCh38
rs551733361 Chr.16: 2091776-2091776 on Build GRCh38
rs749154519 Chr.16: 2091923-2091923 on Build GRCh38
rs963119551 Chr.16: 2091655-2091655 on Build GRCh38
rs753956366 Chr.16: 2092061-2092061 on Build GRCh38
rs750599264 Chr.16: 2091954-2091954 on Build GRCh38
rs374264641 Chr.16: 2091760-2091760 on Build GRCh38
rs776813165 Chr.16: 2091828-2091828 on Build GRCh38
rs758847545 Chr.16: 2091885-2091885 on Build GRCh38
rs147992238 Chr.16: 2092102-2092102 on Build GRCh38
rs777585821 Chr.16: 2092076-2092076 on Build GRCh38
rs899020508 Chr.16: 2092027-2092027 on Build GRCh38
rs145955373 Chr.16: 2092112-2092112 on Build GRCh38
rs774079655 Chr.16: 2091925-2091925 on Build GRCh38
rs752098720 Chr.16: 2092032-2092032 on Build GRCh38
rs371176550 Chr.16: 2091939-2091939 on Build GRCh38
rs749734814 Chr.16: 2091757-2091757 on Build GRCh38
rs1035553327 Chr.16: 2091626-2091626 on Build GRCh38
rs1023153667 Chr.16: 2091649-2091649 on Build GRCh38
rs112276855 Chr.16: 2091919-2091919 on Build GRCh38

View Full Product Details