Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3028610-3029105 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BICDL2
Entrez Gene ID
146439
Gene Name
BICD family like cargo adaptor 2
Gene Aliases
BICDR-2, BICDR2, CCDC64B
UniGene
Hs.513285
Gene Chromosome Location
Chr.16: 3027710-3036985 on Build GRCh38
Transcripts
NM_001103175.1 XM_005255135.4 XM_011522391.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5725845 c.1187C>T p.P396L Chr.16: 3028751-3028751 on Build GRCh38
6622336 c.1127A>C p.E376A Chr.16: 3028811-3028811 on Build GRCh38
6046167 c.1175C>T p.A392V Chr.16: 3028763-3028763 on Build GRCh38
4669986 c.1152_1153delGC p.Q385fs*118 Chr.16: 3028785-3028786 on Build GRCh38
3988432 c.1238A>T p.K413M Chr.16: 3028700-3028700 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770465946 Chr.16: 3028672-3028672 on Build GRCh38
rs371238238 Chr.16: 3028718-3028718 on Build GRCh38
rs940649943 Chr.16: 3028655-3028655 on Build GRCh38
rs976212904 Chr.16: 3028774-3028774 on Build GRCh38
rs766451998 Chr.16: 3028746-3028746 on Build GRCh38
rs377372496 Chr.16: 3028796-3028796 on Build GRCh38
rs1000613828 Chr.16: 3028751-3028751 on Build GRCh38
rs1053546057 Chr.16: 3029002-3029002 on Build GRCh38
rs761812364 Chr.16: 3028728-3028728 on Build GRCh38
rs1046076913 Chr.16: 3029043-3029043 on Build GRCh38
rs745574935 Chr.16: 3028689-3028689 on Build GRCh38
rs985678317 Chr.16: 3028884-3028884 on Build GRCh38
rs950452490 Chr.16: 3028931-3028931 on Build GRCh38
rs750518137 Chr.16: 3028738-3028738 on Build GRCh38
rs180745269 Chr.16: 3029038-3029038 on Build GRCh38
rs530468745 Chr.16: 3028897-3028897 on Build GRCh38
rs755255015 Chr.16: 3028762-3028762 on Build GRCh38
rs761990153 Chr.16: 3029056-3029056 on Build GRCh38
rs918931041 Chr.16: 3028782-3028782 on Build GRCh38
rs754038336 Chr.16: 3028898-3028898 on Build GRCh38
rs909205611 Chr.16: 3028644-3028644 on Build GRCh38
rs902575579 Chr.16: 3029021-3029021 on Build GRCh38
rs751975231 Chr.16: 3028758-3028758 on Build GRCh38
rs986299032 Chr.16: 3028959-3028959 on Build GRCh38
rs528171556 Chr.16: 3028739-3028739 on Build GRCh38
rs1032974828 Chr.16: 3028707-3028707 on Build GRCh38
rs566987864 Chr.16: 3028979-3028979 on Build GRCh38
rs962084454 Chr.16: 3028899-3028899 on Build GRCh38
rs762685630 Chr.16: 3028947-3028947 on Build GRCh38
rs778368485 Chr.16: 3028679-3028679 on Build GRCh38
rs1020968298 Chr.16: 3028669-3028669 on Build GRCh38
rs775104505 Chr.16: 3028883-3028883 on Build GRCh38
rs536064557 Chr.16: 3029000-3029000 on Build GRCh38
rs143969469 Chr.16: 3028943-3028943 on Build GRCh38
rs1002113131 Chr.16: 3029086-3029086 on Build GRCh38
rs775363969 Chr.16: 3028705-3028705 on Build GRCh38
rs1024908453 Chr.16: 3028811-3028811 on Build GRCh38
rs892131385 Chr.16: 3028769-3028769 on Build GRCh38
rs748155113 Chr.16: 3028853-3028853 on Build GRCh38
rs544162173 Chr.16: 3029077-3029077 on Build GRCh38
rs769787589 Chr.16: 3028857-3028857 on Build GRCh38
rs745310796 Chr.16: 3028824-3028824 on Build GRCh38
rs984845333 Chr.16: 3028941-3028941 on Build GRCh38
rs376295108 Chr.16: 3028763-3028763 on Build GRCh38
rs975863725 Chr.16: 3028915-3028916 on Build GRCh38
rs199578448 Chr.16: 3028771-3028771 on Build GRCh38
rs773438871 Chr.16: 3028861-3028861 on Build GRCh38
rs762890895 Chr.16: 3028868-3028868 on Build GRCh38
rs557300600 Chr.16: 3028975-3028975 on Build GRCh38
rs1013690221 Chr.16: 3028842-3028842 on Build GRCh38
rs868651357 Chr.16: 3028797-3028797 on Build GRCh38
rs370499011 Chr.16: 3028837-3028837 on Build GRCh38
rs559310839 Chr.16: 3028680-3028680 on Build GRCh38
rs112741342 Chr.16: 3028915-3028915 on Build GRCh38
rs755520651 Chr.16: 3028652-3028652 on Build GRCh38
rs954261703 Chr.16: 3028876-3028876 on Build GRCh38
rs553583417 Chr.16: 3028974-3028974 on Build GRCh38
rs148635155 Chr.16: 3028957-3028957 on Build GRCh38
rs1054943802 Chr.16: 3028977-3028977 on Build GRCh38
rs373891310 Chr.16: 3028845-3028845 on Build GRCh38
rs902521240 Chr.16: 3028850-3028850 on Build GRCh38
rs1039841744 Chr.16: 3028797-3028817 on Build GRCh38
rs200160527 Chr.16: 3028702-3028702 on Build GRCh38
rs2011561 Chr.16: 3029011-3029011 on Build GRCh38
rs923496415 Chr.16: 3028662-3028662 on Build GRCh38
rs753248638 Chr.16: 3028791-3028791 on Build GRCh38
rs56337734 Chr.16: 3028638-3028638 on Build GRCh38
rs374689387 Chr.16: 3028902-3028902 on Build GRCh38
rs765052854 Chr.16: 3028852-3028852 on Build GRCh38
rs896295018 Chr.16: 3028841-3028841 on Build GRCh38
rs547942362 Chr.16: 3028848-3028848 on Build GRCh38
rs1032636080 Chr.16: 3028875-3028875 on Build GRCh38
rs748824558 Chr.16: 3028658-3028658 on Build GRCh38
rs967090768 Chr.16: 3028685-3028685 on Build GRCh38
rs545588415 Chr.16: 3028656-3028656 on Build GRCh38
rs771772879 Chr.16: 3028826-3028826 on Build GRCh38
rs571693095 Chr.16: 3028640-3028640 on Build GRCh38
rs768940131 Chr.16: 3028686-3028687 on Build GRCh38
rs568169608 Chr.16: 3028891-3028891 on Build GRCh38
rs778464848 Chr.16: 3028804-3028804 on Build GRCh38
rs537537259 Chr.16: 3028914-3028914 on Build GRCh38
rs1049004892 Chr.16: 3028810-3028810 on Build GRCh38
rs937943776 Chr.16: 3028973-3028973 on Build GRCh38
rs776672376 Chr.16: 3028724-3028724 on Build GRCh38
rs765457492 Chr.16: 3028735-3028735 on Build GRCh38
rs965888623 Chr.16: 3028904-3028904 on Build GRCh38
rs760332221 Chr.16: 3028710-3028710 on Build GRCh38
rs906477402 Chr.16: 3029080-3029080 on Build GRCh38
rs933510446 Chr.16: 3028664-3028664 on Build GRCh38

View Full Product Details