Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3204160-3204660 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR1F1
Entrez Gene ID
4992
Gene Name
olfactory receptor family 1 subfamily F member 1
Gene Aliases
OLFMF, OR16-36, OR16-37, OR16-88, OR16-89, OR16-90, OR1F10, OR1F13P, OR1F4, OR1F5, OR1F6, OR1F7, OR1F8, OR1F9, OR3-145, ORL1023
UniGene
Hs.664502
Gene Chromosome Location
Chr.16: 3188195-3206559 on Build GRCh38
Transcripts
NM_012360.1 XM_011522506.2 XM_011522507.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5971787 c.160T>C p.C54R Chr.16: 3204406-3204406 on Build GRCh38
1708656 c.221C>A p.S74Y Chr.16: 3204467-3204467 on Build GRCh38
5951179 c.196C>A p.L66M Chr.16: 3204442-3204442 on Build GRCh38
2918853 c.72G>T p.Q24H Chr.16: 3204318-3204318 on Build GRCh38
4568986 c.142T>G p.S48A Chr.16: 3204388-3204388 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771074425 Chr.16: 3204224-3204224 on Build GRCh38
rs780153971 Chr.16: 3204347-3204347 on Build GRCh38
rs200488860 Chr.16: 3204406-3204406 on Build GRCh38
rs371507036 Chr.16: 3204479-3204479 on Build GRCh38
rs753756127 Chr.16: 3204330-3204330 on Build GRCh38
rs186277679 Chr.16: 3204208-3204208 on Build GRCh38
rs778446982 Chr.16: 3204472-3204472 on Build GRCh38
rs372744779 Chr.16: 3204469-3204471 on Build GRCh38
rs574208759 Chr.16: 3204570-3204570 on Build GRCh38
rs149217288 Chr.16: 3204442-3204442 on Build GRCh38
rs371736211 Chr.16: 3204523-3204523 on Build GRCh38
rs761713816 Chr.16: 3204430-3204430 on Build GRCh38
rs749329943 Chr.16: 3204216-3204216 on Build GRCh38
rs868134628 Chr.16: 3204631-3204631 on Build GRCh38
rs765112481 Chr.16: 3204432-3204432 on Build GRCh38
rs781224614 Chr.16: 3204407-3204407 on Build GRCh38
rs749231346 Chr.16: 3204263-3204263 on Build GRCh38
rs759679524 Chr.16: 3204533-3204533 on Build GRCh38
rs1045433609 Chr.16: 3204225-3204225 on Build GRCh38
rs758873218 Chr.16: 3204586-3204586 on Build GRCh38
rs747242257 Chr.16: 3204349-3204349 on Build GRCh38
rs763843633 Chr.16: 3204385-3204385 on Build GRCh38
rs772025480 Chr.16: 3204621-3204621 on Build GRCh38
rs61731435 Chr.16: 3204566-3204566 on Build GRCh38
rs753687732 Chr.16: 3204387-3204387 on Build GRCh38
rs752952518 Chr.16: 3204536-3204536 on Build GRCh38
rs752460659 Chr.16: 3204257-3204257 on Build GRCh38
rs939954018 Chr.16: 3204202-3204202 on Build GRCh38
rs190411580 Chr.16: 3204226-3204226 on Build GRCh38
rs563043868 Chr.16: 3204605-3204605 on Build GRCh38
rs767631172 Chr.16: 3204575-3204575 on Build GRCh38
rs199866050 Chr.16: 3204458-3204458 on Build GRCh38
rs746532811 Chr.16: 3204480-3204480 on Build GRCh38
rs981396532 Chr.16: 3204532-3204532 on Build GRCh38
rs770312546 Chr.16: 3204367-3204367 on Build GRCh38
rs369884019 Chr.16: 3204569-3204569 on Build GRCh38
rs529350112 Chr.16: 3204185-3204185 on Build GRCh38
rs147872466 Chr.16: 3204465-3204465 on Build GRCh38
rs531758693 Chr.16: 3204403-3204403 on Build GRCh38
rs778899325 Chr.16: 3204310-3204312 on Build GRCh38
rs777586180 Chr.16: 3204609-3204609 on Build GRCh38
rs371331484 Chr.16: 3204422-3204422 on Build GRCh38
rs768707730 Chr.16: 3204625-3204625 on Build GRCh38
rs61731433 Chr.16: 3204424-3204424 on Build GRCh38
rs183120097 Chr.16: 3204416-3204416 on Build GRCh38
rs376639452 Chr.16: 3204502-3204502 on Build GRCh38
rs148405876 Chr.16: 3204280-3204280 on Build GRCh38
rs747854183 Chr.16: 3204563-3204563 on Build GRCh38
rs988700511 Chr.16: 3204363-3204363 on Build GRCh38
rs759357058 Chr.16: 3204574-3204574 on Build GRCh38
rs771490781 Chr.16: 3204478-3204478 on Build GRCh38
rs368624412 Chr.16: 3204467-3204467 on Build GRCh38
rs749787550 Chr.16: 3204417-3204417 on Build GRCh38
rs758484506 Chr.16: 3204342-3204342 on Build GRCh38
rs767191912 Chr.16: 3204291-3204291 on Build GRCh38
rs142486394 Chr.16: 3204293-3204293 on Build GRCh38
rs370072281 Chr.16: 3204227-3204227 on Build GRCh38
rs754976995 Chr.16: 3204449-3204449 on Build GRCh38
rs772604030 Chr.16: 3204230-3204230 on Build GRCh38
rs760341370 Chr.16: 3204382-3204382 on Build GRCh38
rs1034235190 Chr.16: 3204545-3204545 on Build GRCh38
rs764249432 Chr.16: 3204544-3204544 on Build GRCh38
rs57224133 Chr.16: 3204245-3204245 on Build GRCh38
rs1014992426 Chr.16: 3204473-3204473 on Build GRCh38
rs748882517 Chr.16: 3204610-3204610 on Build GRCh38
rs778944660 Chr.16: 3204340-3204340 on Build GRCh38
rs370564788 Chr.16: 3204391-3204391 on Build GRCh38
rs143244133 Chr.16: 3204399-3204399 on Build GRCh38
rs773673344 Chr.16: 3204371-3204371 on Build GRCh38
rs1051334326 Chr.16: 3204184-3204184 on Build GRCh38
rs776484197 Chr.16: 3204497-3204497 on Build GRCh38
rs928316000 Chr.16: 3204499-3204499 on Build GRCh38
rs533281144 Chr.16: 3204618-3204618 on Build GRCh38
rs753890209 Chr.16: 3204211-3204214 on Build GRCh38
rs926962808 Chr.16: 3204397-3204397 on Build GRCh38
rs755868658 Chr.16: 3204261-3204261 on Build GRCh38
rs150333046 Chr.16: 3204508-3204508 on Build GRCh38
rs140644764 Chr.16: 3204408-3204408 on Build GRCh38
rs530728442 Chr.16: 3204233-3204233 on Build GRCh38
rs183463120 Chr.16: 3204606-3204606 on Build GRCh38
rs144582974 Chr.16: 3204267-3204267 on Build GRCh38
rs777026559 Chr.16: 3204360-3204360 on Build GRCh38
rs8045183 Chr.16: 3204622-3204622 on Build GRCh38
rs375366387 Chr.16: 3204567-3204567 on Build GRCh38
rs778178508 Chr.16: 3204588-3204588 on Build GRCh38
rs141236935 Chr.16: 3204556-3204556 on Build GRCh38
rs1834026 Chr.16: 3204470-3204470 on Build GRCh38
rs143258202 Chr.16: 3204444-3204444 on Build GRCh38
rs138114408 Chr.16: 3204579-3204579 on Build GRCh38
rs201584157 Chr.16: 3204285-3204285 on Build GRCh38
rs758952819 Chr.16: 3204598-3204598 on Build GRCh38
rs754597516 Chr.16: 3204255-3204255 on Build GRCh38
rs766373491 Chr.16: 3204514-3204514 on Build GRCh38
rs368384694 Chr.16: 3204421-3204421 on Build GRCh38
rs201720218 Chr.16: 3204568-3204568 on Build GRCh38
rs765515578 Chr.16: 3204593-3204593 on Build GRCh38
rs147055076 Chr.16: 3204505-3204505 on Build GRCh38
rs767992959 Chr.16: 3204490-3204490 on Build GRCh38
rs372434861 Chr.16: 3204591-3204591 on Build GRCh38
rs138845795 Chr.16: 3204549-3204549 on Build GRCh38
rs1006941659 Chr.16: 3204312-3204312 on Build GRCh38
rs566580978 Chr.16: 3204276-3204276 on Build GRCh38
rs749840047 Chr.16: 3204477-3204477 on Build GRCh38
rs186786299 Chr.16: 3204381-3204381 on Build GRCh38
rs371443638 Chr.16: 3204468-3204470 on Build GRCh38
rs889790683 Chr.16: 3204485-3204485 on Build GRCh38
rs557293364 Chr.16: 3204537-3204537 on Build GRCh38
rs897445735 Chr.16: 3204283-3204283 on Build GRCh38
rs947561508 Chr.16: 3204522-3204522 on Build GRCh38
rs996860621 Chr.16: 3204551-3204551 on Build GRCh38
rs757475700 Chr.16: 3204264-3204264 on Build GRCh38
rs762383812 Chr.16: 3204248-3204248 on Build GRCh38
rs146853431 Chr.16: 3204404-3204404 on Build GRCh38
rs181720197 Chr.16: 3204290-3204290 on Build GRCh38
rs775739946 Chr.16: 3204270-3204270 on Build GRCh38
rs540439560 Chr.16: 3204310-3204310 on Build GRCh38
rs267604532 Chr.16: 3204555-3204555 on Build GRCh38
rs560268759 Chr.16: 3204319-3204319 on Build GRCh38
rs546950482 Chr.16: 3204619-3204619 on Build GRCh38
rs200138081 Chr.16: 3204252-3204252 on Build GRCh38
rs145939751 Chr.16: 3204361-3204361 on Build GRCh38
rs185165521 Chr.16: 3204447-3204447 on Build GRCh38
rs141199102 Chr.16: 3204286-3204286 on Build GRCh38
rs376104596 Chr.16: 3204453-3204453 on Build GRCh38
rs780965320 Chr.16: 3204207-3204207 on Build GRCh38
rs781485448 Chr.16: 3204456-3204456 on Build GRCh38
rs757102358 Chr.16: 3204261-3204274 on Build GRCh38
rs574070012 Chr.16: 3204372-3204372 on Build GRCh38
rs768970238 Chr.16: 3204352-3204352 on Build GRCh38
rs201608761 Chr.16: 3204266-3204266 on Build GRCh38
rs61731440 Chr.16: 3204611-3204611 on Build GRCh38
rs769008678 Chr.16: 3204277-3204277 on Build GRCh38
rs60611456 Chr.16: 3204235-3204235 on Build GRCh38
rs545429025 Chr.16: 3204608-3204608 on Build GRCh38
rs61731432 Chr.16: 3204418-3204418 on Build GRCh38
rs113788191 Chr.16: 3204390-3204390 on Build GRCh38
rs766421411 Chr.16: 3204445-3204445 on Build GRCh38
rs61731434 Chr.16: 3204428-3204428 on Build GRCh38
rs983528249 Chr.16: 3204355-3204355 on Build GRCh38
rs758609972 Chr.16: 3204400-3204400 on Build GRCh38
rs780659543 Chr.16: 3204599-3204599 on Build GRCh38
rs750980288 Chr.16: 3204595-3204595 on Build GRCh38
rs748653177 Chr.16: 3204548-3204548 on Build GRCh38
rs777720597 Chr.16: 3204213-3204213 on Build GRCh38
rs898187200 Chr.16: 3204222-3204222 on Build GRCh38
rs751283450 Chr.16: 3204253-3204253 on Build GRCh38
rs757237356 Chr.16: 3204336-3204336 on Build GRCh38

View Full Product Details