Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 8892403-8892906 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USP7
Entrez Gene ID
7874
Gene Name
ubiquitin specific peptidase 7 (herpes virus-associated)
Gene Aliases
HAUSP, TEF1
UniGene
Hs.386939
Gene Chromosome Location
Chr.16: 8892094-8963912 on Build GRCh38
Transcripts
NM_001286457.1 NM_001286458.1 NM_001321858.1 NM_003470.2 NR_135826.1 XM_017023652.1 XM_017023653.1

SNP Detail

SNP ID Chromosome Location
rs530368668 Chr.16: 8892657-8892657 on Build GRCh38
rs755848678 Chr.16: 8892474-8892474 on Build GRCh38
rs945551502 Chr.16: 8892599-8892600 on Build GRCh38
rs916559128 Chr.16: 8892759-8892759 on Build GRCh38
rs982739579 Chr.16: 8892774-8892774 on Build GRCh38
rs764781517 Chr.16: 8892767-8892767 on Build GRCh38
rs779390981 Chr.16: 8892608-8892613 on Build GRCh38
rs998486609 Chr.16: 8892694-8892694 on Build GRCh38
rs560345318 Chr.16: 8892688-8892688 on Build GRCh38
rs553993437 Chr.16: 8892833-8892833 on Build GRCh38
rs553728127 Chr.16: 8892452-8892452 on Build GRCh38
rs572284667 Chr.16: 8892844-8892844 on Build GRCh38
rs139676650 Chr.16: 8892879-8892879 on Build GRCh38
rs557444294 Chr.16: 8892707-8892707 on Build GRCh38
rs190047926 Chr.16: 8892434-8892434 on Build GRCh38
rs138918649 Chr.16: 8892719-8892719 on Build GRCh38
rs867495239 Chr.16: 8892625-8892625 on Build GRCh38
rs574504591 Chr.16: 8892433-8892433 on Build GRCh38
rs967256427 Chr.16: 8892477-8892477 on Build GRCh38
rs888184466 Chr.16: 8892815-8892815 on Build GRCh38
rs55937310 Chr.16: 8892625-8892629 on Build GRCh38
rs930134731 Chr.16: 8892789-8892789 on Build GRCh38
rs765653626 Chr.16: 8892606-8892611 on Build GRCh38
rs978547955 Chr.16: 8892479-8892479 on Build GRCh38
rs954691227 Chr.16: 8892744-8892744 on Build GRCh38
rs372119271 Chr.16: 8892734-8892734 on Build GRCh38
rs918882040 Chr.16: 8892784-8892784 on Build GRCh38
rs368329036 Chr.16: 8892492-8892492 on Build GRCh38
rs76501830 Chr.16: 8892622-8892622 on Build GRCh38
rs866508568 Chr.16: 8892880-8892880 on Build GRCh38
rs776670403 Chr.16: 8892508-8892508 on Build GRCh38
rs186414269 Chr.16: 8892806-8892806 on Build GRCh38
rs906757150 Chr.16: 8892456-8892456 on Build GRCh38
rs756320326 Chr.16: 8892606-8892610 on Build GRCh38
rs916819404 Chr.16: 8892497-8892497 on Build GRCh38
rs563206498 Chr.16: 8892641-8892641 on Build GRCh38
rs544533428 Chr.16: 8892598-8892598 on Build GRCh38
rs749959927 Chr.16: 8892769-8892769 on Build GRCh38
rs576127791 Chr.16: 8892579-8892579 on Build GRCh38
rs548760375 Chr.16: 8892658-8892658 on Build GRCh38
rs930169623 Chr.16: 8892560-8892560 on Build GRCh38
rs374474942 Chr.16: 8892630-8892634 on Build GRCh38
rs536250687 Chr.16: 8892842-8892842 on Build GRCh38
rs760361068 Chr.16: 8892439-8892439 on Build GRCh38
rs748841887 Chr.16: 8892591-8892591 on Build GRCh38
rs571790874 Chr.16: 8892473-8892473 on Build GRCh38
rs905810000 Chr.16: 8892634-8892634 on Build GRCh38
rs1021645866 Chr.16: 8892451-8892451 on Build GRCh38
rs910456039 Chr.16: 8892773-8892773 on Build GRCh38
rs572149227 Chr.16: 8892460-8892460 on Build GRCh38
rs1026739879 Chr.16: 8892438-8892438 on Build GRCh38
rs535411938 Chr.16: 8892831-8892831 on Build GRCh38
rs990842209 Chr.16: 8892495-8892495 on Build GRCh38
rs888237154 Chr.16: 8892626-8892626 on Build GRCh38
rs879206430 Chr.16: 8892623-8892623 on Build GRCh38
rs943005554 Chr.16: 8892523-8892523 on Build GRCh38
rs1042527901 Chr.16: 8892618-8892618 on Build GRCh38
rs919174653 Chr.16: 8892493-8892493 on Build GRCh38
rs961449195 Chr.16: 8892729-8892729 on Build GRCh38
rs1032408525 Chr.16: 8892733-8892733 on Build GRCh38
rs74709536 Chr.16: 8892710-8892710 on Build GRCh38
rs943136581 Chr.16: 8892425-8892425 on Build GRCh38
rs564035609 Chr.16: 8892544-8892544 on Build GRCh38
rs1006547371 Chr.16: 8892448-8892448 on Build GRCh38
rs958136130 Chr.16: 8892740-8892740 on Build GRCh38
rs144861055 Chr.16: 8892750-8892750 on Build GRCh38
rs550399465 Chr.16: 8892426-8892426 on Build GRCh38
rs976600617 Chr.16: 8892465-8892465 on Build GRCh38
rs869169091 Chr.16: 8892623-8892629 on Build GRCh38
rs368811127 Chr.16: 8892607-8892607 on Build GRCh38
rs1048612787 Chr.16: 8892621-8892621 on Build GRCh38
rs978656623 Chr.16: 8892732-8892732 on Build GRCh38
rs376102750 Chr.16: 8892507-8892507 on Build GRCh38
rs1033601241 Chr.16: 8892462-8892462 on Build GRCh38
rs765934502 Chr.16: 8892845-8892845 on Build GRCh38
rs993738622 Chr.16: 8892736-8892736 on Build GRCh38
rs758915094 Chr.16: 8892607-8892613 on Build GRCh38
rs747946767 Chr.16: 8892459-8892459 on Build GRCh38
rs754211240 Chr.16: 8892609-8892613 on Build GRCh38
rs541181996 Chr.16: 8892629-8892629 on Build GRCh38
rs1006495460 Chr.16: 8892650-8892650 on Build GRCh38
rs897274720 Chr.16: 8892530-8892530 on Build GRCh38
rs900862024 Chr.16: 8892683-8892683 on Build GRCh38
rs900808229 Chr.16: 8892859-8892859 on Build GRCh38
rs1005485910 Chr.16: 8892718-8892718 on Build GRCh38
rs149429512 Chr.16: 8892742-8892742 on Build GRCh38
rs925393076 Chr.16: 8892486-8892486 on Build GRCh38
rs189249201 Chr.16: 8892853-8892853 on Build GRCh38
rs1055296226 Chr.16: 8892654-8892654 on Build GRCh38

View Full Product Details