Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 9754240-9754756 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRIN2A
Entrez Gene ID
2903
Gene Name
glutamate ionotropic receptor NMDA type subunit 2A
Gene Aliases
EPND, FESD, GluN2A, LKS, NMDAR2A, NR2A
UniGene
Hs.411472
Gene Chromosome Location
Chr.16: 9753404-10183364 on Build GRCh38
Transcripts
NM_000833.4 NM_001134407.2 NM_001134408.2 XM_011522458.2 XM_011522461.2 XM_017023172.1 XM_017023173.1

SNP Detail

SNP ID Chromosome Location
rs892894248 Chr.16: 9754530-9754530 on Build GRCh38
rs149882531 Chr.16: 9754402-9754402 on Build GRCh38
rs745750621 Chr.16: 9754324-9754324 on Build GRCh38
rs886052514 Chr.16: 9754368-9754368 on Build GRCh38
rs768344069 Chr.16: 9754613-9754613 on Build GRCh38
rs534681142 Chr.16: 9754659-9754659 on Build GRCh38
rs963650718 Chr.16: 9754517-9754517 on Build GRCh38
rs778144489 Chr.16: 9754318-9754318 on Build GRCh38
rs942785597 Chr.16: 9754355-9754355 on Build GRCh38
rs955278370 Chr.16: 9754271-9754271 on Build GRCh38
rs1010033829 Chr.16: 9754531-9754531 on Build GRCh38
rs951467726 Chr.16: 9754637-9754637 on Build GRCh38
rs369312311 Chr.16: 9754613-9754614 on Build GRCh38
rs571467101 Chr.16: 9754435-9754435 on Build GRCh38
rs947301893 Chr.16: 9754548-9754548 on Build GRCh38
rs886052516 Chr.16: 9754607-9754607 on Build GRCh38
rs552102022 Chr.16: 9754645-9754645 on Build GRCh38
rs767748 Chr.16: 9754320-9754320 on Build GRCh38
rs771720272 Chr.16: 9754393-9754393 on Build GRCh38
rs898669746 Chr.16: 9754344-9754344 on Build GRCh38
rs767749 Chr.16: 9754614-9754614 on Build GRCh38
rs771489699 Chr.16: 9754571-9754571 on Build GRCh38
rs760313721 Chr.16: 9754540-9754540 on Build GRCh38
rs189921110 Chr.16: 9754272-9754272 on Build GRCh38
rs548986712 Chr.16: 9754608-9754608 on Build GRCh38
rs935433033 Chr.16: 9754600-9754601 on Build GRCh38
rs905551680 Chr.16: 9754452-9754452 on Build GRCh38
rs545218175 Chr.16: 9754363-9754363 on Build GRCh38
rs59371229 Chr.16: 9754612-9754613 on Build GRCh38
rs1018257126 Chr.16: 9754593-9754593 on Build GRCh38
rs1023859605 Chr.16: 9754302-9754302 on Build GRCh38
rs1033148467 Chr.16: 9754496-9754496 on Build GRCh38
rs567944117 Chr.16: 9754519-9754519 on Build GRCh38
rs974586801 Chr.16: 9754368-9754368 on Build GRCh38
rs1019714834 Chr.16: 9754477-9754477 on Build GRCh38
rs1045362202 Chr.16: 9754443-9754443 on Build GRCh38
rs898784271 Chr.16: 9754672-9754672 on Build GRCh38
rs947998688 Chr.16: 9754652-9754652 on Build GRCh38
rs886052515 Chr.16: 9754523-9754523 on Build GRCh38
rs182174522 Chr.16: 9754396-9754396 on Build GRCh38
rs557950577 Chr.16: 9754701-9754701 on Build GRCh38
rs756310832 Chr.16: 9754265-9754265 on Build GRCh38
rs1001229546 Chr.16: 9754454-9754454 on Build GRCh38
rs896701847 Chr.16: 9754419-9754419 on Build GRCh38
rs955267525 Chr.16: 9754559-9754559 on Build GRCh38
rs911246071 Chr.16: 9754332-9754332 on Build GRCh38
rs775072219 Chr.16: 9754465-9754465 on Build GRCh38
rs1047225195 Chr.16: 9754416-9754416 on Build GRCh38
rs924451768 Chr.16: 9754599-9754599 on Build GRCh38
rs537335552 Chr.16: 9754648-9754648 on Build GRCh38
rs145049184 Chr.16: 9754573-9754573 on Build GRCh38
rs995716662 Chr.16: 9754345-9754345 on Build GRCh38

View Full Product Details