Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 9754589-9755129 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRIN2A
Entrez Gene ID
2903
Gene Name
glutamate ionotropic receptor NMDA type subunit 2A
Gene Aliases
EPND, FESD, GluN2A, LKS, NMDAR2A, NR2A
UniGene
Hs.411472
Gene Chromosome Location
Chr.16: 9753404-10183364 on Build GRCh38
Transcripts
NM_000833.4 NM_001134407.2 NM_001134408.2 XM_011522458.2 XM_011522461.2 XM_017023172.1 XM_017023173.1

SNP Detail

SNP ID Chromosome Location
rs964000248 Chr.16: 9755024-9755024 on Build GRCh38
rs1021237120 Chr.16: 9754800-9754800 on Build GRCh38
rs544326150 Chr.16: 9754787-9754787 on Build GRCh38
rs768344069 Chr.16: 9754613-9754613 on Build GRCh38
rs1018579436 Chr.16: 9755015-9755015 on Build GRCh38
rs926909971 Chr.16: 9754770-9754770 on Build GRCh38
rs534681142 Chr.16: 9754659-9754659 on Build GRCh38
rs918116218 Chr.16: 9754751-9754751 on Build GRCh38
rs936969459 Chr.16: 9754771-9754771 on Build GRCh38
rs865965747 Chr.16: 9754996-9754996 on Build GRCh38
rs142313981 Chr.16: 9754862-9754862 on Build GRCh38
rs995747604 Chr.16: 9754741-9754741 on Build GRCh38
rs1045120956 Chr.16: 9755061-9755061 on Build GRCh38
rs951467726 Chr.16: 9754637-9754637 on Build GRCh38
rs369312311 Chr.16: 9754613-9754614 on Build GRCh38
rs886052519 Chr.16: 9754993-9754993 on Build GRCh38
rs898482347 Chr.16: 9755066-9755066 on Build GRCh38
rs16966243 Chr.16: 9755072-9755072 on Build GRCh38
rs968633942 Chr.16: 9754814-9754814 on Build GRCh38
rs552102022 Chr.16: 9754645-9754645 on Build GRCh38
rs537357132 Chr.16: 9754801-9754801 on Build GRCh38
rs192548671 Chr.16: 9754753-9754753 on Build GRCh38
rs890987963 Chr.16: 9754993-9754993 on Build GRCh38
rs767749 Chr.16: 9754614-9754614 on Build GRCh38
rs975087509 Chr.16: 9754792-9754792 on Build GRCh38
rs1010024001 Chr.16: 9754927-9754927 on Build GRCh38
rs995872112 Chr.16: 9755046-9755046 on Build GRCh38
rs573589434 Chr.16: 9754823-9754823 on Build GRCh38
rs1047279429 Chr.16: 9755089-9755089 on Build GRCh38
rs145933964 Chr.16: 9754893-9754893 on Build GRCh38
rs59371229 Chr.16: 9754612-9754613 on Build GRCh38
rs982855223 Chr.16: 9755095-9755095 on Build GRCh38
rs886052517 Chr.16: 9754751-9754751 on Build GRCh38
rs573015402 Chr.16: 9755060-9755060 on Build GRCh38
rs532201074 Chr.16: 9755032-9755032 on Build GRCh38
rs1041558755 Chr.16: 9754950-9754950 on Build GRCh38
rs373045049 Chr.16: 9754748-9754748 on Build GRCh38
rs898784271 Chr.16: 9754672-9754672 on Build GRCh38
rs947998688 Chr.16: 9754652-9754652 on Build GRCh38
rs1019323535 Chr.16: 9754780-9754780 on Build GRCh38
rs557950577 Chr.16: 9754701-9754701 on Build GRCh38
rs1045703781 Chr.16: 9754761-9754761 on Build GRCh38
rs918021343 Chr.16: 9755098-9755098 on Build GRCh38
rs557021103 Chr.16: 9754808-9754808 on Build GRCh38
rs1054130177 Chr.16: 9754783-9754783 on Build GRCh38
rs951457231 Chr.16: 9755088-9755088 on Build GRCh38
rs557541803 Chr.16: 9754736-9754736 on Build GRCh38
rs537335552 Chr.16: 9754648-9754648 on Build GRCh38
rs768445127 Chr.16: 9754894-9754894 on Build GRCh38
rs886052518 Chr.16: 9754790-9754790 on Build GRCh38
rs1062722 Chr.16: 9754834-9754834 on Build GRCh38

View Full Product Details