Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 24346637-24347036 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNG3
Entrez Gene ID
10368
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 3
Gene Aliases
-
UniGene
Hs.7235
Gene Chromosome Location
Chr.16: 24255553-24362416 on Build GRCh38
Transcripts
NM_006539.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1199389 c.226G>A p.V76M Chr.16: 24346748-24346748 on Build GRCh38
7205029 c.225C>T p.G75G Chr.16: 24346747-24346747 on Build GRCh38
5596432 c.281C>T p.A94V Chr.16: 24346803-24346803 on Build GRCh38
6144039 c.295C>T p.R99* Chr.16: 24346817-24346817 on Build GRCh38
7149861 c.244C>G p.H82D Chr.16: 24346766-24346766 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs113019847 Chr.16: 24346680-24346680 on Build GRCh38
rs9940070 Chr.16: 24346865-24346865 on Build GRCh38
rs190141813 Chr.16: 24346998-24346998 on Build GRCh38
rs1002485686 Chr.16: 24346947-24346947 on Build GRCh38
rs1033869132 Chr.16: 24346713-24346713 on Build GRCh38
rs748289025 Chr.16: 24346707-24346707 on Build GRCh38
rs527836962 Chr.16: 24346964-24346964 on Build GRCh38
rs756163536 Chr.16: 24346781-24346781 on Build GRCh38
rs752757837 Chr.16: 24346774-24346774 on Build GRCh38
rs886414951 Chr.16: 24346894-24346894 on Build GRCh38
rs193097772 Chr.16: 24346837-24346837 on Build GRCh38
rs111513623 Chr.16: 24346754-24346754 on Build GRCh38
rs9929236 Chr.16: 24346923-24346923 on Build GRCh38
rs201670064 Chr.16: 24346699-24346699 on Build GRCh38
rs773069719 Chr.16: 24346836-24346836 on Build GRCh38
rs140855664 Chr.16: 24346747-24346747 on Build GRCh38
rs781463498 Chr.16: 24346771-24346771 on Build GRCh38
rs779398247 Chr.16: 24346813-24346813 on Build GRCh38
rs771900152 Chr.16: 24346731-24346731 on Build GRCh38
rs772990500 Chr.16: 24346838-24346838 on Build GRCh38
rs771606027 Chr.16: 24346811-24346811 on Build GRCh38
rs551077857 Chr.16: 24346717-24346717 on Build GRCh38
rs962508717 Chr.16: 24346703-24346703 on Build GRCh38
rs749791046 Chr.16: 24346802-24346802 on Build GRCh38
rs1016795337 Chr.16: 24346700-24346700 on Build GRCh38
rs771845014 Chr.16: 24346822-24346822 on Build GRCh38
rs200755015 Chr.16: 24346817-24346817 on Build GRCh38
rs5816249 Chr.16: 24346677-24346678 on Build GRCh38
rs759233611 Chr.16: 24346730-24346730 on Build GRCh38
rs978141428 Chr.16: 24346994-24346994 on Build GRCh38
rs368528326 Chr.16: 24346789-24346789 on Build GRCh38
rs1022507800 Chr.16: 24346978-24346978 on Build GRCh38
rs375344869 Chr.16: 24346743-24346743 on Build GRCh38
rs9929298 Chr.16: 24346897-24346897 on Build GRCh38
rs776196785 Chr.16: 24346896-24346896 on Build GRCh38
rs765043632 Chr.16: 24346762-24346762 on Build GRCh38
rs111726266 Chr.16: 24346685-24346685 on Build GRCh38
rs776588148 Chr.16: 24346830-24346830 on Build GRCh38
rs767672155 Chr.16: 24346864-24346864 on Build GRCh38
rs755183039 Chr.16: 24346766-24346766 on Build GRCh38
rs377185822 Chr.16: 24346715-24346715 on Build GRCh38
rs548079872 Chr.16: 24346989-24346989 on Build GRCh38
rs1002029917 Chr.16: 24346712-24346712 on Build GRCh38
rs556129837 Chr.16: 24346661-24346661 on Build GRCh38
rs138667202 Chr.16: 24346892-24346892 on Build GRCh38
rs1006731150 Chr.16: 24346925-24346925 on Build GRCh38
rs757005088 Chr.16: 24346683-24346683 on Build GRCh38
rs371386260 Chr.16: 24346828-24346828 on Build GRCh38
rs386384490 Chr.16: 24346678-24346679 on Build GRCh38
rs960747036 Chr.16: 24346982-24346982 on Build GRCh38
rs772013394 Chr.16: 24346704-24346704 on Build GRCh38
rs774108625 Chr.16: 24346724-24346724 on Build GRCh38
rs1005237343 Chr.16: 24346869-24346869 on Build GRCh38
rs185111555 Chr.16: 24346893-24346893 on Build GRCh38
rs9929254 Chr.16: 24346662-24346662 on Build GRCh38
rs915320855 Chr.16: 24346695-24346695 on Build GRCh38
rs751799107 Chr.16: 24346702-24346702 on Build GRCh38
rs150135113 Chr.16: 24346748-24346748 on Build GRCh38
rs181334604 Chr.16: 24347007-24347007 on Build GRCh38
rs762744084 Chr.16: 24346849-24346849 on Build GRCh38
rs775284966 Chr.16: 24346827-24346827 on Build GRCh38
rs750272065 Chr.16: 24346763-24346763 on Build GRCh38
rs370375292 Chr.16: 24346720-24346720 on Build GRCh38
rs1038324198 Chr.16: 24346905-24346905 on Build GRCh38
rs577728796 Chr.16: 24346708-24346708 on Build GRCh38
rs1036670517 Chr.16: 24346876-24346876 on Build GRCh38
rs530419346 Chr.16: 24347004-24347004 on Build GRCh38
rs768070765 Chr.16: 24346829-24346829 on Build GRCh38
rs35638594 Chr.16: 24346679-24346680 on Build GRCh38

View Full Product Details