Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30902986-30903481 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTF1
Entrez Gene ID
1489
Gene Name
cardiotrophin 1
Gene Aliases
CT-1, CT1
UniGene
Hs.483811
Gene Chromosome Location
Chr.16: 30895824-30903560 on Build GRCh38
Transcripts
NM_001142544.1 NM_001330.3 XM_011545759.2 XM_011545760.2

SNP Detail

SNP ID Chromosome Location
rs114618084 Chr.16: 30903018-30903018 on Build GRCh38
rs938100118 Chr.16: 30903126-30903126 on Build GRCh38
rs767914238 Chr.16: 30903198-30903198 on Build GRCh38
rs928068442 Chr.16: 30903113-30903113 on Build GRCh38
rs1033608204 Chr.16: 30903446-30903446 on Build GRCh38
rs996571880 Chr.16: 30903267-30903267 on Build GRCh38
rs766429142 Chr.16: 30903325-30903325 on Build GRCh38
rs1042829713 Chr.16: 30903398-30903398 on Build GRCh38
rs61735542 Chr.16: 30903041-30903041 on Build GRCh38
rs886051929 Chr.16: 30903448-30903448 on Build GRCh38
rs910401969 Chr.16: 30903421-30903439 on Build GRCh38
rs911553568 Chr.16: 30903284-30903284 on Build GRCh38
rs1870292 Chr.16: 30903074-30903074 on Build GRCh38
rs1035191400 Chr.16: 30903397-30903399 on Build GRCh38
rs547167699 Chr.16: 30903089-30903089 on Build GRCh38
rs982284869 Chr.16: 30903095-30903095 on Build GRCh38
rs536632807 Chr.16: 30903424-30903424 on Build GRCh38
rs1020950214 Chr.16: 30903040-30903040 on Build GRCh38
rs553684900 Chr.16: 30903445-30903445 on Build GRCh38
rs921494685 Chr.16: 30903306-30903306 on Build GRCh38
rs565672356 Chr.16: 30903193-30903193 on Build GRCh38
rs1053862193 Chr.16: 30903335-30903335 on Build GRCh38
rs137870930 Chr.16: 30903169-30903169 on Build GRCh38
rs539240936 Chr.16: 30903182-30903182 on Build GRCh38
rs893876736 Chr.16: 30903025-30903025 on Build GRCh38
rs957675131 Chr.16: 30903304-30903304 on Build GRCh38
rs944340406 Chr.16: 30903287-30903287 on Build GRCh38
rs190346372 Chr.16: 30903019-30903019 on Build GRCh38
rs1046276 Chr.16: 30903305-30903305 on Build GRCh38
rs983562699 Chr.16: 30903244-30903244 on Build GRCh38
rs753801790 Chr.16: 30903382-30903382 on Build GRCh38
rs920607764 Chr.16: 30903430-30903430 on Build GRCh38
rs1010005329 Chr.16: 30903027-30903027 on Build GRCh38
rs749251054 Chr.16: 30903318-30903318 on Build GRCh38
rs555747301 Chr.16: 30903347-30903347 on Build GRCh38
rs965137300 Chr.16: 30903021-30903021 on Build GRCh38
rs900959893 Chr.16: 30903245-30903246 on Build GRCh38
rs61733966 Chr.16: 30903383-30903383 on Build GRCh38
rs8045936 Chr.16: 30903243-30903243 on Build GRCh38
rs886956348 Chr.16: 30903225-30903225 on Build GRCh38
rs900021166 Chr.16: 30903016-30903016 on Build GRCh38
rs886051928 Chr.16: 30903290-30903290 on Build GRCh38
rs8061920 Chr.16: 30903416-30903416 on Build GRCh38
rs547270645 Chr.16: 30903094-30903094 on Build GRCh38
rs996943751 Chr.16: 30903022-30903022 on Build GRCh38

View Full Product Details