Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 31131343-31131841 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KAT8
Entrez Gene ID
84148
Gene Name
lysine acetyltransferase 8
Gene Aliases
MOF, MYST1, ZC2HC8, hMOF
UniGene
Hs.533803
Gene Chromosome Location
Chr.16: 31117664-31131393 on Build GRCh38
Transcripts
NM_032188.2 NM_182958.2 XM_011545970.1 XM_011545971.1 NM_002773.4

Gene Symbol
PRSS8
Entrez Gene ID
5652
Gene Name
protease, serine 8
Gene Aliases
CAP1, PROSTASIN
UniGene
Hs.75799
Gene Chromosome Location
Chr.16: 31131433-31135762 on Build GRCh38
Transcripts
NM_032188.2 NM_182958.2 XM_011545970.1 XM_011545971.1 NM_002773.4

SNP Detail

SNP ID Chromosome Location
rs866113492 Chr.16: 31131673-31131673 on Build GRCh38
rs934118491 Chr.16: 31131671-31131671 on Build GRCh38
rs562177063 Chr.16: 31131513-31131513 on Build GRCh38
rs375119310 Chr.16: 31131371-31131371 on Build GRCh38
rs909820788 Chr.16: 31131605-31131605 on Build GRCh38
rs901776361 Chr.16: 31131720-31131720 on Build GRCh38
rs1051240844 Chr.16: 31131427-31131427 on Build GRCh38
rs549472044 Chr.16: 31131666-31131666 on Build GRCh38
rs1051189028 Chr.16: 31131716-31131716 on Build GRCh38
rs79988879 Chr.16: 31131810-31131810 on Build GRCh38
rs140647741 Chr.16: 31131542-31131542 on Build GRCh38
rs192276914 Chr.16: 31131396-31131396 on Build GRCh38
rs779276225 Chr.16: 31131573-31131573 on Build GRCh38
rs1022494368 Chr.16: 31131543-31131543 on Build GRCh38
rs897139894 Chr.16: 31131460-31131460 on Build GRCh38
rs78264150 Chr.16: 31131401-31131401 on Build GRCh38
rs916315837 Chr.16: 31131656-31131656 on Build GRCh38
rs529440004 Chr.16: 31131548-31131548 on Build GRCh38
rs951267008 Chr.16: 31131806-31131806 on Build GRCh38
rs117502363 Chr.16: 31131768-31131768 on Build GRCh38
rs561465962 Chr.16: 31131489-31131489 on Build GRCh38
rs183961459 Chr.16: 31131639-31131639 on Build GRCh38
rs533454694 Chr.16: 31131549-31131549 on Build GRCh38
rs983076561 Chr.16: 31131590-31131590 on Build GRCh38
rs572356871 Chr.16: 31131395-31131395 on Build GRCh38
rs904127800 Chr.16: 31131757-31131757 on Build GRCh38
rs567778191 Chr.16: 31131668-31131668 on Build GRCh38
rs940547533 Chr.16: 31131672-31131672 on Build GRCh38
rs552092450 Chr.16: 31131569-31131569 on Build GRCh38
rs1034050174 Chr.16: 31131773-31131773 on Build GRCh38
rs557468105 Chr.16: 31131426-31131426 on Build GRCh38
rs1022550082 Chr.16: 31131451-31131451 on Build GRCh38
rs892592242 Chr.16: 31131748-31131748 on Build GRCh38
rs188294450 Chr.16: 31131800-31131800 on Build GRCh38
rs1036209206 Chr.16: 31131683-31131683 on Build GRCh38
rs117174323 Chr.16: 31131521-31131521 on Build GRCh38
rs998154747 Chr.16: 31131476-31131476 on Build GRCh38
rs951254453 Chr.16: 31131555-31131555 on Build GRCh38
rs968759448 Chr.16: 31131488-31131488 on Build GRCh38
rs576031368 Chr.16: 31131433-31131433 on Build GRCh38
rs998297124 Chr.16: 31131383-31131383 on Build GRCh38
rs544693958 Chr.16: 31131670-31131670 on Build GRCh38
rs757834422 Chr.16: 31131561-31131561 on Build GRCh38
rs878996764 Chr.16: 31131493-31131493 on Build GRCh38
rs933953291 Chr.16: 31131724-31131724 on Build GRCh38
rs1013979449 Chr.16: 31131787-31131787 on Build GRCh38
rs1004787134 Chr.16: 31131440-31131440 on Build GRCh38
rs117219760 Chr.16: 31131446-31131446 on Build GRCh38
rs535158681 Chr.16: 31131749-31131749 on Build GRCh38
rs1022865696 Chr.16: 31131783-31131783 on Build GRCh38
rs958441492 Chr.16: 31131775-31131775 on Build GRCh38
rs1039709913 Chr.16: 31131665-31131665 on Build GRCh38
rs553989509 Chr.16: 31131762-31131762 on Build GRCh38
rs1044343168 Chr.16: 31131754-31131754 on Build GRCh38
rs369812168 Chr.16: 31131706-31131706 on Build GRCh38
rs569910615 Chr.16: 31131609-31131609 on Build GRCh38
rs565931158 Chr.16: 31131771-31131771 on Build GRCh38
rs755318160 Chr.16: 31131522-31131522 on Build GRCh38
rs1051142241 Chr.16: 31131734-31131734 on Build GRCh38
rs978360862 Chr.16: 31131552-31131552 on Build GRCh38

View Full Product Details