Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 31916599-31917110 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF267
Entrez Gene ID
10308
Gene Name
zinc finger protein 267
Gene Aliases
HZF2
UniGene
Hs.460645
Gene Chromosome Location
Chr.16: 31873758-31917951 on Build GRCh38
Transcripts
NM_001265588.1 NM_003414.5 NR_049749.1 XM_017022865.1 XM_017022866.1

SNP Detail

SNP ID Chromosome Location
rs542705483 Chr.16: 31916730-31916730 on Build GRCh38
rs1032392084 Chr.16: 31916866-31916866 on Build GRCh38
rs917810465 Chr.16: 31916650-31916650 on Build GRCh38
rs956787419 Chr.16: 31916868-31916868 on Build GRCh38
rs751239275 Chr.16: 31916776-31916776 on Build GRCh38
rs895991904 Chr.16: 31916819-31916819 on Build GRCh38
rs1045013253 Chr.16: 31916655-31916655 on Build GRCh38
rs756811002 Chr.16: 31916806-31916806 on Build GRCh38
rs926598951 Chr.16: 31916959-31916959 on Build GRCh38
rs929039563 Chr.16: 31917056-31917056 on Build GRCh38
rs913359726 Chr.16: 31917054-31917054 on Build GRCh38
rs565213342 Chr.16: 31916887-31916887 on Build GRCh38
rs968778379 Chr.16: 31917080-31917080 on Build GRCh38
rs1005934455 Chr.16: 31916870-31916870 on Build GRCh38
rs1030228816 Chr.16: 31916962-31916962 on Build GRCh38
rs947426453 Chr.16: 31916768-31916768 on Build GRCh38
rs964378063 Chr.16: 31916939-31916939 on Build GRCh38
rs779139384 Chr.16: 31916864-31916864 on Build GRCh38
rs1057357736 Chr.16: 31916809-31916809 on Build GRCh38
rs550712461 Chr.16: 31916955-31916955 on Build GRCh38
rs1047238346 Chr.16: 31916777-31916777 on Build GRCh38
rs545189982 Chr.16: 31916667-31916667 on Build GRCh38
rs559958882 Chr.16: 31917039-31917039 on Build GRCh38
rs1000872634 Chr.16: 31916859-31916860 on Build GRCh38
rs1018565057 Chr.16: 31916800-31916800 on Build GRCh38
rs954544560 Chr.16: 31916686-31916686 on Build GRCh38
rs543973578 Chr.16: 31916999-31916999 on Build GRCh38
rs569428752 Chr.16: 31917068-31917068 on Build GRCh38
rs753927561 Chr.16: 31916694-31916697 on Build GRCh38
rs949343629 Chr.16: 31916652-31916652 on Build GRCh38
rs368038327 Chr.16: 31916965-31916965 on Build GRCh38
rs563483705 Chr.16: 31916700-31916700 on Build GRCh38
rs1008884192 Chr.16: 31916785-31916785 on Build GRCh38
rs973057161 Chr.16: 31917019-31917019 on Build GRCh38
rs987947172 Chr.16: 31916895-31916895 on Build GRCh38
rs987979924 Chr.16: 31916629-31916629 on Build GRCh38
rs755536113 Chr.16: 31916992-31916992 on Build GRCh38
rs994329504 Chr.16: 31916838-31916838 on Build GRCh38
rs181982743 Chr.16: 31916769-31916769 on Build GRCh38
rs988877752 Chr.16: 31916714-31916714 on Build GRCh38
rs926268806 Chr.16: 31916678-31916678 on Build GRCh38
rs546220550 Chr.16: 31916877-31916877 on Build GRCh38
rs954729558 Chr.16: 31916977-31916977 on Build GRCh38
rs878908532 Chr.16: 31916937-31916941 on Build GRCh38
rs1047544902 Chr.16: 31916846-31916846 on Build GRCh38
rs780785306 Chr.16: 31916812-31916812 on Build GRCh38
rs970614900 Chr.16: 31916947-31916947 on Build GRCh38
rs886186059 Chr.16: 31916853-31916853 on Build GRCh38
rs116511896 Chr.16: 31916906-31916906 on Build GRCh38
rs748449986 Chr.16: 31917027-31917027 on Build GRCh38
rs897588445 Chr.16: 31916766-31916766 on Build GRCh38
rs528076123 Chr.16: 31916852-31916852 on Build GRCh38
rs891318231 Chr.16: 31916779-31916779 on Build GRCh38
rs575203674 Chr.16: 31916724-31916724 on Build GRCh38
rs927260320 Chr.16: 31916801-31916801 on Build GRCh38

View Full Product Details