Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 71635363-71635872 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MARVELD3
Entrez Gene ID
91862
Gene Name
MARVEL domain containing 3
Gene Aliases
MARVD3, MRVLDC3
UniGene
Hs.513706
Gene Chromosome Location
Chr.16: 71626137-71641965 on Build GRCh38
Transcripts
NM_001017967.3 NM_001271329.1 NM_052858.5 XM_011523449.2

SNP Detail

SNP ID Chromosome Location
rs185872554 Chr.16: 71635536-71635536 on Build GRCh38
rs950130989 Chr.16: 71635662-71635662 on Build GRCh38
rs936063674 Chr.16: 71635448-71635448 on Build GRCh38
rs955970985 Chr.16: 71635437-71635437 on Build GRCh38
rs144275428 Chr.16: 71635425-71635425 on Build GRCh38
rs573144138 Chr.16: 71635584-71635584 on Build GRCh38
rs938873388 Chr.16: 71635523-71635523 on Build GRCh38
rs565831026 Chr.16: 71635715-71635715 on Build GRCh38
rs754671983 Chr.16: 71635685-71635686 on Build GRCh38
rs938457775 Chr.16: 71635829-71635829 on Build GRCh38
rs561134720 Chr.16: 71635544-71635544 on Build GRCh38
rs540749087 Chr.16: 71635585-71635585 on Build GRCh38
rs376362936 Chr.16: 71635810-71635810 on Build GRCh38
rs558183359 Chr.16: 71635412-71635412 on Build GRCh38
rs928807097 Chr.16: 71635499-71635499 on Build GRCh38
rs113823163 Chr.16: 71635552-71635552 on Build GRCh38
rs926139830 Chr.16: 71635600-71635600 on Build GRCh38
rs1345868 Chr.16: 71635721-71635721 on Build GRCh38
rs765883593 Chr.16: 71635793-71635793 on Build GRCh38
rs533516832 Chr.16: 71635832-71635832 on Build GRCh38
rs983991026 Chr.16: 71635830-71635830 on Build GRCh38
rs979858958 Chr.16: 71635791-71635791 on Build GRCh38
rs141313724 Chr.16: 71635750-71635750 on Build GRCh38
rs147801483 Chr.16: 71635435-71635435 on Build GRCh38
rs189861020 Chr.16: 71635596-71635596 on Build GRCh38
rs950043398 Chr.16: 71635518-71635518 on Build GRCh38
rs73576292 Chr.16: 71635628-71635628 on Build GRCh38
rs948470938 Chr.16: 71635472-71635472 on Build GRCh38
rs997834692 Chr.16: 71635822-71635822 on Build GRCh38
rs1044572638 Chr.16: 71635497-71635497 on Build GRCh38
rs989306289 Chr.16: 71635616-71635617 on Build GRCh38
rs779972257 Chr.16: 71635418-71635418 on Build GRCh38
rs1029264172 Chr.16: 71635823-71635823 on Build GRCh38
rs187944342 Chr.16: 71635740-71635740 on Build GRCh38
rs954280292 Chr.16: 71635599-71635599 on Build GRCh38
rs1037665521 Chr.16: 71635556-71635556 on Build GRCh38
rs796686641 Chr.16: 71635616-71635616 on Build GRCh38
rs900540421 Chr.16: 71635570-71635570 on Build GRCh38
rs762681501 Chr.16: 71635784-71635784 on Build GRCh38
rs879004070 Chr.16: 71635646-71635646 on Build GRCh38
rs547347475 Chr.16: 71635566-71635566 on Build GRCh38
rs192508525 Chr.16: 71635799-71635799 on Build GRCh38
rs78688791 Chr.16: 71635627-71635628 on Build GRCh38
rs1031475595 Chr.16: 71635660-71635660 on Build GRCh38
rs1005345605 Chr.16: 71635770-71635770 on Build GRCh38
rs886661947 Chr.16: 71635541-71635541 on Build GRCh38
rs1028152131 Chr.16: 71635402-71635402 on Build GRCh38
rs953689049 Chr.16: 71635849-71635849 on Build GRCh38
rs989742743 Chr.16: 71635445-71635445 on Build GRCh38
rs990066954 Chr.16: 71635732-71635732 on Build GRCh38
rs1045749599 Chr.16: 71635665-71635665 on Build GRCh38
rs548826137 Chr.16: 71635778-71635778 on Build GRCh38
rs574641197 Chr.16: 71635463-71635463 on Build GRCh38
rs1054948097 Chr.16: 71635554-71635554 on Build GRCh38
rs183365853 Chr.16: 71635433-71635433 on Build GRCh38

View Full Product Details