Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 71647590-71648089 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHLPP2
Entrez Gene ID
23035
Gene Name
PH domain and leucine rich repeat protein phosphatase 2
Gene Aliases
PHLPPL, PPM3B
UniGene
Hs.709458
Gene Chromosome Location
Chr.16: 71644924-71724701 on Build GRCh38
Transcripts
NM_001289003.1 NM_015020.3

SNP Detail

SNP ID Chromosome Location
rs899149465 Chr.16: 71647971-71647971 on Build GRCh38
rs184110649 Chr.16: 71647782-71647782 on Build GRCh38
rs959121135 Chr.16: 71647839-71647839 on Build GRCh38
rs1029452455 Chr.16: 71648015-71648015 on Build GRCh38
rs536083222 Chr.16: 71647781-71647781 on Build GRCh38
rs963419338 Chr.16: 71647632-71647632 on Build GRCh38
rs941612861 Chr.16: 71647625-71647625 on Build GRCh38
rs1004837816 Chr.16: 71647622-71647622 on Build GRCh38
rs2052585 Chr.16: 71647679-71647679 on Build GRCh38
rs973425965 Chr.16: 71647932-71647932 on Build GRCh38
rs1054866494 Chr.16: 71647721-71647721 on Build GRCh38
rs912465388 Chr.16: 71647722-71647722 on Build GRCh38
rs545367103 Chr.16: 71647964-71647964 on Build GRCh38
rs557222841 Chr.16: 71648006-71648006 on Build GRCh38
rs759394877 Chr.16: 71647717-71647717 on Build GRCh38
rs566894637 Chr.16: 71647861-71647861 on Build GRCh38
rs77559569 Chr.16: 71647813-71647813 on Build GRCh38
rs189132896 Chr.16: 71648026-71648026 on Build GRCh38
rs181117882 Chr.16: 71647681-71647681 on Build GRCh38
rs922060571 Chr.16: 71647646-71647646 on Build GRCh38
rs4788822 Chr.16: 71647612-71647612 on Build GRCh38
rs1034785690 Chr.16: 71647821-71647821 on Build GRCh38
rs944001355 Chr.16: 71647740-71647740 on Build GRCh38
rs1014862014 Chr.16: 71647959-71647959 on Build GRCh38
rs909581080 Chr.16: 71647872-71647872 on Build GRCh38
rs934134101 Chr.16: 71647666-71647666 on Build GRCh38
rs534582133 Chr.16: 71647891-71647891 on Build GRCh38
rs373247552 Chr.16: 71647945-71647945 on Build GRCh38
rs554468124 Chr.16: 71647946-71647946 on Build GRCh38
rs905621717 Chr.16: 71647869-71647869 on Build GRCh38
rs149295364 Chr.16: 71647701-71647701 on Build GRCh38
rs566284502 Chr.16: 71647811-71647811 on Build GRCh38
rs889893545 Chr.16: 71647705-71647705 on Build GRCh38
rs994813113 Chr.16: 71647990-71647990 on Build GRCh38
rs865949954 Chr.16: 71647812-71647812 on Build GRCh38
rs1051279069 Chr.16: 71648024-71648024 on Build GRCh38
rs942800009 Chr.16: 71648053-71648053 on Build GRCh38
rs912925021 Chr.16: 71647798-71647798 on Build GRCh38
rs1003347788 Chr.16: 71647815-71647816 on Build GRCh38
rs973485289 Chr.16: 71647645-71647645 on Build GRCh38
rs550430747 Chr.16: 71647699-71647699 on Build GRCh38
rs896049092 Chr.16: 71647898-71647898 on Build GRCh38
rs558783898 Chr.16: 71647904-71647904 on Build GRCh38
rs577986764 Chr.16: 71647924-71647924 on Build GRCh38
rs1057341156 Chr.16: 71647895-71647895 on Build GRCh38
rs747045576 Chr.16: 71647980-71647980 on Build GRCh38
rs80274051 Chr.16: 71647626-71647626 on Build GRCh38
rs893132888 Chr.16: 71647759-71647759 on Build GRCh38
rs1011273832 Chr.16: 71647760-71647762 on Build GRCh38
rs767542547 Chr.16: 71648009-71648009 on Build GRCh38
rs774538105 Chr.16: 71647875-71647875 on Build GRCh38

View Full Product Details