Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 81785856-81786359 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLCG2
Entrez Gene ID
5336
Gene Name
phospholipase C gamma 2
Gene Aliases
APLAID, FCAS3, PLC-IV, PLC-gamma-2
UniGene
Hs.372303
Gene Chromosome Location
Chr.16: 81779258-81962693 on Build GRCh38
Transcripts
NM_002661.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
704621 c.138C>T p.I46I Chr.16: 81786127-81786127 on Build GRCh38
51195 c.1-4G>A p.? Chr.16: 81785986-81785986 on Build GRCh38
4484386 c.27C>T p.S9S Chr.16: 81786016-81786016 on Build GRCh38
4483431 c.26C>T p.S9F Chr.16: 81786015-81786015 on Build GRCh38
4788147 c.166A>C p.K56Q Chr.16: 81786155-81786155 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753845661 Chr.16: 81786162-81786162 on Build GRCh38
rs552409251 Chr.16: 81785958-81785958 on Build GRCh38
rs1034788127 Chr.16: 81786253-81786253 on Build GRCh38
rs761554137 Chr.16: 81785988-81785988 on Build GRCh38
rs763818041 Chr.16: 81785948-81785948 on Build GRCh38
rs1143685 Chr.16: 81786163-81786163 on Build GRCh38
rs34850739 Chr.16: 81785883-81785884 on Build GRCh38
rs4354929 Chr.16: 81785881-81785881 on Build GRCh38
rs760188007 Chr.16: 81786088-81786088 on Build GRCh38
rs757139873 Chr.16: 81786195-81786195 on Build GRCh38
rs143618645 Chr.16: 81785897-81785897 on Build GRCh38
rs781194589 Chr.16: 81786200-81786200 on Build GRCh38
rs780153793 Chr.16: 81785999-81785999 on Build GRCh38
rs184923289 Chr.16: 81785916-81785916 on Build GRCh38
rs369542354 Chr.16: 81786046-81786046 on Build GRCh38
rs4294811 Chr.16: 81786215-81786215 on Build GRCh38
rs201282710 Chr.16: 81786194-81786194 on Build GRCh38
rs563795527 Chr.16: 81785936-81785936 on Build GRCh38
rs368137889 Chr.16: 81786122-81786122 on Build GRCh38
rs777730567 Chr.16: 81786129-81786129 on Build GRCh38
rs763960656 Chr.16: 81785954-81785954 on Build GRCh38
rs530412276 Chr.16: 81785888-81785888 on Build GRCh38
rs779108574 Chr.16: 81786118-81786118 on Build GRCh38
rs553657822 Chr.16: 81786172-81786172 on Build GRCh38
rs1020348923 Chr.16: 81786305-81786305 on Build GRCh38
rs369426093 Chr.16: 81786214-81786214 on Build GRCh38
rs556174475 Chr.16: 81786002-81786002 on Build GRCh38
rs769990892 Chr.16: 81785998-81785998 on Build GRCh38
rs139488059 Chr.16: 81786303-81786303 on Build GRCh38
rs1050133080 Chr.16: 81786033-81786033 on Build GRCh38
rs114215669 Chr.16: 81785927-81785927 on Build GRCh38
rs775349663 Chr.16: 81785952-81785952 on Build GRCh38
rs765042969 Chr.16: 81786098-81786098 on Build GRCh38
rs199972098 Chr.16: 81786000-81786000 on Build GRCh38
rs1143685 Chr.16: 81786163-81786163 on Build GRCh38
rs764524085 Chr.16: 81786035-81786035 on Build GRCh38
rs919214759 Chr.16: 81786220-81786220 on Build GRCh38
rs752018966 Chr.16: 81786037-81786037 on Build GRCh38
rs749945394 Chr.16: 81785960-81785960 on Build GRCh38
rs752145398 Chr.16: 81785981-81785981 on Build GRCh38
rs370242901 Chr.16: 81786138-81786138 on Build GRCh38
rs374354863 Chr.16: 81786160-81786160 on Build GRCh38
rs202002041 Chr.16: 81786006-81786006 on Build GRCh38
rs769959186 Chr.16: 81785950-81785950 on Build GRCh38
rs767741418 Chr.16: 81786148-81786148 on Build GRCh38
rs758690442 Chr.16: 81786192-81786192 on Build GRCh38
rs748139734 Chr.16: 81786188-81786194 on Build GRCh38
rs376949064 Chr.16: 81786001-81786001 on Build GRCh38
rs778378424 Chr.16: 81785980-81785980 on Build GRCh38
rs763370826 Chr.16: 81786025-81786025 on Build GRCh38
rs370683432 Chr.16: 81786139-81786139 on Build GRCh38
rs753043745 Chr.16: 81785973-81785973 on Build GRCh38
rs748925816 Chr.16: 81786208-81786208 on Build GRCh38
rs755775476 Chr.16: 81785964-81785964 on Build GRCh38
rs141718763 Chr.16: 81785949-81785949 on Build GRCh38
rs189301790 Chr.16: 81786066-81786066 on Build GRCh38
rs969672855 Chr.16: 81785963-81785963 on Build GRCh38
rs770382676 Chr.16: 81786089-81786089 on Build GRCh38
rs777408502 Chr.16: 81786193-81786193 on Build GRCh38
rs750388097 Chr.16: 81786040-81786040 on Build GRCh38
rs762228969 Chr.16: 81786144-81786144 on Build GRCh38
rs766981162 Chr.16: 81785959-81785959 on Build GRCh38
rs148066456 Chr.16: 81785907-81785907 on Build GRCh38
rs372331728 Chr.16: 81785921-81785921 on Build GRCh38
rs759758361 Chr.16: 81785934-81785934 on Build GRCh38
rs751247647 Chr.16: 81786228-81786228 on Build GRCh38
rs773040139 Chr.16: 81786086-81786086 on Build GRCh38
rs1045529685 Chr.16: 81786326-81786326 on Build GRCh38
rs780660956 Chr.16: 81786056-81786096 on Build GRCh38
rs753458249 Chr.16: 81786021-81786021 on Build GRCh38
rs988309398 Chr.16: 81786182-81786182 on Build GRCh38
rs778662619 Chr.16: 81786074-81786074 on Build GRCh38
rs759241889 Chr.16: 81786093-81786093 on Build GRCh38
rs746891285 Chr.16: 81786131-81786131 on Build GRCh38
rs749437141 Chr.16: 81786067-81786067 on Build GRCh38
rs764445057 Chr.16: 81786227-81786227 on Build GRCh38
rs753972799 Chr.16: 81786115-81786115 on Build GRCh38
rs772051902 Chr.16: 81786013-81786013 on Build GRCh38
rs758215832 Chr.16: 81786128-81786128 on Build GRCh38
rs917270590 Chr.16: 81785883-81785883 on Build GRCh38
rs745994157 Chr.16: 81785986-81785986 on Build GRCh38
rs777617388 Chr.16: 81786222-81786222 on Build GRCh38
rs756167862 Chr.16: 81786061-81786061 on Build GRCh38
rs1048773758 Chr.16: 81786190-81786190 on Build GRCh38
rs887568232 Chr.16: 81786278-81786278 on Build GRCh38
rs766974268 Chr.16: 81786161-81786161 on Build GRCh38
rs181655438 Chr.16: 81786207-81786207 on Build GRCh38
rs1001450591 Chr.16: 81786248-81786248 on Build GRCh38
rs9936371 Chr.16: 81786011-81786011 on Build GRCh38
rs957371373 Chr.16: 81786292-81786292 on Build GRCh38
rs766002121 Chr.16: 81786017-81786017 on Build GRCh38
rs546366286 Chr.16: 81786323-81786323 on Build GRCh38
rs750924322 Chr.16: 81786103-81786103 on Build GRCh38
rs372502550 Chr.16: 81786077-81786077 on Build GRCh38
rs944338936 Chr.16: 81785946-81785946 on Build GRCh38
rs769712189 Chr.16: 81786200-81786201 on Build GRCh38
rs762659564 Chr.16: 81785953-81785953 on Build GRCh38
rs757096580 Chr.16: 81786233-81786233 on Build GRCh38
rs765272795 Chr.16: 81786219-81786219 on Build GRCh38
rs61749044 Chr.16: 81786071-81786071 on Build GRCh38
rs537204469 Chr.16: 81786090-81786090 on Build GRCh38
rs1051325187 Chr.16: 81785931-81785931 on Build GRCh38
rs773671139 Chr.16: 81786154-81786154 on Build GRCh38
rs759576530 Chr.16: 81786217-81786217 on Build GRCh38
rs772778136 Chr.16: 81786016-81786016 on Build GRCh38
rs150565405 Chr.16: 81785982-81785982 on Build GRCh38
rs770951220 Chr.16: 81786137-81786137 on Build GRCh38
rs752952252 Chr.16: 81786178-81786178 on Build GRCh38
rs138699122 Chr.16: 81785985-81785985 on Build GRCh38
rs752823843 Chr.16: 81786226-81786226 on Build GRCh38
rs374235189 Chr.16: 81786052-81786052 on Build GRCh38
rs532678378 Chr.16: 81786146-81786146 on Build GRCh38
rs530142575 Chr.16: 81785920-81785920 on Build GRCh38
rs147349332 Chr.16: 81786099-81786099 on Build GRCh38
rs1023636697 Chr.16: 81786127-81786127 on Build GRCh38
rs969768555 Chr.16: 81786136-81786136 on Build GRCh38
rs757826017 Chr.16: 81786039-81786039 on Build GRCh38
rs775303395 Chr.16: 81786142-81786142 on Build GRCh38
rs371524184 Chr.16: 81786210-81786210 on Build GRCh38
rs182080456 Chr.16: 81785886-81785886 on Build GRCh38
rs370352962 Chr.16: 81786116-81786116 on Build GRCh38
rs758705382 Chr.16: 81785975-81785975 on Build GRCh38
rs375123176 Chr.16: 81785956-81785956 on Build GRCh38

View Full Product Details