Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 1646432-1646930 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RILP
Entrez Gene ID
83547
Gene Name
Rab interacting lysosomal protein
Gene Aliases
PP10141
UniGene
Hs.534497
Gene Chromosome Location
Chr.17: 1646150-1650098 on Build GRCh38
Transcripts
NM_031430.2 XM_005256811.1 XM_017025195.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6209338 c.1165G>A p.E389K Chr.17: 1646483-1646483 on Build GRCh38
3889345 c.1083C>T p.S361S Chr.17: 1646565-1646565 on Build GRCh38
4064102 c.1055C>A p.A352D Chr.17: 1646593-1646593 on Build GRCh38
6766488 c.1179G>A p.L393L Chr.17: 1646469-1646469 on Build GRCh38
4394160 c.1090C>T p.P364S Chr.17: 1646558-1646558 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2293067 Chr.17: 1646812-1646812 on Build GRCh38
rs755001670 Chr.17: 1646872-1646872 on Build GRCh38
rs372893048 Chr.17: 1646667-1646667 on Build GRCh38
rs1039683490 Chr.17: 1646784-1646784 on Build GRCh38
rs1022317698 Chr.17: 1646875-1646875 on Build GRCh38
rs978790843 Chr.17: 1646713-1646713 on Build GRCh38
rs954954872 Chr.17: 1646637-1646637 on Build GRCh38
rs200883518 Chr.17: 1646568-1646568 on Build GRCh38
rs775154367 Chr.17: 1646620-1646620 on Build GRCh38
rs538809606 Chr.17: 1646708-1646708 on Build GRCh38
rs569195701 Chr.17: 1646780-1646780 on Build GRCh38
rs1052463022 Chr.17: 1646825-1646825 on Build GRCh38
rs112166919 Chr.17: 1646502-1646502 on Build GRCh38
rs766645693 Chr.17: 1646864-1646864 on Build GRCh38
rs368935398 Chr.17: 1646863-1646863 on Build GRCh38
rs556577180 Chr.17: 1646498-1646498 on Build GRCh38
rs773982070 Chr.17: 1646527-1646527 on Build GRCh38
rs774341024 Chr.17: 1646478-1646479 on Build GRCh38
rs770211325 Chr.17: 1646512-1646512 on Build GRCh38
rs141613365 Chr.17: 1646786-1646786 on Build GRCh38
rs751649564 Chr.17: 1646576-1646576 on Build GRCh38
rs550992717 Chr.17: 1646625-1646625 on Build GRCh38
rs141024997 Chr.17: 1646473-1646473 on Build GRCh38
rs776665812 Chr.17: 1646759-1646759 on Build GRCh38
rs755094527 Chr.17: 1646586-1646586 on Build GRCh38
rs909747805 Chr.17: 1646775-1646775 on Build GRCh38
rs11867411 Chr.17: 1646838-1646838 on Build GRCh38
rs757406116 Chr.17: 1646499-1646499 on Build GRCh38
rs992515834 Chr.17: 1646707-1646707 on Build GRCh38
rs771065621 Chr.17: 1646470-1646472 on Build GRCh38
rs1038002344 Chr.17: 1646855-1646855 on Build GRCh38
rs949976862 Chr.17: 1646807-1646807 on Build GRCh38
rs757622744 Chr.17: 1646904-1646904 on Build GRCh38
rs760643404 Chr.17: 1646453-1646453 on Build GRCh38
rs942480518 Chr.17: 1646813-1646813 on Build GRCh38
rs530867328 Chr.17: 1646611-1646611 on Build GRCh38
rs993641218 Chr.17: 1646610-1646610 on Build GRCh38
rs901083616 Chr.17: 1646787-1646787 on Build GRCh38
rs374979618 Chr.17: 1646513-1646513 on Build GRCh38
rs750685599 Chr.17: 1646483-1646483 on Build GRCh38
rs910555622 Chr.17: 1646468-1646468 on Build GRCh38
rs573268040 Chr.17: 1646844-1646844 on Build GRCh38
rs747831343 Chr.17: 1646781-1646781 on Build GRCh38
rs530327038 Chr.17: 1646826-1646826 on Build GRCh38
rs771536892 Chr.17: 1646531-1646531 on Build GRCh38
rs753110765 Chr.17: 1646876-1646876 on Build GRCh38
rs778148008 Chr.17: 1646605-1646605 on Build GRCh38
rs772839965 Chr.17: 1646905-1646905 on Build GRCh38
rs772340146 Chr.17: 1646521-1646522 on Build GRCh38
rs776635030 Chr.17: 1646559-1646559 on Build GRCh38
rs751449949 Chr.17: 1646494-1646494 on Build GRCh38
rs773093080 Chr.17: 1646665-1646665 on Build GRCh38
rs75242198 Chr.17: 1646602-1646602 on Build GRCh38
rs746437448 Chr.17: 1646623-1646623 on Build GRCh38
rs780774829 Chr.17: 1646699-1646699 on Build GRCh38
rs1033090322 Chr.17: 1646737-1646737 on Build GRCh38
rs763983256 Chr.17: 1646883-1646889 on Build GRCh38
rs775025801 Chr.17: 1646538-1646538 on Build GRCh38
rs138782493 Chr.17: 1646493-1646493 on Build GRCh38
rs896875300 Chr.17: 1646856-1646856 on Build GRCh38
rs561295774 Chr.17: 1646631-1646631 on Build GRCh38
rs760467943 Chr.17: 1646882-1646883 on Build GRCh38
rs7212294 Chr.17: 1646692-1646692 on Build GRCh38
rs1047866997 Chr.17: 1646884-1646884 on Build GRCh38
rs759771291 Chr.17: 1646506-1646521 on Build GRCh38
rs748397950 Chr.17: 1646503-1646503 on Build GRCh38
rs530201573 Chr.17: 1646643-1646643 on Build GRCh38
rs552744925 Chr.17: 1646709-1646709 on Build GRCh38
rs1009580694 Chr.17: 1646668-1646668 on Build GRCh38
rs775209590 Chr.17: 1646603-1646603 on Build GRCh38
rs1002318513 Chr.17: 1646891-1646891 on Build GRCh38
rs559730233 Chr.17: 1646574-1646574 on Build GRCh38
rs761456082 Chr.17: 1646562-1646562 on Build GRCh38
rs140403152 Chr.17: 1646477-1646477 on Build GRCh38
rs927243855 Chr.17: 1646832-1646832 on Build GRCh38
rs373358901 Chr.17: 1646871-1646871 on Build GRCh38
rs200277401 Chr.17: 1646603-1646603 on Build GRCh38
rs750644769 Chr.17: 1646564-1646564 on Build GRCh38
rs145604071 Chr.17: 1646594-1646594 on Build GRCh38
rs929760213 Chr.17: 1646712-1646712 on Build GRCh38
rs778270569 Chr.17: 1646899-1646899 on Build GRCh38
rs368307428 Chr.17: 1646558-1646558 on Build GRCh38
rs368515236 Chr.17: 1646900-1646900 on Build GRCh38
rs765136532 Chr.17: 1646563-1646563 on Build GRCh38
rs771659833 Chr.17: 1646613-1646613 on Build GRCh38
rs763922701 Chr.17: 1646456-1646456 on Build GRCh38
rs201946575 Chr.17: 1646484-1646484 on Build GRCh38
rs748750573 Chr.17: 1646599-1646599 on Build GRCh38
rs1048144408 Chr.17: 1646725-1646725 on Build GRCh38
rs998196161 Chr.17: 1646546-1646546 on Build GRCh38
rs760251391 Chr.17: 1646542-1646542 on Build GRCh38
rs756690973 Chr.17: 1646896-1646896 on Build GRCh38
rs757106622 Chr.17: 1646465-1646465 on Build GRCh38
rs535869108 Chr.17: 1646873-1646873 on Build GRCh38
rs779177436 Chr.17: 1646609-1646609 on Build GRCh38
rs917085317 Chr.17: 1646803-1646803 on Build GRCh38
rs745585158 Chr.17: 1646528-1646528 on Build GRCh38
rs368151611 Chr.17: 1646457-1646457 on Build GRCh38

View Full Product Details