Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 2060540-2061043 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMG6
Entrez Gene ID
23293
Gene Name
SMG6, nonsense mediated mRNA decay factor
Gene Aliases
C17orf31, EST1A, SMG-6, hSMG5/7a
UniGene
Hs.448342
Gene Chromosome Location
Chr.17: 2059839-2303836 on Build GRCh38
Transcripts
NM_001256827.1 NM_001256828.1 NM_001282326.1 NM_017575.4 XM_005256569.3 XM_005256571.4 XM_011523769.2 XM_011523775.2 XM_017024399.1

SNP Detail

SNP ID Chromosome Location
rs942560981 Chr.17: 2060639-2060639 on Build GRCh38
rs1008296950 Chr.17: 2060984-2060984 on Build GRCh38
rs545050733 Chr.17: 2060972-2060972 on Build GRCh38
rs947795551 Chr.17: 2060668-2060668 on Build GRCh38
rs536825457 Chr.17: 2060632-2060632 on Build GRCh38
rs573776992 Chr.17: 2060935-2060935 on Build GRCh38
rs962930037 Chr.17: 2060575-2060575 on Build GRCh38
rs11553525 Chr.17: 2060576-2060576 on Build GRCh38
rs567774344 Chr.17: 2060611-2060611 on Build GRCh38
rs542773146 Chr.17: 2060947-2060947 on Build GRCh38
rs1019738548 Chr.17: 2060987-2060987 on Build GRCh38
rs561159404 Chr.17: 2060817-2060817 on Build GRCh38
rs759891824 Chr.17: 2060702-2060705 on Build GRCh38
rs573071108 Chr.17: 2060970-2060970 on Build GRCh38
rs964032185 Chr.17: 2060995-2060995 on Build GRCh38
rs965171837 Chr.17: 2060774-2060774 on Build GRCh38
rs1044817993 Chr.17: 2060687-2060687 on Build GRCh38
rs141530811 Chr.17: 2060953-2060953 on Build GRCh38
rs913731682 Chr.17: 2060909-2060909 on Build GRCh38
rs918710862 Chr.17: 2060606-2060606 on Build GRCh38
rs1046152114 Chr.17: 2060926-2060926 on Build GRCh38
rs1003701724 Chr.17: 2060700-2060700 on Build GRCh38
rs903158 Chr.17: 2060724-2060724 on Build GRCh38
rs1007644939 Chr.17: 2060766-2060766 on Build GRCh38
rs1028055966 Chr.17: 2060751-2060751 on Build GRCh38
rs558592697 Chr.17: 2060636-2060636 on Build GRCh38
rs184838840 Chr.17: 2060978-2060978 on Build GRCh38
rs372026347 Chr.17: 2060757-2060757 on Build GRCh38
rs1034453480 Chr.17: 2060568-2060568 on Build GRCh38
rs748942904 Chr.17: 2060963-2060963 on Build GRCh38
rs930873716 Chr.17: 2060954-2060954 on Build GRCh38
rs188944682 Chr.17: 2060653-2060653 on Build GRCh38
rs1052604337 Chr.17: 2060692-2060692 on Build GRCh38
rs1026146820 Chr.17: 2060782-2060782 on Build GRCh38
rs550804441 Chr.17: 2060590-2060590 on Build GRCh38
rs995724324 Chr.17: 2060779-2060779 on Build GRCh38
rs911039981 Chr.17: 2060596-2060596 on Build GRCh38
rs530062335 Chr.17: 2060754-2060754 on Build GRCh38
rs934971663 Chr.17: 2060645-2060645 on Build GRCh38
rs12946564 Chr.17: 2060667-2060667 on Build GRCh38
rs906708664 Chr.17: 2060691-2060691 on Build GRCh38
rs904851298 Chr.17: 2060938-2060938 on Build GRCh38
rs927726353 Chr.17: 2060857-2060857 on Build GRCh38
rs939482645 Chr.17: 2060876-2060876 on Build GRCh38
rs964062286 Chr.17: 2060805-2060805 on Build GRCh38
rs566417029 Chr.17: 2060610-2060611 on Build GRCh38
rs914875652 Chr.17: 2060814-2060814 on Build GRCh38
rs980482333 Chr.17: 2060832-2060832 on Build GRCh38
rs777484755 Chr.17: 2060807-2060807 on Build GRCh38
rs986839150 Chr.17: 2060617-2060617 on Build GRCh38
rs757357842 Chr.17: 2060578-2060578 on Build GRCh38
rs975477210 Chr.17: 2061003-2061003 on Build GRCh38
rs923861754 Chr.17: 2060637-2060637 on Build GRCh38
rs368164366 Chr.17: 2060746-2060746 on Build GRCh38
rs192262828 Chr.17: 2060801-2060801 on Build GRCh38
rs1018999057 Chr.17: 2060979-2060979 on Build GRCh38
rs1032757823 Chr.17: 2061004-2061004 on Build GRCh38
rs976582796 Chr.17: 2060792-2060792 on Build GRCh38
rs949109409 Chr.17: 2060916-2060916 on Build GRCh38
rs765392253 Chr.17: 2060615-2060615 on Build GRCh38
rs956846565 Chr.17: 2060802-2060802 on Build GRCh38
rs889111741 Chr.17: 2060964-2060964 on Build GRCh38
rs994825380 Chr.17: 2060747-2060747 on Build GRCh38
rs953382630 Chr.17: 2060762-2060762 on Build GRCh38
rs1045524 Chr.17: 2060976-2060976 on Build GRCh38
rs951404925 Chr.17: 2060791-2060791 on Build GRCh38
rs534628118 Chr.17: 2060701-2060701 on Build GRCh38
rs571406691 Chr.17: 2060769-2060769 on Build GRCh38

View Full Product Details