Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 3498765-3499302 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASPA
Entrez Gene ID
443
Gene Name
aspartoacylase
Gene Aliases
ACY2, ASP
UniGene
Hs.171142
Gene Chromosome Location
Chr.17: 3474106-3502534 on Build GRCh38
Transcripts
NM_000049.2 NM_001128085.1 XM_017024661.1 NM_001321336.1 NM_001321337.1

Gene Symbol
SPATA22
Entrez Gene ID
84690
Gene Name
spermatogenesis associated 22
Gene Aliases
NYD-SP20, NYDSP20
UniGene
Hs.351068
Gene Chromosome Location
Chr.17: 3440011-3513852 on Build GRCh38
Transcripts
NM_000049.2 NM_001128085.1 XM_017024661.1 NM_001321336.1 NM_001321337.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
148150 c.831C>T p.T277T Chr.17: 3498977-3498977 on Build GRCh38
7356634 c.854A>C p.E285A Chr.17: 3499000-3499000 on Build GRCh38
5747787 c.905C>T p.T302M Chr.17: 3499051-3499051 on Build GRCh38
1640651 c.745-6T>C p.? Chr.17: 3498885-3498885 on Build GRCh38
6605948 c.806C>T p.T269M Chr.17: 3498952-3498952 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1057517450 Chr.17: 3499022-3499025 on Build GRCh38
rs1057517085 Chr.17: 3498973-3498974 on Build GRCh38
rs750505963 Chr.17: 3498984-3498984 on Build GRCh38
rs145717248 Chr.17: 3498981-3498981 on Build GRCh38
rs368363489 Chr.17: 3498868-3498868 on Build GRCh38
rs759018576 Chr.17: 3499007-3499007 on Build GRCh38
rs776283439 Chr.17: 3498865-3498869 on Build GRCh38
rs370915844 Chr.17: 3498998-3498998 on Build GRCh38
rs1050452949 Chr.17: 3499031-3499031 on Build GRCh38
rs752336086 Chr.17: 3498965-3498965 on Build GRCh38
rs993086856 Chr.17: 3498864-3498864 on Build GRCh38
rs1041721728 Chr.17: 3499273-3499273 on Build GRCh38
rs757344785 Chr.17: 3498843-3498843 on Build GRCh38
rs78677072 Chr.17: 3498977-3498977 on Build GRCh38
rs1056264045 Chr.17: 3499233-3499237 on Build GRCh38
rs779134574 Chr.17: 3498860-3498860 on Build GRCh38
rs1031834384 Chr.17: 3499126-3499126 on Build GRCh38
rs148081446 Chr.17: 3498986-3498986 on Build GRCh38
rs890496947 Chr.17: 3499045-3499045 on Build GRCh38
rs774323189 Chr.17: 3499005-3499005 on Build GRCh38
rs780003521 Chr.17: 3499062-3499062 on Build GRCh38
rs567712382 Chr.17: 3499044-3499044 on Build GRCh38
rs1057517066 Chr.17: 3498889-3498889 on Build GRCh38
rs959111875 Chr.17: 3499077-3499077 on Build GRCh38
rs778770959 Chr.17: 3499082-3499082 on Build GRCh38
rs773153121 Chr.17: 3498919-3498919 on Build GRCh38
rs755140673 Chr.17: 3499071-3499071 on Build GRCh38
rs568031094 Chr.17: 3498885-3498885 on Build GRCh38
rs755061005 Chr.17: 3498934-3498934 on Build GRCh38
rs1050505982 Chr.17: 3499259-3499259 on Build GRCh38
rs921130696 Chr.17: 3499188-3499188 on Build GRCh38
rs528643312 Chr.17: 3499267-3499267 on Build GRCh38
rs761064915 Chr.17: 3498966-3498966 on Build GRCh38
rs765819351 Chr.17: 3499056-3499056 on Build GRCh38
rs28940574 Chr.17: 3499060-3499060 on Build GRCh38
rs374525029 Chr.17: 3499140-3499140 on Build GRCh38
rs772408129 Chr.17: 3498891-3498891 on Build GRCh38
rs542964746 Chr.17: 3499135-3499135 on Build GRCh38
rs1018213095 Chr.17: 3498946-3498946 on Build GRCh38
rs762812945 Chr.17: 3498942-3498942 on Build GRCh38
rs970053573 Chr.17: 3499219-3499219 on Build GRCh38
rs375736464 Chr.17: 3499013-3499013 on Build GRCh38
rs766720790 Chr.17: 3499016-3499019 on Build GRCh38
rs759106494 Chr.17: 3498953-3498953 on Build GRCh38
rs771525119 Chr.17: 3499247-3499247 on Build GRCh38
rs750991168 Chr.17: 3499058-3499058 on Build GRCh38
rs9896885 Chr.17: 3499091-3499091 on Build GRCh38
rs765810524 Chr.17: 3498950-3498950 on Build GRCh38
rs757610508 Chr.17: 3499146-3499146 on Build GRCh38
rs141858640 Chr.17: 3498987-3498987 on Build GRCh38
rs765729393 Chr.17: 3499132-3499132 on Build GRCh38
rs763869261 Chr.17: 3499051-3499051 on Build GRCh38
rs376854191 Chr.17: 3499074-3499074 on Build GRCh38
rs747218271 Chr.17: 3498989-3498989 on Build GRCh38
rs746496601 Chr.17: 3498856-3498860 on Build GRCh38
rs768368098 Chr.17: 3499268-3499268 on Build GRCh38
rs778121276 Chr.17: 3499072-3499072 on Build GRCh38
rs1552830 Chr.17: 3499227-3499227 on Build GRCh38
rs200126822 Chr.17: 3498955-3498955 on Build GRCh38
rs373410075 Chr.17: 3498899-3498899 on Build GRCh38
rs371394875 Chr.17: 3499052-3499052 on Build GRCh38
rs1009128846 Chr.17: 3498827-3498827 on Build GRCh38
rs773849106 Chr.17: 3498952-3498952 on Build GRCh38
rs768121711 Chr.17: 3498864-3498864 on Build GRCh38
rs747943040 Chr.17: 3498915-3498915 on Build GRCh38
rs775443252 Chr.17: 3498847-3498848 on Build GRCh38
rs866693885 Chr.17: 3498805-3498805 on Build GRCh38
rs762310899 Chr.17: 3499055-3499055 on Build GRCh38
rs890549402 Chr.17: 3499261-3499261 on Build GRCh38
rs555537960 Chr.17: 3499068-3499068 on Build GRCh38
rs745832703 Chr.17: 3499087-3499087 on Build GRCh38
rs761390319 Chr.17: 3498942-3498946 on Build GRCh38
rs952434769 Chr.17: 3498875-3498875 on Build GRCh38
rs939271414 Chr.17: 3499231-3499231 on Build GRCh38
rs1057517260 Chr.17: 3499070-3499070 on Build GRCh38
rs762221059 Chr.17: 3499117-3499117 on Build GRCh38
rs914573514 Chr.17: 3499211-3499211 on Build GRCh38
rs776474192 Chr.17: 3499116-3499116 on Build GRCh38
rs954697326 Chr.17: 3498798-3498798 on Build GRCh38
rs990214187 Chr.17: 3499189-3499189 on Build GRCh38
rs559875524 Chr.17: 3499250-3499250 on Build GRCh38
rs151133430 Chr.17: 3499115-3499115 on Build GRCh38
rs28940279 Chr.17: 3499000-3499000 on Build GRCh38
rs557594000 Chr.17: 3499105-3499105 on Build GRCh38
rs749148738 Chr.17: 3499001-3499001 on Build GRCh38
rs140581464 Chr.17: 3498978-3498978 on Build GRCh38
rs377217076 Chr.17: 3498916-3498916 on Build GRCh38
rs528972033 Chr.17: 3499221-3499221 on Build GRCh38
rs779350543 Chr.17: 3499155-3499155 on Build GRCh38
rs974368086 Chr.17: 3499161-3499161 on Build GRCh38
rs373219949 Chr.17: 3499099-3499099 on Build GRCh38
rs104894552 Chr.17: 3498892-3498892 on Build GRCh38
rs775163203 Chr.17: 3499041-3499041 on Build GRCh38
rs774852992 Chr.17: 3499102-3499102 on Build GRCh38
rs1057516309 Chr.17: 3499068-3499068 on Build GRCh38
rs1008846130 Chr.17: 3499050-3499050 on Build GRCh38
rs1020492620 Chr.17: 3498865-3498865 on Build GRCh38
rs749049390 Chr.17: 3499073-3499073 on Build GRCh38
rs138062143 Chr.17: 3499004-3499004 on Build GRCh38

View Full Product Details