Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4559468-4559960 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GGT6
Entrez Gene ID
124975
Gene Name
gamma-glutamyltransferase 6
Gene Aliases
-
UniGene
Hs.130749
Gene Chromosome Location
Chr.17: 4556927-4560596 on Build GRCh38
Transcripts
NM_001122890.2 NM_001288702.1 NM_001288703.1 NM_001288704.1 NM_153338.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4950460 c.275C>T p.A92V Chr.17: 4559626-4559626 on Build GRCh38
6588761 c.175C>G p.R59G Chr.17: 4559726-4559726 on Build GRCh38
4686886 c.156G>A p.G52G Chr.17: 4559745-4559745 on Build GRCh38
3518408 c.330C>T p.I110I Chr.17: 4559571-4559571 on Build GRCh38
5363826 c.141-8delC p.? Chr.17: 4559768-4559768 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775016920 Chr.17: 4559491-4559491 on Build GRCh38
rs773604892 Chr.17: 4559542-4559542 on Build GRCh38
rs907637094 Chr.17: 4559925-4559925 on Build GRCh38
rs549836107 Chr.17: 4559888-4559888 on Build GRCh38
rs377439398 Chr.17: 4559492-4559492 on Build GRCh38
rs777268961 Chr.17: 4559660-4559660 on Build GRCh38
rs775757530 Chr.17: 4559767-4559768 on Build GRCh38
rs145923439 Chr.17: 4559649-4559649 on Build GRCh38
rs766277687 Chr.17: 4559585-4559585 on Build GRCh38
rs754670720 Chr.17: 4559513-4559513 on Build GRCh38
rs142207873 Chr.17: 4559600-4559600 on Build GRCh38
rs930418779 Chr.17: 4559685-4559685 on Build GRCh38
rs377047398 Chr.17: 4559571-4559571 on Build GRCh38
rs765028133 Chr.17: 4559576-4559576 on Build GRCh38
rs780219052 Chr.17: 4559686-4559686 on Build GRCh38
rs148111494 Chr.17: 4559580-4559580 on Build GRCh38
rs753736237 Chr.17: 4559489-4559489 on Build GRCh38
rs1011866252 Chr.17: 4559791-4559791 on Build GRCh38
rs760937160 Chr.17: 4559768-4559768 on Build GRCh38
rs11657054 Chr.17: 4559728-4559728 on Build GRCh38
rs776710223 Chr.17: 4559726-4559726 on Build GRCh38
rs747034693 Chr.17: 4559713-4559713 on Build GRCh38
rs774614008 Chr.17: 4559553-4559553 on Build GRCh38
rs367928800 Chr.17: 4559615-4559615 on Build GRCh38
rs1015737774 Chr.17: 4559822-4559822 on Build GRCh38
rs749586621 Chr.17: 4559604-4559604 on Build GRCh38
rs768975988 Chr.17: 4559725-4559725 on Build GRCh38
rs748528438 Chr.17: 4559518-4559518 on Build GRCh38
rs541178788 Chr.17: 4559826-4559826 on Build GRCh38
rs983496185 Chr.17: 4559864-4559864 on Build GRCh38
rs760239453 Chr.17: 4559806-4559806 on Build GRCh38
rs1047478589 Chr.17: 4559714-4559714 on Build GRCh38
rs751124076 Chr.17: 4559591-4559591 on Build GRCh38
rs370479021 Chr.17: 4559625-4559625 on Build GRCh38
rs779375707 Chr.17: 4559767-4559767 on Build GRCh38
rs752324097 Chr.17: 4559652-4559652 on Build GRCh38
rs970470774 Chr.17: 4559807-4559807 on Build GRCh38
rs759304747 Chr.17: 4559775-4559775 on Build GRCh38
rs564167421 Chr.17: 4559831-4559831 on Build GRCh38
rs751430836 Chr.17: 4559637-4559637 on Build GRCh38
rs951756382 Chr.17: 4559835-4559835 on Build GRCh38
rs755832863 Chr.17: 4559658-4559658 on Build GRCh38
rs766367569 Chr.17: 4559632-4559632 on Build GRCh38
rs771168466 Chr.17: 4559547-4559547 on Build GRCh38
rs780408274 Chr.17: 4559769-4559769 on Build GRCh38
rs912645326 Chr.17: 4559906-4559906 on Build GRCh38
rs562529644 Chr.17: 4559939-4559939 on Build GRCh38
rs748319913 Chr.17: 4559772-4559772 on Build GRCh38
rs759644525 Chr.17: 4559545-4559545 on Build GRCh38
rs183838731 Chr.17: 4559852-4559852 on Build GRCh38
rs780808194 Chr.17: 4559514-4559514 on Build GRCh38
rs750355595 Chr.17: 4559579-4559579 on Build GRCh38
rs754286886 Chr.17: 4559661-4559661 on Build GRCh38
rs371984536 Chr.17: 4559539-4559539 on Build GRCh38
rs373030143 Chr.17: 4559738-4559738 on Build GRCh38
rs985469103 Chr.17: 4559903-4559903 on Build GRCh38
rs763875416 Chr.17: 4559751-4559751 on Build GRCh38
rs757065791 Chr.17: 4559491-4559491 on Build GRCh38
rs778161148 Chr.17: 4559601-4559601 on Build GRCh38
rs779496692 Chr.17: 4559494-4559494 on Build GRCh38
rs757711515 Chr.17: 4559763-4559763 on Build GRCh38
rs780826617 Chr.17: 4559596-4559596 on Build GRCh38
rs772245514 Chr.17: 4559667-4559668 on Build GRCh38
rs886270920 Chr.17: 4559506-4559506 on Build GRCh38
rs773106199 Chr.17: 4559629-4559629 on Build GRCh38
rs944114480 Chr.17: 4559924-4559924 on Build GRCh38
rs920250853 Chr.17: 4559682-4559682 on Build GRCh38
rs779571047 Chr.17: 4559669-4559669 on Build GRCh38
rs775286737 Chr.17: 4559796-4559796 on Build GRCh38
rs774986374 Chr.17: 4559612-4559612 on Build GRCh38
rs556101605 Chr.17: 4559496-4559496 on Build GRCh38
rs749756369 Chr.17: 4559546-4559546 on Build GRCh38
rs902448379 Chr.17: 4559757-4559757 on Build GRCh38
rs752301690 Chr.17: 4559748-4559748 on Build GRCh38
rs762719587 Chr.17: 4559631-4559631 on Build GRCh38
rs754480140 Chr.17: 4559592-4559592 on Build GRCh38
rs954064362 Chr.17: 4559894-4559894 on Build GRCh38
rs146789997 Chr.17: 4559736-4559736 on Build GRCh38
rs1056679593 Chr.17: 4559732-4559732 on Build GRCh38
rs772303187 Chr.17: 4559681-4559681 on Build GRCh38
rs767002789 Chr.17: 4559650-4559650 on Build GRCh38
rs757695092 Chr.17: 4559666-4559666 on Build GRCh38
rs961635303 Chr.17: 4559823-4559823 on Build GRCh38
rs149456893 Chr.17: 4559739-4559739 on Build GRCh38
rs753477664 Chr.17: 4559761-4559761 on Build GRCh38
rs374352398 Chr.17: 4559613-4559613 on Build GRCh38
rs1021327581 Chr.17: 4559599-4559599 on Build GRCh38
rs775591313 Chr.17: 4559616-4559616 on Build GRCh38
rs759666966 Chr.17: 4559561-4559561 on Build GRCh38
rs890980683 Chr.17: 4559541-4559541 on Build GRCh38
rs772108116 Chr.17: 4559787-4559787 on Build GRCh38
rs759427541 Chr.17: 4559740-4559740 on Build GRCh38
rs751001528 Chr.17: 4559768-4559768 on Build GRCh38
rs764287843 Chr.17: 4559795-4559796 on Build GRCh38
rs572173915 Chr.17: 4559774-4559774 on Build GRCh38
rs142643623 Chr.17: 4559575-4559575 on Build GRCh38
rs771616002 Chr.17: 4559607-4559607 on Build GRCh38
rs1018441629 Chr.17: 4559783-4559783 on Build GRCh38
rs768549067 Chr.17: 4559770-4559770 on Build GRCh38
rs781489485 Chr.17: 4559771-4559771 on Build GRCh38
rs772922247 Chr.17: 4559747-4559747 on Build GRCh38
rs767912501 Chr.17: 4559564-4559564 on Build GRCh38

View Full Product Details