Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4672712-4673224 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PELP1
Entrez Gene ID
27043
Gene Name
proline, glutamate and leucine rich protein 1
Gene Aliases
MNAR, P160
UniGene
Hs.744899
Gene Chromosome Location
Chr.17: 4671384-4704337 on Build GRCh38
Transcripts
NM_001278241.1 NM_014389.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4632153 c.2646C>T p.P882P Chr.17: 4672777-4672777 on Build GRCh38
7169568 c.2204C>T p.S735L Chr.17: 4672937-4672937 on Build GRCh38
4067268 c.2424G>A p.P808P Chr.17: 4672999-4672999 on Build GRCh38
3518580 c.2381C>T p.P794L Chr.17: 4673042-4673042 on Build GRCh38
6894325 c.2638G>A p.E880K Chr.17: 4672785-4672785 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200062536 Chr.17: 4672980-4672980 on Build GRCh38
rs749062848 Chr.17: 4673039-4673039 on Build GRCh38
rs745916555 Chr.17: 4672958-4672958 on Build GRCh38
rs756319868 Chr.17: 4672948-4672965 on Build GRCh38
rs781554929 Chr.17: 4672800-4672800 on Build GRCh38
rs866178242 Chr.17: 4672833-4672833 on Build GRCh38
rs922844974 Chr.17: 4672889-4672889 on Build GRCh38
rs760238191 Chr.17: 4672992-4672992 on Build GRCh38
rs566672712 Chr.17: 4673037-4673037 on Build GRCh38
rs759121133 Chr.17: 4672987-4672987 on Build GRCh38
rs770325846 Chr.17: 4672809-4672809 on Build GRCh38
rs761698628 Chr.17: 4672896-4672896 on Build GRCh38
rs372080166 Chr.17: 4672926-4672926 on Build GRCh38
rs751103310 Chr.17: 4672915-4672915 on Build GRCh38
rs754406795 Chr.17: 4672949-4672949 on Build GRCh38
rs1003370912 Chr.17: 4673014-4673014 on Build GRCh38
rs961726343 Chr.17: 4672779-4672779 on Build GRCh38
rs868239336 Chr.17: 4672908-4672908 on Build GRCh38
rs760421299 Chr.17: 4673145-4673145 on Build GRCh38
rs140079854 Chr.17: 4673188-4673188 on Build GRCh38
rs548162502 Chr.17: 4672969-4672969 on Build GRCh38
rs759684711 Chr.17: 4672836-4672836 on Build GRCh38
rs559141289 Chr.17: 4673148-4673148 on Build GRCh38
rs113477436 Chr.17: 4673064-4673064 on Build GRCh38
rs376160582 Chr.17: 4672814-4672814 on Build GRCh38
rs778556653 Chr.17: 4672910-4672927 on Build GRCh38
rs764712338 Chr.17: 4672768-4672768 on Build GRCh38
rs750844787 Chr.17: 4673017-4673017 on Build GRCh38
rs765188029 Chr.17: 4672848-4672848 on Build GRCh38
rs752154570 Chr.17: 4672957-4672957 on Build GRCh38
rs910296508 Chr.17: 4673084-4673084 on Build GRCh38
rs749925559 Chr.17: 4672770-4672770 on Build GRCh38
rs368444449 Chr.17: 4672871-4672871 on Build GRCh38
rs763366335 Chr.17: 4673184-4673184 on Build GRCh38
rs769427960 Chr.17: 4672898-4672898 on Build GRCh38
rs774512776 Chr.17: 4672743-4672743 on Build GRCh38
rs529623797 Chr.17: 4672745-4672745 on Build GRCh38
rs772314522 Chr.17: 4672892-4672892 on Build GRCh38
rs776472829 Chr.17: 4672840-4672840 on Build GRCh38
rs766876710 Chr.17: 4672988-4672988 on Build GRCh38
rs1046199362 Chr.17: 4673091-4673091 on Build GRCh38
rs759881968 Chr.17: 4673170-4673171 on Build GRCh38
rs532240356 Chr.17: 4672878-4672878 on Build GRCh38
rs752256184 Chr.17: 4672929-4672929 on Build GRCh38
rs982460213 Chr.17: 4672737-4672737 on Build GRCh38
rs757747506 Chr.17: 4672953-4672953 on Build GRCh38
rs766662075 Chr.17: 4672953-4672970 on Build GRCh38
rs758163168 Chr.17: 4672774-4672774 on Build GRCh38
rs758520016 Chr.17: 4673020-4673020 on Build GRCh38
rs868431825 Chr.17: 4673035-4673035 on Build GRCh38
rs775208212 Chr.17: 4672990-4672990 on Build GRCh38
rs765528664 Chr.17: 4673173-4673173 on Build GRCh38
rs893799341 Chr.17: 4673187-4673187 on Build GRCh38
rs910218414 Chr.17: 4673049-4673049 on Build GRCh38
rs773295937 Chr.17: 4672900-4672900 on Build GRCh38
rs375481592 Chr.17: 4673099-4673099 on Build GRCh38
rs779932015 Chr.17: 4673025-4673025 on Build GRCh38
rs560242080 Chr.17: 4672830-4672830 on Build GRCh38
rs993347563 Chr.17: 4673033-4673033 on Build GRCh38
rs575727361 Chr.17: 4673170-4673170 on Build GRCh38
rs531659496 Chr.17: 4672944-4672944 on Build GRCh38
rs771675044 Chr.17: 4673095-4673095 on Build GRCh38
rs759162666 Chr.17: 4672916-4672916 on Build GRCh38
rs746144990 Chr.17: 4672887-4672887 on Build GRCh38
rs377113067 Chr.17: 4672742-4672742 on Build GRCh38
rs920228740 Chr.17: 4672801-4672801 on Build GRCh38
rs780347388 Chr.17: 4672960-4672960 on Build GRCh38
rs761703296 Chr.17: 4672847-4672847 on Build GRCh38
rs567669309 Chr.17: 4672970-4672970 on Build GRCh38
rs763887774 Chr.17: 4672748-4672748 on Build GRCh38
rs778957081 Chr.17: 4672886-4672886 on Build GRCh38
rs938068817 Chr.17: 4673125-4673125 on Build GRCh38
rs770813119 Chr.17: 4673065-4673065 on Build GRCh38
rs747332132 Chr.17: 4672910-4672945 on Build GRCh38
rs779885967 Chr.17: 4672777-4672777 on Build GRCh38
rs749338627 Chr.17: 4672826-4672826 on Build GRCh38
rs758369823 Chr.17: 4672959-4672959 on Build GRCh38
rs147763003 Chr.17: 4672921-4672921 on Build GRCh38
rs750346544 Chr.17: 4672850-4672850 on Build GRCh38
rs748673923 Chr.17: 4672802-4672802 on Build GRCh38
rs764497135 Chr.17: 4672996-4672996 on Build GRCh38
rs770972452 Chr.17: 4672978-4672978 on Build GRCh38
rs776226691 Chr.17: 4673163-4673163 on Build GRCh38
rs935581314 Chr.17: 4673164-4673164 on Build GRCh38
rs766568560 Chr.17: 4672739-4672739 on Build GRCh38
rs770272132 Chr.17: 4672933-4672934 on Build GRCh38
rs915267975 Chr.17: 4673200-4673200 on Build GRCh38
rs919201701 Chr.17: 4673076-4673076 on Build GRCh38
rs922730427 Chr.17: 4673109-4673109 on Build GRCh38
rs370376466 Chr.17: 4672894-4672894 on Build GRCh38
rs967267031 Chr.17: 4672999-4672999 on Build GRCh38
rs751262670 Chr.17: 4672864-4672864 on Build GRCh38
rs778363267 Chr.17: 4672872-4672872 on Build GRCh38
rs768977236 Chr.17: 4672921-4672922 on Build GRCh38
rs961943979 Chr.17: 4673024-4673024 on Build GRCh38
rs906365531 Chr.17: 4673118-4673118 on Build GRCh38
rs751209564 Chr.17: 4672796-4672796 on Build GRCh38
rs753719637 Chr.17: 4672754-4672754 on Build GRCh38
rs774456693 Chr.17: 4673067-4673067 on Build GRCh38
rs756510631 Chr.17: 4672931-4672931 on Build GRCh38
rs954452018 Chr.17: 4673023-4673023 on Build GRCh38
rs202062234 Chr.17: 4672799-4672799 on Build GRCh38
rs781470306 Chr.17: 4672867-4672867 on Build GRCh38
rs765763814 Chr.17: 4673010-4673010 on Build GRCh38
rs558960670 Chr.17: 4672901-4672901 on Build GRCh38
rs375701025 Chr.17: 4672939-4672939 on Build GRCh38
rs755507936 Chr.17: 4672965-4672965 on Build GRCh38
rs765929440 Chr.17: 4672858-4672858 on Build GRCh38
rs958032388 Chr.17: 4672946-4672946 on Build GRCh38
rs200607294 Chr.17: 4673000-4673000 on Build GRCh38
rs763220675 Chr.17: 4672736-4672736 on Build GRCh38
rs750944376 Chr.17: 4673182-4673182 on Build GRCh38
rs754658068 Chr.17: 4672865-4672865 on Build GRCh38
rs751865342 Chr.17: 4673088-4673089 on Build GRCh38
rs757213322 Chr.17: 4672758-4672758 on Build GRCh38
rs757798889 Chr.17: 4673008-4673008 on Build GRCh38
rs750760710 Chr.17: 4672909-4672909 on Build GRCh38
rs941889300 Chr.17: 4672860-4672860 on Build GRCh38
rs776909062 Chr.17: 4672922-4672939 on Build GRCh38
rs753088632 Chr.17: 4672868-4672868 on Build GRCh38
rs755239224 Chr.17: 4673036-4673036 on Build GRCh38
rs773277043 Chr.17: 4672939-4672940 on Build GRCh38
rs745722648 Chr.17: 4673090-4673090 on Build GRCh38
rs929235668 Chr.17: 4673089-4673089 on Build GRCh38
rs762576153 Chr.17: 4672854-4672854 on Build GRCh38
rs766054972 Chr.17: 4672913-4672913 on Build GRCh38
rs748093664 Chr.17: 4673074-4673074 on Build GRCh38
rs973305453 Chr.17: 4673071-4673071 on Build GRCh38
rs768401873 Chr.17: 4672838-4672838 on Build GRCh38
rs1027297007 Chr.17: 4673066-4673066 on Build GRCh38
rs763140306 Chr.17: 4672940-4672957 on Build GRCh38
rs749300100 Chr.17: 4672973-4672973 on Build GRCh38

View Full Product Details