Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4940803-4941302 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF167
Entrez Gene ID
26001
Gene Name
ring finger protein 167
Gene Aliases
5730408C10Rik, LP2254, RING105
UniGene
Hs.7158
Gene Chromosome Location
Chr.17: 4940008-4945223 on Build GRCh38
Transcripts
NM_001320356.1 NM_001320357.1 NM_001320358.1 NM_001320359.1 NM_001320360.1 NM_001320361.1 NM_001320362.1 NM_001320363.1 NM_001320364.1 NM_001320365.1 NM_015528.2 XM_017024427.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7387716 c.118G>T p.D40Y Chr.17: 4941110-4941110 on Build GRCh38
5420801 c.144C>T p.T48T Chr.17: 4941136-4941136 on Build GRCh38
7402456 c.1A>G p.M1V Chr.17: 4940910-4940910 on Build GRCh38
1191734 c.62C>A p.P21Q Chr.17: 4940971-4940971 on Build GRCh38
4834091 c.147G>A p.L49L Chr.17: 4941139-4941139 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772733360 Chr.17: 4941197-4941197 on Build GRCh38
rs755828116 Chr.17: 4940917-4940917 on Build GRCh38
rs1051775967 Chr.17: 4940990-4940990 on Build GRCh38
rs537507611 Chr.17: 4941208-4941208 on Build GRCh38
rs772142671 Chr.17: 4940884-4940884 on Build GRCh38
rs759447136 Chr.17: 4941009-4941009 on Build GRCh38
rs927872055 Chr.17: 4941228-4941228 on Build GRCh38
rs751136689 Chr.17: 4941164-4941166 on Build GRCh38
rs1051864937 Chr.17: 4941201-4941201 on Build GRCh38
rs780859215 Chr.17: 4941195-4941196 on Build GRCh38
rs767018823 Chr.17: 4941146-4941146 on Build GRCh38
rs34430513 Chr.17: 4940947-4940947 on Build GRCh38
rs753245742 Chr.17: 4941041-4941041 on Build GRCh38
rs765799830 Chr.17: 4941163-4941166 on Build GRCh38
rs369142735 Chr.17: 4940885-4940885 on Build GRCh38
rs946148643 Chr.17: 4941210-4941211 on Build GRCh38
rs773228461 Chr.17: 4941095-4941095 on Build GRCh38
rs145210225 Chr.17: 4941085-4941085 on Build GRCh38
rs745782809 Chr.17: 4941097-4941097 on Build GRCh38
rs182848539 Chr.17: 4941166-4941166 on Build GRCh38
rs1010679678 Chr.17: 4941016-4941016 on Build GRCh38
rs775215703 Chr.17: 4941106-4941106 on Build GRCh38
rs11548909 Chr.17: 4940845-4940845 on Build GRCh38
rs563205605 Chr.17: 4941175-4941175 on Build GRCh38
rs867600313 Chr.17: 4940929-4940929 on Build GRCh38
rs890519845 Chr.17: 4941013-4941013 on Build GRCh38
rs751935165 Chr.17: 4941154-4941154 on Build GRCh38
rs937978989 Chr.17: 4941253-4941253 on Build GRCh38
rs749941210 Chr.17: 4941135-4941135 on Build GRCh38
rs528847193 Chr.17: 4940890-4940890 on Build GRCh38
rs549962523 Chr.17: 4940936-4940936 on Build GRCh38
rs758896165 Chr.17: 4941055-4941055 on Build GRCh38
rs560001343 Chr.17: 4941218-4941219 on Build GRCh38
rs776917805 Chr.17: 4940896-4940896 on Build GRCh38
rs919067074 Chr.17: 4941198-4941198 on Build GRCh38
rs780624527 Chr.17: 4940955-4940955 on Build GRCh38
rs200305741 Chr.17: 4941042-4941042 on Build GRCh38
rs565177578 Chr.17: 4940898-4940898 on Build GRCh38
rs199653542 Chr.17: 4941050-4941050 on Build GRCh38
rs909511815 Chr.17: 4941178-4941178 on Build GRCh38
rs776359072 Chr.17: 4941184-4941184 on Build GRCh38
rs1034244722 Chr.17: 4941233-4941233 on Build GRCh38
rs922574012 Chr.17: 4940826-4940826 on Build GRCh38
rs761663167 Chr.17: 4940894-4940895 on Build GRCh38
rs753080925 Chr.17: 4940830-4940830 on Build GRCh38
rs778274211 Chr.17: 4941123-4941126 on Build GRCh38
rs1021118055 Chr.17: 4941053-4941053 on Build GRCh38
rs149864707 Chr.17: 4941074-4941074 on Build GRCh38
rs757654757 Chr.17: 4940934-4940934 on Build GRCh38
rs1046314808 Chr.17: 4940952-4940952 on Build GRCh38
rs751266201 Chr.17: 4940908-4940908 on Build GRCh38
rs755206343 Chr.17: 4941082-4941082 on Build GRCh38
rs866146808 Chr.17: 4941156-4941156 on Build GRCh38
rs894383788 Chr.17: 4941278-4941278 on Build GRCh38
rs762586495 Chr.17: 4941202-4941202 on Build GRCh38
rs772046058 Chr.17: 4941098-4941098 on Build GRCh38
rs145789550 Chr.17: 4941183-4941183 on Build GRCh38
rs539931954 Chr.17: 4940935-4940935 on Build GRCh38
rs539737019 Chr.17: 4941236-4941236 on Build GRCh38
rs763794898 Chr.17: 4940872-4940872 on Build GRCh38
rs761496408 Chr.17: 4941187-4941187 on Build GRCh38
rs747231088 Chr.17: 4940964-4940964 on Build GRCh38
rs748111891 Chr.17: 4941090-4941090 on Build GRCh38
rs761978619 Chr.17: 4940976-4940976 on Build GRCh38
rs779123468 Chr.17: 4940859-4940859 on Build GRCh38
rs746264966 Chr.17: 4940871-4940871 on Build GRCh38
rs774356298 Chr.17: 4940981-4940981 on Build GRCh38
rs190591845 Chr.17: 4941142-4941142 on Build GRCh38
rs577524032 Chr.17: 4941113-4941113 on Build GRCh38
rs767017587 Chr.17: 4940912-4940912 on Build GRCh38
rs752525089 Chr.17: 4941025-4941025 on Build GRCh38
rs760298093 Chr.17: 4941033-4941033 on Build GRCh38
rs763668375 Chr.17: 4941118-4941118 on Build GRCh38
rs761431506 Chr.17: 4941132-4941132 on Build GRCh38
rs981959765 Chr.17: 4941224-4941224 on Build GRCh38
rs1055188467 Chr.17: 4941259-4941259 on Build GRCh38
rs762955694 Chr.17: 4941161-4941162 on Build GRCh38
rs777882252 Chr.17: 4941173-4941173 on Build GRCh38
rs775861531 Chr.17: 4940892-4940892 on Build GRCh38
rs760779651 Chr.17: 4940894-4940894 on Build GRCh38
rs753394485 Chr.17: 4940928-4940928 on Build GRCh38
rs756928778 Chr.17: 4941195-4941195 on Build GRCh38
rs187752946 Chr.17: 4941252-4941252 on Build GRCh38
rs777233679 Chr.17: 4940924-4940924 on Build GRCh38
rs770736329 Chr.17: 4940980-4940980 on Build GRCh38
rs781726560 Chr.17: 4941162-4941162 on Build GRCh38
rs752329246 Chr.17: 4940915-4940915 on Build GRCh38
rs769044376 Chr.17: 4940974-4940974 on Build GRCh38
rs139697970 Chr.17: 4941078-4941078 on Build GRCh38
rs768844064 Chr.17: 4940895-4940895 on Build GRCh38
rs764800755 Chr.17: 4941134-4941134 on Build GRCh38
rs150319818 Chr.17: 4941192-4941195 on Build GRCh38
rs766391792 Chr.17: 4940903-4940903 on Build GRCh38
rs929152691 Chr.17: 4941216-4941216 on Build GRCh38
rs749896816 Chr.17: 4941267-4941267 on Build GRCh38
rs973654308 Chr.17: 4941192-4941192 on Build GRCh38
rs1037488624 Chr.17: 4940923-4940923 on Build GRCh38
rs186588602 Chr.17: 4941051-4941051 on Build GRCh38
rs769902541 Chr.17: 4941093-4941093 on Build GRCh38
rs937680142 Chr.17: 4941008-4941008 on Build GRCh38
rs759409703 Chr.17: 4940909-4940909 on Build GRCh38
rs752377798 Chr.17: 4941205-4941208 on Build GRCh38
rs764863432 Chr.17: 4941206-4941206 on Build GRCh38
rs141088909 Chr.17: 4941035-4941035 on Build GRCh38
rs775410136 Chr.17: 4941180-4941180 on Build GRCh38
rs764497812 Chr.17: 4940835-4940835 on Build GRCh38
rs776361019 Chr.17: 4941124-4941124 on Build GRCh38
rs767292101 Chr.17: 4941014-4941014 on Build GRCh38
rs765149644 Chr.17: 4941084-4941086 on Build GRCh38

View Full Product Details