Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 27819529-27820032 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STX12
Entrez Gene ID
23673
Gene Name
syntaxin 12
Gene Aliases
STX13, STX14
UniGene
Hs.523855
Gene Chromosome Location
Chr.1: 27773183-27824452 on Build GRCh38
Transcripts
NM_177424.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7318925 c.732G>C p.Q244H Chr.1: 27819732-27819732 on Build GRCh38
907917 c.653G>T p.S218I Chr.1: 27819653-27819653 on Build GRCh38
6221836 c.693A>C p.E231D Chr.1: 27819693-27819693 on Build GRCh38
5483177 c.700A>G p.T234A Chr.1: 27819700-27819700 on Build GRCh38
5621940 c.683T>C p.V228A Chr.1: 27819683-27819683 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774987462 Chr.1: 27819620-27819620 on Build GRCh38
rs983657163 Chr.1: 27819848-27819848 on Build GRCh38
rs115867877 Chr.1: 27819966-27819966 on Build GRCh38
rs757788037 Chr.1: 27819673-27819673 on Build GRCh38
rs755681038 Chr.1: 27819716-27819716 on Build GRCh38
rs767253924 Chr.1: 27819743-27819743 on Build GRCh38
rs904718028 Chr.1: 27819985-27819985 on Build GRCh38
rs1021795049 Chr.1: 27819770-27819770 on Build GRCh38
rs757735639 Chr.1: 27819762-27819763 on Build GRCh38
rs929243191 Chr.1: 27819845-27819845 on Build GRCh38
rs892001829 Chr.1: 27819964-27819964 on Build GRCh38
rs1001609133 Chr.1: 27820002-27820002 on Build GRCh38
rs774011676 Chr.1: 27819740-27819740 on Build GRCh38
rs941759446 Chr.1: 27819874-27819874 on Build GRCh38
rs1038646435 Chr.1: 27819674-27819674 on Build GRCh38
rs34528653 Chr.1: 27819956-27819957 on Build GRCh38
rs116417958 Chr.1: 27819913-27819913 on Build GRCh38
rs556505958 Chr.1: 27819971-27819971 on Build GRCh38
rs145114259 Chr.1: 27819688-27819688 on Build GRCh38
rs527262415 Chr.1: 27819568-27819568 on Build GRCh38
rs17162817 Chr.1: 27819795-27819795 on Build GRCh38
rs917866730 Chr.1: 27819837-27819837 on Build GRCh38
rs770959374 Chr.1: 27819733-27819733 on Build GRCh38
rs887490248 Chr.1: 27819592-27819592 on Build GRCh38
rs558162440 Chr.1: 27819894-27819894 on Build GRCh38
rs559915110 Chr.1: 27819748-27819748 on Build GRCh38
rs747252142 Chr.1: 27819695-27819695 on Build GRCh38
rs764512723 Chr.1: 27819597-27819597 on Build GRCh38
rs550102857 Chr.1: 27819690-27819690 on Build GRCh38
rs968762536 Chr.1: 27819781-27819781 on Build GRCh38
rs370700701 Chr.1: 27819783-27819783 on Build GRCh38
rs549183964 Chr.1: 27819794-27819794 on Build GRCh38
rs909062315 Chr.1: 27819859-27819859 on Build GRCh38
rs759442277 Chr.1: 27819741-27819741 on Build GRCh38
rs1052016940 Chr.1: 27819963-27819963 on Build GRCh38
rs900163620 Chr.1: 27819924-27819924 on Build GRCh38
rs755731936 Chr.1: 27819653-27819653 on Build GRCh38
rs569877251 Chr.1: 27819691-27819691 on Build GRCh38
rs746355894 Chr.1: 27819807-27819807 on Build GRCh38
rs900194639 Chr.1: 27819675-27819675 on Build GRCh38
rs879159687 Chr.1: 27819994-27819994 on Build GRCh38
rs562871559 Chr.1: 27819814-27819814 on Build GRCh38
rs905794603 Chr.1: 27819601-27819601 on Build GRCh38
rs1021779875 Chr.1: 27819970-27819970 on Build GRCh38
rs1001918095 Chr.1: 27819622-27819622 on Build GRCh38
rs143032204 Chr.1: 27819723-27819723 on Build GRCh38
rs570146523 Chr.1: 27819687-27819687 on Build GRCh38
rs17162815 Chr.1: 27819582-27819582 on Build GRCh38
rs768679047 Chr.1: 27819712-27819712 on Build GRCh38
rs536052325 Chr.1: 27819752-27819752 on Build GRCh38
rs933012672 Chr.1: 27819951-27819951 on Build GRCh38
rs77275809 Chr.1: 27819647-27819647 on Build GRCh38
rs762212047 Chr.1: 27819655-27819655 on Build GRCh38
rs752223005 Chr.1: 27819609-27819609 on Build GRCh38
rs373301735 Chr.1: 27819683-27819683 on Build GRCh38
rs749719506 Chr.1: 27819664-27819664 on Build GRCh38

View Full Product Details