Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 28668632-28669157 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GMEB1
Entrez Gene ID
10691
Gene Name
glucocorticoid modulatory element binding protein 1
Gene Aliases
P96PIF, PIF96
UniGene
Hs.632373
Gene Chromosome Location
Chr.1: 28668728-28715603 on Build GRCh38
Transcripts
NM_001319674.1 NM_006582.3 NM_024482.2 XM_011540519.2 XM_011540522.2

SNP Detail

SNP ID Chromosome Location
rs925276575 Chr.1: 28668764-28668764 on Build GRCh38
rs1057464102 Chr.1: 28668857-28668857 on Build GRCh38
rs1049001915 Chr.1: 28669040-28669040 on Build GRCh38
rs960887934 Chr.1: 28668948-28668948 on Build GRCh38
rs931846274 Chr.1: 28669128-28669128 on Build GRCh38
rs996059392 Chr.1: 28668888-28668888 on Build GRCh38
rs908033990 Chr.1: 28668737-28668737 on Build GRCh38
rs925556691 Chr.1: 28668676-28668676 on Build GRCh38
rs974358638 Chr.1: 28668707-28668708 on Build GRCh38
rs944083952 Chr.1: 28669106-28669106 on Build GRCh38
rs1013370834 Chr.1: 28668881-28668881 on Build GRCh38
rs1000417269 Chr.1: 28668901-28668901 on Build GRCh38
rs900400528 Chr.1: 28668879-28668879 on Build GRCh38
rs1015117821 Chr.1: 28668947-28668948 on Build GRCh38
rs925548100 Chr.1: 28669075-28669075 on Build GRCh38
rs1031860148 Chr.1: 28668982-28668982 on Build GRCh38
rs866405915 Chr.1: 28668706-28668706 on Build GRCh38
rs1027292354 Chr.1: 28668711-28668711 on Build GRCh38
rs1043712761 Chr.1: 28668759-28668759 on Build GRCh38
rs925263727 Chr.1: 28668977-28668977 on Build GRCh38
rs865923717 Chr.1: 28669107-28669107 on Build GRCh38
rs1009168801 Chr.1: 28668794-28668794 on Build GRCh38
rs951754119 Chr.1: 28668940-28668940 on Build GRCh38
rs529722900 Chr.1: 28668740-28668740 on Build GRCh38
rs983609700 Chr.1: 28668736-28668736 on Build GRCh38
rs912574043 Chr.1: 28669095-28669095 on Build GRCh38
rs536054274 Chr.1: 28668672-28668672 on Build GRCh38
rs900349573 Chr.1: 28669037-28669037 on Build GRCh38
rs1004980612 Chr.1: 28669047-28669047 on Build GRCh38
rs1013338940 Chr.1: 28669060-28669060 on Build GRCh38
rs992705374 Chr.1: 28668948-28668948 on Build GRCh38
rs887431262 Chr.1: 28669044-28669044 on Build GRCh38
rs878922434 Chr.1: 28668773-28668773 on Build GRCh38
rs969589246 Chr.1: 28668953-28668953 on Build GRCh38
rs1022902336 Chr.1: 28668887-28668887 on Build GRCh38
rs539476297 Chr.1: 28669134-28669134 on Build GRCh38
rs35229620 Chr.1: 28668742-28668743 on Build GRCh38
rs1025711163 Chr.1: 28668908-28668935 on Build GRCh38
rs951767199 Chr.1: 28668715-28668715 on Build GRCh38
rs976939220 Chr.1: 28669059-28669059 on Build GRCh38
rs966688392 Chr.1: 28668669-28668669 on Build GRCh38
rs540342560 Chr.1: 28668758-28668758 on Build GRCh38
rs916299410 Chr.1: 28668698-28668698 on Build GRCh38
rs544539958 Chr.1: 28668702-28668702 on Build GRCh38
rs912615987 Chr.1: 28668992-28668994 on Build GRCh38
rs140690565 Chr.1: 28668656-28668656 on Build GRCh38
rs895890911 Chr.1: 28668876-28668876 on Build GRCh38
rs1004629120 Chr.1: 28668942-28668942 on Build GRCh38
rs949202922 Chr.1: 28668700-28668700 on Build GRCh38
rs916756896 Chr.1: 28668950-28668950 on Build GRCh38
rs1040226262 Chr.1: 28668849-28668849 on Build GRCh38
rs975510950 Chr.1: 28669108-28669108 on Build GRCh38
rs866946960 Chr.1: 28668796-28668796 on Build GRCh38
rs979665211 Chr.1: 28669073-28669073 on Build GRCh38
rs956412493 Chr.1: 28669042-28669042 on Build GRCh38
rs868638719 Chr.1: 28668904-28668904 on Build GRCh38
rs986483913 Chr.1: 28669064-28669072 on Build GRCh38
rs532182960 Chr.1: 28669062-28669062 on Build GRCh38
rs936813655 Chr.1: 28668679-28668679 on Build GRCh38
rs988520237 Chr.1: 28669083-28669083 on Build GRCh38
rs935356220 Chr.1: 28668984-28668984 on Build GRCh38
rs1052510441 Chr.1: 28668991-28668992 on Build GRCh38
rs956994207 Chr.1: 28669079-28669079 on Build GRCh38
rs990977130 Chr.1: 28668693-28668693 on Build GRCh38
rs979717529 Chr.1: 28668967-28668967 on Build GRCh38
rs891611067 Chr.1: 28668772-28668772 on Build GRCh38
rs904811908 Chr.1: 28668895-28668895 on Build GRCh38
rs745708005 Chr.1: 28668718-28668718 on Build GRCh38
rs935424092 Chr.1: 28668765-28668765 on Build GRCh38
rs543842076 Chr.1: 28668667-28668667 on Build GRCh38
rs1052981138 Chr.1: 28668768-28668768 on Build GRCh38
rs1014603582 Chr.1: 28669057-28669057 on Build GRCh38
rs921760618 Chr.1: 28669112-28669112 on Build GRCh38
rs559865781 Chr.1: 28668771-28668771 on Build GRCh38
rs886072843 Chr.1: 28668712-28668712 on Build GRCh38
rs944151843 Chr.1: 28669035-28669035 on Build GRCh38

View Full Product Details