Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 29475547-29476091 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAOK1
Entrez Gene ID
57551
Gene Name
TAO kinase 1
Gene Aliases
KFC-B, MAP3K16, MARKK, PSK-2, PSK2, TAO1, hKFC-B, hTAOK1
UniGene
Hs.597434
Gene Chromosome Location
Chr.17: 29390925-29551904 on Build GRCh38
Transcripts
NM_020791.2 NM_025142.1 XM_011525060.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2801696 c.279C>T p.G93G Chr.17: 29475744-29475744 on Build GRCh38
3421378 c.241C>G p.Q81E Chr.17: 29475706-29475706 on Build GRCh38
3515627 c.306+2T>C p.? Chr.17: 29475773-29475773 on Build GRCh38
2801695 c.255T>C p.H85H Chr.17: 29475720-29475720 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs762313690 Chr.17: 29475689-29475689 on Build GRCh38
rs375273313 Chr.17: 29475641-29475641 on Build GRCh38
rs530656196 Chr.17: 29475732-29475732 on Build GRCh38
rs774433786 Chr.17: 29475666-29475666 on Build GRCh38
rs768391759 Chr.17: 29475904-29475904 on Build GRCh38
rs935370154 Chr.17: 29476048-29476048 on Build GRCh38
rs933343754 Chr.17: 29475873-29475873 on Build GRCh38
rs924008693 Chr.17: 29476037-29476037 on Build GRCh38
rs567389817 Chr.17: 29475774-29475774 on Build GRCh38
rs757823706 Chr.17: 29475966-29475966 on Build GRCh38
rs78807458 Chr.17: 29475798-29475798 on Build GRCh38
rs147203802 Chr.17: 29475765-29475765 on Build GRCh38
rs563594248 Chr.17: 29475612-29475612 on Build GRCh38
rs963947113 Chr.17: 29475927-29475927 on Build GRCh38
rs752326115 Chr.17: 29475768-29475768 on Build GRCh38
rs769749101 Chr.17: 29475812-29475812 on Build GRCh38
rs372410494 Chr.17: 29475790-29475790 on Build GRCh38
rs1043851437 Chr.17: 29476036-29476036 on Build GRCh38
rs951111209 Chr.17: 29475863-29475863 on Build GRCh38
rs372221258 Chr.17: 29475958-29475958 on Build GRCh38
rs529322022 Chr.17: 29475837-29475837 on Build GRCh38
rs146822918 Chr.17: 29476049-29476049 on Build GRCh38
rs745777189 Chr.17: 29475799-29475799 on Build GRCh38
rs547405594 Chr.17: 29475860-29475860 on Build GRCh38
rs376512377 Chr.17: 29475794-29475794 on Build GRCh38
rs768079772 Chr.17: 29475715-29475715 on Build GRCh38
rs947168499 Chr.17: 29476033-29476033 on Build GRCh38
rs995272831 Chr.17: 29475842-29475842 on Build GRCh38
rs184683020 Chr.17: 29476035-29476035 on Build GRCh38
rs1019998031 Chr.17: 29475573-29475573 on Build GRCh38
rs919316173 Chr.17: 29475866-29475866 on Build GRCh38
rs559283923 Chr.17: 29475890-29475890 on Build GRCh38
rs763202541 Chr.17: 29475619-29475619 on Build GRCh38
rs760799793 Chr.17: 29475792-29475792 on Build GRCh38
rs570321287 Chr.17: 29476018-29476018 on Build GRCh38
rs758017007 Chr.17: 29475778-29475778 on Build GRCh38
rs765959656 Chr.17: 29475779-29475779 on Build GRCh38
rs551836273 Chr.17: 29475937-29475937 on Build GRCh38
rs112664157 Chr.17: 29475979-29475979 on Build GRCh38
rs113540803 Chr.17: 29475763-29475763 on Build GRCh38
rs1039621775 Chr.17: 29475990-29475990 on Build GRCh38
rs1012482326 Chr.17: 29475781-29475781 on Build GRCh38
rs912500885 Chr.17: 29475926-29475926 on Build GRCh38
rs750920986 Chr.17: 29475744-29475744 on Build GRCh38
rs533451070 Chr.17: 29475930-29475930 on Build GRCh38
rs775314561 Chr.17: 29475816-29475816 on Build GRCh38
rs775666201 Chr.17: 29475614-29475614 on Build GRCh38
rs756523875 Chr.17: 29475750-29475750 on Build GRCh38

View Full Product Details