Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 29480026-29480575 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAOK1
Entrez Gene ID
57551
Gene Name
TAO kinase 1
Gene Aliases
KFC-B, MAP3K16, MARKK, PSK-2, PSK2, TAO1, hKFC-B, hTAOK1
UniGene
Hs.597434
Gene Chromosome Location
Chr.17: 29390925-29551904 on Build GRCh38
Transcripts
NM_020791.2 NM_025142.1 XM_011525060.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7158710 c.496A>G p.K166E Chr.17: 29480414-29480414 on Build GRCh38
5672131 c.557C>T p.P186L Chr.17: 29480475-29480475 on Build GRCh38
6573518 c.495G>A p.V165V Chr.17: 29480413-29480413 on Build GRCh38
1710068 c.451G>C p.D151H Chr.17: 29480369-29480369 on Build GRCh38
5385956 c.521C>T p.S174F Chr.17: 29480439-29480439 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs56387760 Chr.17: 29480398-29480398 on Build GRCh38
rs986037497 Chr.17: 29480057-29480057 on Build GRCh38
rs760282156 Chr.17: 29480505-29480505 on Build GRCh38
rs770897827 Chr.17: 29480449-29480449 on Build GRCh38
rs903695137 Chr.17: 29480123-29480124 on Build GRCh38
rs529433124 Chr.17: 29480107-29480107 on Build GRCh38
rs942206748 Chr.17: 29480397-29480397 on Build GRCh38
rs1007892550 Chr.17: 29480533-29480533 on Build GRCh38
rs868114481 Chr.17: 29480068-29480068 on Build GRCh38
rs569346615 Chr.17: 29480173-29480173 on Build GRCh38
rs188683307 Chr.17: 29480119-29480119 on Build GRCh38
rs1015696441 Chr.17: 29480362-29480362 on Build GRCh38
rs760000002 Chr.17: 29480329-29480329 on Build GRCh38
rs145506168 Chr.17: 29480437-29480437 on Build GRCh38
rs80173580 Chr.17: 29480353-29480353 on Build GRCh38
rs761562887 Chr.17: 29480498-29480498 on Build GRCh38
rs773525136 Chr.17: 29480224-29480224 on Build GRCh38
rs140166555 Chr.17: 29480141-29480141 on Build GRCh38
rs921251467 Chr.17: 29480090-29480090 on Build GRCh38
rs553851015 Chr.17: 29480518-29480521 on Build GRCh38
rs1002463692 Chr.17: 29480265-29480265 on Build GRCh38
rs772227735 Chr.17: 29480441-29480441 on Build GRCh38
rs928589185 Chr.17: 29480194-29480194 on Build GRCh38
rs777831998 Chr.17: 29480442-29480442 on Build GRCh38
rs776897242 Chr.17: 29480139-29480139 on Build GRCh38
rs776995057 Chr.17: 29480325-29480325 on Build GRCh38
rs1052202069 Chr.17: 29480134-29480136 on Build GRCh38
rs552485341 Chr.17: 29480076-29480076 on Build GRCh38
rs770223672 Chr.17: 29480473-29480473 on Build GRCh38
rs765495165 Chr.17: 29480333-29480333 on Build GRCh38
rs527286712 Chr.17: 29480071-29480071 on Build GRCh38
rs904853482 Chr.17: 29480226-29480226 on Build GRCh38
rs937595119 Chr.17: 29480309-29480309 on Build GRCh38
rs780098181 Chr.17: 29480357-29480357 on Build GRCh38
rs770362778 Chr.17: 29480212-29480212 on Build GRCh38
rs957893994 Chr.17: 29480082-29480082 on Build GRCh38
rs528533125 Chr.17: 29480550-29480550 on Build GRCh38
rs191111852 Chr.17: 29480176-29480176 on Build GRCh38
rs368082855 Chr.17: 29480506-29480506 on Build GRCh38
rs976803317 Chr.17: 29480072-29480072 on Build GRCh38
rs763388165 Chr.17: 29480289-29480289 on Build GRCh38
rs761515791 Chr.17: 29480347-29480347 on Build GRCh38
rs555008031 Chr.17: 29480288-29480288 on Build GRCh38
rs767157503 Chr.17: 29480351-29480351 on Build GRCh38
rs562380732 Chr.17: 29480521-29480521 on Build GRCh38
rs549680039 Chr.17: 29480169-29480169 on Build GRCh38
rs757420579 Chr.17: 29480279-29480279 on Build GRCh38
rs750980623 Chr.17: 29480528-29480528 on Build GRCh38
rs1050729649 Chr.17: 29480122-29480122 on Build GRCh38
rs866335866 Chr.17: 29480390-29480390 on Build GRCh38
rs559239646 Chr.17: 29480549-29480549 on Build GRCh38
rs753697188 Chr.17: 29480372-29480372 on Build GRCh38
rs977235145 Chr.17: 29480394-29480394 on Build GRCh38
rs766797369 Chr.17: 29480341-29480341 on Build GRCh38
rs201854324 Chr.17: 29480457-29480457 on Build GRCh38
rs936459731 Chr.17: 29480124-29480124 on Build GRCh38
rs958138889 Chr.17: 29480543-29480543 on Build GRCh38
rs777241118 Chr.17: 29480455-29480455 on Build GRCh38
rs376864746 Chr.17: 29480354-29480354 on Build GRCh38
rs747156223 Chr.17: 29480445-29480445 on Build GRCh38
rs772986529 Chr.17: 29480504-29480504 on Build GRCh38
rs1027161383 Chr.17: 29480308-29480308 on Build GRCh38
rs767347683 Chr.17: 29480499-29480499 on Build GRCh38
rs1032251596 Chr.17: 29480538-29480538 on Build GRCh38
rs765779366 Chr.17: 29480516-29480516 on Build GRCh38
rs371833053 Chr.17: 29480425-29480425 on Build GRCh38
rs373849651 Chr.17: 29480476-29480476 on Build GRCh38
rs376345627 Chr.17: 29480324-29480324 on Build GRCh38
rs747298489 Chr.17: 29480112-29480112 on Build GRCh38

View Full Product Details