Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 30333922-30334436 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMIGD1
Entrez Gene ID
388364
Gene Name
transmembrane and immunoglobulin domain containing 1
Gene Aliases
TMIGD, UNQ9372
UniGene
Hs.371005
Gene Chromosome Location
Chr.17: 30316329-30334076 on Build GRCh38
Transcripts
NM_001319942.1 NM_206832.2 XM_011524787.1

SNP Detail

SNP ID Chromosome Location
rs75234423 Chr.17: 30334117-30334117 on Build GRCh38
rs545687830 Chr.17: 30333976-30333976 on Build GRCh38
rs988970368 Chr.17: 30334351-30334351 on Build GRCh38
rs578152976 Chr.17: 30333960-30333960 on Build GRCh38
rs1044382581 Chr.17: 30334118-30334118 on Build GRCh38
rs1027503428 Chr.17: 30334168-30334168 on Build GRCh38
rs191731808 Chr.17: 30333993-30333993 on Build GRCh38
rs1033035050 Chr.17: 30334248-30334248 on Build GRCh38
rs117986747 Chr.17: 30334252-30334252 on Build GRCh38
rs1018691811 Chr.17: 30334201-30334201 on Build GRCh38
rs1043234925 Chr.17: 30333978-30333978 on Build GRCh38
rs954120172 Chr.17: 30334241-30334241 on Build GRCh38
rs940376707 Chr.17: 30334383-30334383 on Build GRCh38
rs183137755 Chr.17: 30334352-30334352 on Build GRCh38
rs1028826436 Chr.17: 30334232-30334232 on Build GRCh38
rs925914277 Chr.17: 30334071-30334071 on Build GRCh38
rs927430610 Chr.17: 30334347-30334347 on Build GRCh38
rs148612180 Chr.17: 30334192-30334192 on Build GRCh38
rs932863521 Chr.17: 30333965-30333965 on Build GRCh38
rs1049475959 Chr.17: 30334387-30334387 on Build GRCh38
rs1028577075 Chr.17: 30334249-30334249 on Build GRCh38
rs965760205 Chr.17: 30334244-30334244 on Build GRCh38
rs895901408 Chr.17: 30334081-30334081 on Build GRCh38
rs917482954 Chr.17: 30334109-30334109 on Build GRCh38
rs3110093 Chr.17: 30334374-30334374 on Build GRCh38
rs901745654 Chr.17: 30334212-30334212 on Build GRCh38
rs978823231 Chr.17: 30333994-30333994 on Build GRCh38
rs930650338 Chr.17: 30334386-30334386 on Build GRCh38
rs974388322 Chr.17: 30334247-30334247 on Build GRCh38
rs996042793 Chr.17: 30334223-30334223 on Build GRCh38
rs1044434192 Chr.17: 30334360-30334360 on Build GRCh38
rs905781516 Chr.17: 30334126-30334126 on Build GRCh38
rs1018894702 Chr.17: 30334207-30334207 on Build GRCh38
rs950302031 Chr.17: 30334294-30334294 on Build GRCh38
rs934862966 Chr.17: 30334339-30334339 on Build GRCh38
rs774334806 Chr.17: 30333987-30333987 on Build GRCh38
rs934832372 Chr.17: 30334027-30334027 on Build GRCh38
rs759551741 Chr.17: 30334373-30334373 on Build GRCh38
rs16965686 Chr.17: 30334355-30334355 on Build GRCh38
rs143850536 Chr.17: 30334120-30334120 on Build GRCh38
rs112764764 Chr.17: 30334102-30334102 on Build GRCh38
rs926084350 Chr.17: 30333999-30333999 on Build GRCh38
rs767657356 Chr.17: 30334276-30334276 on Build GRCh38
rs905661118 Chr.17: 30334141-30334141 on Build GRCh38
rs572619322 Chr.17: 30333992-30333992 on Build GRCh38
rs1053217136 Chr.17: 30334029-30334029 on Build GRCh38
rs540091854 Chr.17: 30334311-30334311 on Build GRCh38
rs930350762 Chr.17: 30334184-30334184 on Build GRCh38
rs753570603 Chr.17: 30334346-30334346 on Build GRCh38
rs144250775 Chr.17: 30334199-30334199 on Build GRCh38
rs995138188 Chr.17: 30334140-30334140 on Build GRCh38
rs1048728625 Chr.17: 30334187-30334187 on Build GRCh38
rs989447191 Chr.17: 30334108-30334108 on Build GRCh38
rs988815317 Chr.17: 30334274-30334274 on Build GRCh38

View Full Product Details