Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 17202890-17203386 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLD6
Entrez Gene ID
201164
Gene Name
phospholipase D family member 6
Gene Aliases
ZUC
UniGene
Hs.729079
Gene Chromosome Location
Chr.17: 17200990-17206332 on Build GRCh38
Transcripts
NM_178836.3 XM_017024310.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5481349 c.460T>C p.Y154H Chr.17: 17203066-17203066 on Build GRCh38
1221016 c.610C>T p.R204C Chr.17: 17202916-17202916 on Build GRCh38
6748641 c.506C>A p.T169N Chr.17: 17203020-17203020 on Build GRCh38
560007 c.445G>A p.D149N Chr.17: 17203081-17203081 on Build GRCh38
5055262 c.573C>T p.D191D Chr.17: 17202953-17202953 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763115121 Chr.17: 17203122-17203122 on Build GRCh38
rs1008571243 Chr.17: 17203358-17203358 on Build GRCh38
rs201208806 Chr.17: 17202939-17202939 on Build GRCh38
rs373555063 Chr.17: 17203031-17203031 on Build GRCh38
rs909073336 Chr.17: 17203337-17203337 on Build GRCh38
rs1000122328 Chr.17: 17203205-17203205 on Build GRCh38
rs780960567 Chr.17: 17203039-17203039 on Build GRCh38
rs769670654 Chr.17: 17202986-17202986 on Build GRCh38
rs760231888 Chr.17: 17203115-17203116 on Build GRCh38
rs770332478 Chr.17: 17203148-17203148 on Build GRCh38
rs774000285 Chr.17: 17203003-17203003 on Build GRCh38
rs768493867 Chr.17: 17202963-17202963 on Build GRCh38
rs745734555 Chr.17: 17203313-17203313 on Build GRCh38
rs978109062 Chr.17: 17203198-17203198 on Build GRCh38
rs1025025098 Chr.17: 17203152-17203152 on Build GRCh38
rs542678889 Chr.17: 17203239-17203239 on Build GRCh38
rs750641276 Chr.17: 17203014-17203014 on Build GRCh38
rs903336963 Chr.17: 17203353-17203353 on Build GRCh38
rs765522425 Chr.17: 17203095-17203095 on Build GRCh38
rs766619600 Chr.17: 17203124-17203124 on Build GRCh38
rs752426772 Chr.17: 17203056-17203056 on Build GRCh38
rs968490076 Chr.17: 17203160-17203160 on Build GRCh38
rs771629386 Chr.17: 17203039-17203039 on Build GRCh38
rs370282805 Chr.17: 17203285-17203285 on Build GRCh38
rs116123204 Chr.17: 17203259-17203259 on Build GRCh38
rs116547825 Chr.17: 17202965-17202965 on Build GRCh38
rs761183704 Chr.17: 17203009-17203009 on Build GRCh38
rs748880961 Chr.17: 17203077-17203077 on Build GRCh38
rs540237848 Chr.17: 17203185-17203185 on Build GRCh38
rs751848703 Chr.17: 17203126-17203126 on Build GRCh38
rs769604058 Chr.17: 17203317-17203317 on Build GRCh38
rs886205971 Chr.17: 17203356-17203356 on Build GRCh38
rs964612186 Chr.17: 17203131-17203131 on Build GRCh38
rs142631639 Chr.17: 17202944-17202944 on Build GRCh38
rs9914950 Chr.17: 17203013-17203013 on Build GRCh38
rs772976619 Chr.17: 17202992-17202992 on Build GRCh38
rs150565127 Chr.17: 17202950-17202950 on Build GRCh38
rs966780760 Chr.17: 17203193-17203193 on Build GRCh38
rs144490572 Chr.17: 17202998-17202998 on Build GRCh38
rs780051488 Chr.17: 17203022-17203022 on Build GRCh38
rs368055624 Chr.17: 17202952-17202952 on Build GRCh38
rs924026875 Chr.17: 17203244-17203244 on Build GRCh38
rs1047359292 Chr.17: 17203338-17203338 on Build GRCh38
rs758461726 Chr.17: 17202959-17202959 on Build GRCh38
rs775281929 Chr.17: 17203097-17203097 on Build GRCh38
rs1013533671 Chr.17: 17203150-17203150 on Build GRCh38
rs777649728 Chr.17: 17203148-17203148 on Build GRCh38
rs372737833 Chr.17: 17203143-17203143 on Build GRCh38
rs772808546 Chr.17: 17202916-17202916 on Build GRCh38
rs573794391 Chr.17: 17203238-17203238 on Build GRCh38
rs1054950429 Chr.17: 17203139-17203139 on Build GRCh38
rs762521324 Chr.17: 17202926-17202926 on Build GRCh38
rs992346980 Chr.17: 17203280-17203280 on Build GRCh38
rs765454108 Chr.17: 17203046-17203046 on Build GRCh38
rs527533472 Chr.17: 17203364-17203364 on Build GRCh38
rs139543758 Chr.17: 17202960-17202960 on Build GRCh38
rs751662295 Chr.17: 17203028-17203028 on Build GRCh38
rs762454262 Chr.17: 17203091-17203091 on Build GRCh38
rs757940831 Chr.17: 17203090-17203090 on Build GRCh38
rs145002892 Chr.17: 17203086-17203086 on Build GRCh38
rs762750008 Chr.17: 17202996-17202996 on Build GRCh38
rs560151308 Chr.17: 17203215-17203215 on Build GRCh38
rs750800824 Chr.17: 17203103-17203103 on Build GRCh38
rs1055002737 Chr.17: 17203347-17203347 on Build GRCh38
rs61738638 Chr.17: 17203007-17203007 on Build GRCh38
rs376910993 Chr.17: 17202940-17202940 on Build GRCh38
rs958434766 Chr.17: 17203225-17203226 on Build GRCh38
rs369608720 Chr.17: 17203082-17203082 on Build GRCh38
rs373078017 Chr.17: 17203141-17203141 on Build GRCh38
rs550504699 Chr.17: 17203334-17203334 on Build GRCh38
rs531772328 Chr.17: 17203226-17203226 on Build GRCh38
rs916724024 Chr.17: 17203289-17203289 on Build GRCh38
rs748165661 Chr.17: 17203043-17203043 on Build GRCh38
rs564880519 Chr.17: 17203359-17203359 on Build GRCh38
rs769462749 Chr.17: 17203047-17203047 on Build GRCh38
rs551274091 Chr.17: 17203301-17203301 on Build GRCh38
rs371619441 Chr.17: 17202953-17202953 on Build GRCh38
rs1018708395 Chr.17: 17203099-17203099 on Build GRCh38
rs756079987 Chr.17: 17203144-17203144 on Build GRCh38
rs1032482627 Chr.17: 17203214-17203214 on Build GRCh38
rs1038657901 Chr.17: 17203295-17203295 on Build GRCh38
rs764527127 Chr.17: 17203010-17203010 on Build GRCh38
rs754989451 Chr.17: 17203136-17203136 on Build GRCh38
rs186327050 Chr.17: 17203081-17203081 on Build GRCh38
rs375857325 Chr.17: 17202977-17202977 on Build GRCh38
rs948191351 Chr.17: 17203290-17203290 on Build GRCh38
rs898856295 Chr.17: 17202999-17202999 on Build GRCh38
rs777776148 Chr.17: 17203061-17203061 on Build GRCh38
rs754186732 Chr.17: 17203011-17203011 on Build GRCh38
rs756758567 Chr.17: 17203245-17203245 on Build GRCh38
rs781168317 Chr.17: 17203273-17203273 on Build GRCh38
rs375940521 Chr.17: 17202915-17202915 on Build GRCh38

View Full Product Details