Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28398036-28398537 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SARM1
Entrez Gene ID
23098
Gene Name
sterile alpha and TIR motif containing 1
Gene Aliases
MyD88-5, SAMD2, SARM
UniGene
Hs.743510
Gene Chromosome Location
Chr.17: 28371662-28401045 on Build GRCh38
Transcripts
NM_015077.3 NM_001242366.2 NM_080669.5 XM_005277786.3 XM_017024110.1

Gene Symbol
SLC46A1
Entrez Gene ID
113235
Gene Name
solute carrier family 46 member 1
Gene Aliases
G21, HCP1, PCFT
UniGene
Hs.446689
Gene Chromosome Location
Chr.17: 28394642-28406630 on Build GRCh38
Transcripts
NM_015077.3 NM_001242366.2 NM_080669.5 XM_005277786.3 XM_017024110.1

SNP Detail

SNP ID Chromosome Location
rs781824816 Chr.17: 28398407-28398407 on Build GRCh38
rs141940033 Chr.17: 28398264-28398264 on Build GRCh38
rs560836860 Chr.17: 28398273-28398273 on Build GRCh38
rs913151703 Chr.17: 28398148-28398148 on Build GRCh38
rs997790463 Chr.17: 28398436-28398436 on Build GRCh38
rs953556650 Chr.17: 28398068-28398068 on Build GRCh38
rs928501627 Chr.17: 28398307-28398307 on Build GRCh38
rs531209426 Chr.17: 28398460-28398460 on Build GRCh38
rs370834754 Chr.17: 28398346-28398346 on Build GRCh38
rs986248835 Chr.17: 28398102-28398102 on Build GRCh38
rs41297911 Chr.17: 28398148-28398148 on Build GRCh38
rs139970165 Chr.17: 28398358-28398358 on Build GRCh38
rs577268750 Chr.17: 28398113-28398113 on Build GRCh38
rs988138947 Chr.17: 28398135-28398135 on Build GRCh38
rs576841097 Chr.17: 28398276-28398276 on Build GRCh38
rs369226733 Chr.17: 28398271-28398271 on Build GRCh38
rs886052748 Chr.17: 28398436-28398436 on Build GRCh38
rs146041886 Chr.17: 28398324-28398324 on Build GRCh38
rs562847087 Chr.17: 28398150-28398150 on Build GRCh38
rs782514608 Chr.17: 28398267-28398268 on Build GRCh38
rs782497027 Chr.17: 28398412-28398412 on Build GRCh38
rs62065282 Chr.17: 28398505-28398505 on Build GRCh38
rs2239908 Chr.17: 28398249-28398249 on Build GRCh38
rs781900716 Chr.17: 28398067-28398067 on Build GRCh38
rs549805854 Chr.17: 28398486-28398486 on Build GRCh38
rs958017325 Chr.17: 28398133-28398133 on Build GRCh38
rs41297909 Chr.17: 28398413-28398413 on Build GRCh38
rs781874858 Chr.17: 28398417-28398417 on Build GRCh38

View Full Product Details