Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28400256-28400777 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SARM1
Entrez Gene ID
23098
Gene Name
sterile alpha and TIR motif containing 1
Gene Aliases
MyD88-5, SAMD2, SARM
UniGene
Hs.743510
Gene Chromosome Location
Chr.17: 28371662-28401045 on Build GRCh38
Transcripts
NM_015077.3 NM_001242366.2 NM_080669.5 XM_005277786.3 XM_017024110.1

Gene Symbol
SLC46A1
Entrez Gene ID
113235
Gene Name
solute carrier family 46 member 1
Gene Aliases
G21, HCP1, PCFT
UniGene
Hs.446689
Gene Chromosome Location
Chr.17: 28394642-28406630 on Build GRCh38
Transcripts
NM_015077.3 NM_001242366.2 NM_080669.5 XM_005277786.3 XM_017024110.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3362040 c.1171C>A p.L391I Chr.17: 28400761-28400761 on Build GRCh38
3889448 c.1276C>T p.L426F Chr.17: 28400653-28400653 on Build GRCh38
3680121 c.1304C>T p.P435L Chr.17: 28400625-28400625 on Build GRCh38
5586609 c.1278C>T p.L426L Chr.17: 28400651-28400651 on Build GRCh38
5852084 c.1264G>A p.G422R Chr.17: 28400665-28400665 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782287467 Chr.17: 28400687-28400687 on Build GRCh38
rs782027732 Chr.17: 28400610-28400610 on Build GRCh38
rs781998047 Chr.17: 28400691-28400691 on Build GRCh38
rs781803952 Chr.17: 28400636-28400636 on Build GRCh38
rs897179345 Chr.17: 28400524-28400524 on Build GRCh38
rs782576278 Chr.17: 28400651-28400651 on Build GRCh38
rs80338774 Chr.17: 28400658-28400658 on Build GRCh38
rs782188751 Chr.17: 28400671-28400671 on Build GRCh38
rs41297113 Chr.17: 28400488-28400488 on Build GRCh38
rs569945738 Chr.17: 28400352-28400352 on Build GRCh38
rs138861714 Chr.17: 28400426-28400426 on Build GRCh38
rs539619751 Chr.17: 28400541-28400541 on Build GRCh38
rs939505362 Chr.17: 28400427-28400427 on Build GRCh38
rs561780114 Chr.17: 28400706-28400706 on Build GRCh38
rs374232525 Chr.17: 28400727-28400727 on Build GRCh38
rs533154330 Chr.17: 28400303-28400303 on Build GRCh38
rs761549800 Chr.17: 28400717-28400717 on Build GRCh38
rs782344527 Chr.17: 28400573-28400573 on Build GRCh38
rs782189198 Chr.17: 28400604-28400604 on Build GRCh38
rs781949017 Chr.17: 28400625-28400625 on Build GRCh38
rs890413466 Chr.17: 28400612-28400612 on Build GRCh38
rs562411490 Chr.17: 28400669-28400669 on Build GRCh38
rs941432545 Chr.17: 28400494-28400494 on Build GRCh38
rs1048095279 Chr.17: 28400585-28400585 on Build GRCh38
rs368561206 Chr.17: 28400630-28400630 on Build GRCh38
rs576590466 Chr.17: 28400583-28400583 on Build GRCh38
rs557962388 Chr.17: 28400565-28400565 on Build GRCh38
rs370083652 Chr.17: 28400605-28400605 on Build GRCh38
rs782782249 Chr.17: 28400723-28400723 on Build GRCh38
rs782044138 Chr.17: 28400745-28400745 on Build GRCh38
rs782527604 Chr.17: 28400620-28400620 on Build GRCh38
rs781853191 Chr.17: 28400639-28400639 on Build GRCh38
rs782811181 Chr.17: 28400638-28400638 on Build GRCh38
rs782352629 Chr.17: 28400696-28400696 on Build GRCh38
rs991041226 Chr.17: 28400431-28400431 on Build GRCh38
rs113278826 Chr.17: 28400593-28400593 on Build GRCh38
rs782530326 Chr.17: 28400642-28400642 on Build GRCh38
rs782201123 Chr.17: 28400695-28400695 on Build GRCh38
rs201732528 Chr.17: 28400706-28400706 on Build GRCh38
rs199727773 Chr.17: 28400724-28400724 on Build GRCh38
rs367590729 Chr.17: 28400314-28400314 on Build GRCh38
rs782595969 Chr.17: 28400646-28400646 on Build GRCh38
rs141992236 Chr.17: 28400676-28400676 on Build GRCh38
rs199895119 Chr.17: 28400705-28400705 on Build GRCh38
rs782534694 Chr.17: 28400637-28400637 on Build GRCh38
rs782796227 Chr.17: 28400733-28400733 on Build GRCh38
rs374502830 Chr.17: 28400705-28400705 on Build GRCh38
rs568315880 Chr.17: 28400404-28400404 on Build GRCh38
rs1021928823 Chr.17: 28400369-28400369 on Build GRCh38
rs185918175 Chr.17: 28400419-28400419 on Build GRCh38
rs541069777 Chr.17: 28400718-28400718 on Build GRCh38
rs782590413 Chr.17: 28400678-28400678 on Build GRCh38
rs553839009 Chr.17: 28400424-28400424 on Build GRCh38
rs756768 Chr.17: 28400361-28400361 on Build GRCh38
rs782089869 Chr.17: 28400629-28400629 on Build GRCh38
rs1038352110 Chr.17: 28400513-28400513 on Build GRCh38
rs201932339 Chr.17: 28400703-28400703 on Build GRCh38
rs183886521 Chr.17: 28400388-28400388 on Build GRCh38
rs960988954 Chr.17: 28400725-28400725 on Build GRCh38
rs782456428 Chr.17: 28400741-28400741 on Build GRCh38
rs782259423 Chr.17: 28400566-28400566 on Build GRCh38
rs776702709 Chr.17: 28400598-28400598 on Build GRCh38

View Full Product Details