Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28719902-28720395 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPL23A
Entrez Gene ID
6147
Gene Name
ribosomal protein L23a
Gene Aliases
L23A, MDA20
UniGene
Hs.419463
Gene Chromosome Location
Chr.17: 28719982-28724356 on Build GRCh38
Transcripts
NM_000984.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3819144 c.21G>A p.K7K Chr.17: 28720026-28720026 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs539168874 Chr.17: 28720026-28720026 on Build GRCh38
rs867866996 Chr.17: 28720181-28720181 on Build GRCh38
rs1054663540 Chr.17: 28720138-28720138 on Build GRCh38
rs773360501 Chr.17: 28720152-28720152 on Build GRCh38
rs748668556 Chr.17: 28720042-28720042 on Build GRCh38
rs573745975 Chr.17: 28720331-28720331 on Build GRCh38
rs935850154 Chr.17: 28720135-28720135 on Build GRCh38
rs977647073 Chr.17: 28719997-28719997 on Build GRCh38
rs537101000 Chr.17: 28720291-28720291 on Build GRCh38
rs564602460 Chr.17: 28720172-28720172 on Build GRCh38
rs572440256 Chr.17: 28720157-28720157 on Build GRCh38
rs1015023383 Chr.17: 28720033-28720033 on Build GRCh38
rs760486105 Chr.17: 28720229-28720229 on Build GRCh38
rs377608220 Chr.17: 28720234-28720234 on Build GRCh38
rs964456542 Chr.17: 28719979-28719979 on Build GRCh38
rs935781470 Chr.17: 28719986-28719986 on Build GRCh38
rs748923717 Chr.17: 28720292-28720292 on Build GRCh38
rs1015075954 Chr.17: 28720201-28720201 on Build GRCh38
rs1006401451 Chr.17: 28720199-28720199 on Build GRCh38
rs569460754 Chr.17: 28720235-28720235 on Build GRCh38
rs1008087692 Chr.17: 28720141-28720141 on Build GRCh38
rs752763961 Chr.17: 28720374-28720374 on Build GRCh38
rs186231161 Chr.17: 28720344-28720344 on Build GRCh38
rs891509006 Chr.17: 28719989-28719989 on Build GRCh38
rs759748355 Chr.17: 28720364-28720364 on Build GRCh38
rs142109717 Chr.17: 28720176-28720176 on Build GRCh38
rs774606221 Chr.17: 28720198-28720198 on Build GRCh38
rs760968797 Chr.17: 28720227-28720228 on Build GRCh38
rs1030368783 Chr.17: 28720225-28720225 on Build GRCh38
rs923248754 Chr.17: 28720075-28720075 on Build GRCh38
rs756297747 Chr.17: 28720001-28720001 on Build GRCh38
rs985384260 Chr.17: 28720109-28720109 on Build GRCh38
rs372944738 Chr.17: 28720049-28720049 on Build GRCh38
rs779006363 Chr.17: 28720038-28720038 on Build GRCh38
rs997639789 Chr.17: 28720367-28720367 on Build GRCh38
rs981149322 Chr.17: 28720253-28720253 on Build GRCh38
rs912998228 Chr.17: 28720148-28720148 on Build GRCh38
rs373298218 Chr.17: 28719995-28719995 on Build GRCh38
rs960268652 Chr.17: 28719956-28719956 on Build GRCh38
rs778193730 Chr.17: 28720046-28720046 on Build GRCh38
rs550112443 Chr.17: 28720183-28720183 on Build GRCh38
rs773846925 Chr.17: 28720301-28720301 on Build GRCh38
rs376842436 Chr.17: 28719988-28719988 on Build GRCh38
rs970663002 Chr.17: 28720243-28720243 on Build GRCh38
rs566010875 Chr.17: 28720228-28720228 on Build GRCh38
rs922525094 Chr.17: 28719953-28719953 on Build GRCh38
rs763194820 Chr.17: 28720174-28720174 on Build GRCh38
rs565305140 Chr.17: 28720005-28720005 on Build GRCh38
rs553446098 Chr.17: 28720058-28720058 on Build GRCh38
rs201418245 Chr.17: 28719983-28719983 on Build GRCh38
rs977386913 Chr.17: 28720196-28720196 on Build GRCh38
rs1023181730 Chr.17: 28720240-28720240 on Build GRCh38
rs768492968 Chr.17: 28720299-28720299 on Build GRCh38
rs1009867709 Chr.17: 28719991-28719991 on Build GRCh38
rs887052606 Chr.17: 28720228-28720229 on Build GRCh38
rs911042294 Chr.17: 28719980-28719980 on Build GRCh38
rs35537717 Chr.17: 28720234-28720235 on Build GRCh38
rs765950505 Chr.17: 28720226-28720226 on Build GRCh38
rs1034960016 Chr.17: 28720022-28720022 on Build GRCh38
rs548045167 Chr.17: 28720227-28720227 on Build GRCh38
rs1044794588 Chr.17: 28720076-28720076 on Build GRCh38
rs539944052 Chr.17: 28720233-28720233 on Build GRCh38
rs557459539 Chr.17: 28720027-28720027 on Build GRCh38
rs989962100 Chr.17: 28720232-28720232 on Build GRCh38
rs769648454 Chr.17: 28720010-28720010 on Build GRCh38
rs367799633 Chr.17: 28719972-28719972 on Build GRCh38
rs767896889 Chr.17: 28720343-28720343 on Build GRCh38
rs147254826 Chr.17: 28720231-28720231 on Build GRCh38
rs937027054 Chr.17: 28720315-28720315 on Build GRCh38
rs754224166 Chr.17: 28720231-28720232 on Build GRCh38
rs1045346045 Chr.17: 28719992-28719992 on Build GRCh38
rs769512437 Chr.17: 28720068-28720068 on Build GRCh38
rs573855856 Chr.17: 28720122-28720122 on Build GRCh38
rs199954719 Chr.17: 28720045-28720045 on Build GRCh38
rs776469237 Chr.17: 28720363-28720363 on Build GRCh38
rs887988907 Chr.17: 28720197-28720197 on Build GRCh38
rs560494257 Chr.17: 28720145-28720145 on Build GRCh38
rs775301342 Chr.17: 28720082-28720082 on Build GRCh38
rs976098648 Chr.17: 28720065-28720065 on Build GRCh38
rs759642555 Chr.17: 28720290-28720290 on Build GRCh38
rs899667613 Chr.17: 28720229-28720230 on Build GRCh38
rs781717968 Chr.17: 28720275-28720275 on Build GRCh38
rs951199921 Chr.17: 28719948-28719948 on Build GRCh38
rs562271033 Chr.17: 28720193-28720193 on Build GRCh38
rs753866318 Chr.17: 28720009-28720009 on Build GRCh38
rs556811788 Chr.17: 28719963-28719963 on Build GRCh38
rs369473946 Chr.17: 28720025-28720025 on Build GRCh38
rs757723603 Chr.17: 28720247-28720247 on Build GRCh38
rs754030643 Chr.17: 28720230-28720230 on Build GRCh38
rs777414001 Chr.17: 28720158-28720158 on Build GRCh38
rs997588924 Chr.17: 28720222-28720222 on Build GRCh38
rs200840240 Chr.17: 28720041-28720041 on Build GRCh38
rs149264208 Chr.17: 28720226-28720227 on Build GRCh38
rs757887187 Chr.17: 28720298-28720299 on Build GRCh38
rs529619980 Chr.17: 28720225-28720226 on Build GRCh38
rs928344363 Chr.17: 28720314-28720314 on Build GRCh38
rs376623510 Chr.17: 28720139-28720139 on Build GRCh38
rs955499573 Chr.17: 28720080-28720080 on Build GRCh38
rs1036561690 Chr.17: 28720345-28720345 on Build GRCh38
rs747346621 Chr.17: 28720237-28720237 on Build GRCh38
rs1055838216 Chr.17: 28720317-28720317 on Build GRCh38
rs529577889 Chr.17: 28720220-28720220 on Build GRCh38
rs113797834 Chr.17: 28720323-28720323 on Build GRCh38
rs546194736 Chr.17: 28720169-28720169 on Build GRCh38
rs766142352 Chr.17: 28720347-28720347 on Build GRCh38
rs536101438 Chr.17: 28719987-28719987 on Build GRCh38
rs898156517 Chr.17: 28720106-28720106 on Build GRCh38
rs375194558 Chr.17: 28720023-28720023 on Build GRCh38
rs765381231 Chr.17: 28720373-28720373 on Build GRCh38

View Full Product Details