Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 43877177-43877672 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MPP2
Entrez Gene ID
4355
Gene Name
membrane palmitoylated protein 2
Gene Aliases
DLG2
UniGene
Hs.514208
Gene Chromosome Location
Chr.17: 43875357-43909711 on Build GRCh38
Transcripts
NM_001278370.1 NM_001278371.1 NM_001278372.1 NM_001278373.1 NM_001278374.1 NM_001278375.1 NM_001278376.2 NM_001278381.1 NM_005374.4 XM_011524827.2

SNP Detail

SNP ID Chromosome Location
rs892046455 Chr.17: 43877368-43877368 on Build GRCh38
rs892057635 Chr.17: 43877549-43877549 on Build GRCh38
rs901417685 Chr.17: 43877445-43877460 on Build GRCh38
rs1034902321 Chr.17: 43877311-43877311 on Build GRCh38
rs993831999 Chr.17: 43877310-43877310 on Build GRCh38
rs925006442 Chr.17: 43877458-43877458 on Build GRCh38
rs917665677 Chr.17: 43877315-43877318 on Build GRCh38
rs1030200810 Chr.17: 43877204-43877204 on Build GRCh38
rs947809310 Chr.17: 43877452-43877452 on Build GRCh38
rs572086331 Chr.17: 43877525-43877525 on Build GRCh38
rs1035113462 Chr.17: 43877645-43877645 on Build GRCh38
rs951242613 Chr.17: 43877437-43877437 on Build GRCh38
rs990642060 Chr.17: 43877344-43877344 on Build GRCh38
rs560859639 Chr.17: 43877404-43877404 on Build GRCh38
rs936214845 Chr.17: 43877462-43877463 on Build GRCh38
rs962958999 Chr.17: 43877279-43877279 on Build GRCh38
rs779899677 Chr.17: 43877199-43877199 on Build GRCh38
rs192640487 Chr.17: 43877353-43877353 on Build GRCh38
rs564899988 Chr.17: 43877520-43877520 on Build GRCh38
rs1044850409 Chr.17: 43877453-43877453 on Build GRCh38
rs1029632160 Chr.17: 43877381-43877381 on Build GRCh38
rs768432527 Chr.17: 43877254-43877254 on Build GRCh38
rs551564949 Chr.17: 43877559-43877559 on Build GRCh38
rs955478619 Chr.17: 43877229-43877229 on Build GRCh38
rs376528343 Chr.17: 43877290-43877290 on Build GRCh38
rs1005706888 Chr.17: 43877578-43877578 on Build GRCh38
rs761476970 Chr.17: 43877581-43877581 on Build GRCh38
rs1018761193 Chr.17: 43877420-43877420 on Build GRCh38
rs543624947 Chr.17: 43877469-43877469 on Build GRCh38
rs934255289 Chr.17: 43877499-43877499 on Build GRCh38
rs974013121 Chr.17: 43877291-43877291 on Build GRCh38
rs1054545970 Chr.17: 43877501-43877501 on Build GRCh38
rs973898526 Chr.17: 43877425-43877425 on Build GRCh38
rs555437092 Chr.17: 43877367-43877367 on Build GRCh38
rs918541061 Chr.17: 43877436-43877436 on Build GRCh38
rs1044976350 Chr.17: 43877328-43877328 on Build GRCh38
rs1016666056 Chr.17: 43877266-43877266 on Build GRCh38
rs1053533633 Chr.17: 43877459-43877459 on Build GRCh38
rs896910804 Chr.17: 43877591-43877591 on Build GRCh38
rs571703965 Chr.17: 43877319-43877319 on Build GRCh38
rs566919894 Chr.17: 43877585-43877585 on Build GRCh38
rs911032378 Chr.17: 43877276-43877276 on Build GRCh38
rs1051545306 Chr.17: 43877577-43877577 on Build GRCh38
rs889936677 Chr.17: 43877548-43877548 on Build GRCh38
rs764869631 Chr.17: 43877606-43877606 on Build GRCh38
rs527694631 Chr.17: 43877644-43877644 on Build GRCh38
rs1010229296 Chr.17: 43877373-43877374 on Build GRCh38
rs761532365 Chr.17: 43877424-43877424 on Build GRCh38
rs533504919 Chr.17: 43877541-43877541 on Build GRCh38
rs967900395 Chr.17: 43877351-43877351 on Build GRCh38
rs231522 Chr.17: 43877234-43877234 on Build GRCh38
rs1054661564 Chr.17: 43877362-43877362 on Build GRCh38
rs916420944 Chr.17: 43877345-43877345 on Build GRCh38
rs531454984 Chr.17: 43877451-43877451 on Build GRCh38
rs1010407474 Chr.17: 43877564-43877564 on Build GRCh38
rs926061770 Chr.17: 43877386-43877386 on Build GRCh38
rs906003475 Chr.17: 43877336-43877336 on Build GRCh38

View Full Product Details