Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 43881007-43881503 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MPP2
Entrez Gene ID
4355
Gene Name
membrane palmitoylated protein 2
Gene Aliases
DLG2
UniGene
Hs.514208
Gene Chromosome Location
Chr.17: 43875357-43909711 on Build GRCh38
Transcripts
NM_001278370.1 NM_001278371.1 NM_001278372.1 NM_001278373.1 NM_001278374.1 NM_001278375.1 NM_001278376.2 NM_001278381.1 NM_005374.4 XM_011524827.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6714798 c.917T>C p.L306P Chr.17: 43881318-43881318 on Build GRCh38
4919357 c.863A>C p.Q288P Chr.17: 43881480-43881480 on Build GRCh38
6714801 c.1002C>T p.C334C Chr.17: 43881148-43881148 on Build GRCh38
4267501 c.826G>A p.E276K Chr.17: 43881337-43881337 on Build GRCh38
4558469 c.841G>A p.D281N Chr.17: 43881502-43881502 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755718411 Chr.17: 43881434-43881434 on Build GRCh38
rs901943034 Chr.17: 43881439-43881439 on Build GRCh38
rs543822481 Chr.17: 43881417-43881417 on Build GRCh38
rs976276330 Chr.17: 43881269-43881269 on Build GRCh38
rs766998382 Chr.17: 43881363-43881363 on Build GRCh38
rs903036441 Chr.17: 43881160-43881160 on Build GRCh38
rs764410772 Chr.17: 43881086-43881086 on Build GRCh38
rs546398187 Chr.17: 43881076-43881076 on Build GRCh38
rs535812148 Chr.17: 43881029-43881029 on Build GRCh38
rs771548976 Chr.17: 43881462-43881462 on Build GRCh38
rs752423998 Chr.17: 43881268-43881268 on Build GRCh38
rs894392201 Chr.17: 43881202-43881202 on Build GRCh38
rs373647309 Chr.17: 43881299-43881299 on Build GRCh38
rs757662744 Chr.17: 43881120-43881120 on Build GRCh38
rs985340470 Chr.17: 43881232-43881232 on Build GRCh38
rs767091436 Chr.17: 43881070-43881070 on Build GRCh38
rs755578453 Chr.17: 43881430-43881431 on Build GRCh38
rs67843533 Chr.17: 43881448-43881448 on Build GRCh38
rs771848334 Chr.17: 43881185-43881185 on Build GRCh38
rs370958204 Chr.17: 43881150-43881150 on Build GRCh38
rs753803946 Chr.17: 43881382-43881382 on Build GRCh38
rs761223680 Chr.17: 43881362-43881362 on Build GRCh38
rs768070525 Chr.17: 43881121-43881121 on Build GRCh38
rs747987226 Chr.17: 43881046-43881046 on Build GRCh38
rs185295203 Chr.17: 43881075-43881075 on Build GRCh38
rs923510493 Chr.17: 43881072-43881072 on Build GRCh38
rs374546641 Chr.17: 43881241-43881241 on Build GRCh38
rs759199112 Chr.17: 43881056-43881065 on Build GRCh38
rs779210820 Chr.17: 43881267-43881267 on Build GRCh38
rs772644031 Chr.17: 43881074-43881074 on Build GRCh38
rs780418329 Chr.17: 43881157-43881157 on Build GRCh38
rs928409671 Chr.17: 43881288-43881288 on Build GRCh38
rs751973227 Chr.17: 43881119-43881119 on Build GRCh38
rs769515223 Chr.17: 43881213-43881213 on Build GRCh38
rs781123034 Chr.17: 43881424-43881424 on Build GRCh38
rs774509683 Chr.17: 43881214-43881214 on Build GRCh38
rs772604062 Chr.17: 43881356-43881356 on Build GRCh38
rs751432334 Chr.17: 43881055-43881057 on Build GRCh38
rs766518786 Chr.17: 43881244-43881244 on Build GRCh38
rs745359371 Chr.17: 43881426-43881426 on Build GRCh38
rs919340410 Chr.17: 43881405-43881405 on Build GRCh38
rs770872796 Chr.17: 43881289-43881289 on Build GRCh38
rs777499053 Chr.17: 43881409-43881409 on Build GRCh38
rs770472087 Chr.17: 43881201-43881201 on Build GRCh38
rs878945334 Chr.17: 43881233-43881233 on Build GRCh38
rs752455203 Chr.17: 43881193-43881193 on Build GRCh38
rs747002600 Chr.17: 43881284-43881284 on Build GRCh38
rs909304477 Chr.17: 43881123-43881123 on Build GRCh38
rs56196501 Chr.17: 43881215-43881215 on Build GRCh38
rs769270640 Chr.17: 43881333-43881333 on Build GRCh38
rs893065387 Chr.17: 43881085-43881085 on Build GRCh38
rs758282842 Chr.17: 43881277-43881277 on Build GRCh38
rs947303739 Chr.17: 43881380-43881380 on Build GRCh38
rs749562275 Chr.17: 43881161-43881161 on Build GRCh38
rs372591611 Chr.17: 43881061-43881061 on Build GRCh38
rs777660208 Chr.17: 43881282-43881282 on Build GRCh38
rs745384701 Chr.17: 43881316-43881316 on Build GRCh38
rs867281597 Chr.17: 43881134-43881134 on Build GRCh38
rs894403512 Chr.17: 43881410-43881410 on Build GRCh38
rs573008913 Chr.17: 43881148-43881148 on Build GRCh38
rs962554122 Chr.17: 43881258-43881258 on Build GRCh38
rs1040396874 Chr.17: 43881427-43881427 on Build GRCh38
rs777259610 Chr.17: 43881468-43881468 on Build GRCh38
rs972586234 Chr.17: 43881391-43881391 on Build GRCh38
rs368052813 Chr.17: 43881248-43881248 on Build GRCh38
rs370767185 Chr.17: 43881219-43881219 on Build GRCh38
rs377255507 Chr.17: 43881414-43881414 on Build GRCh38
rs773833327 Chr.17: 43881357-43881357 on Build GRCh38
rs762079137 Chr.17: 43881343-43881343 on Build GRCh38
rs370335404 Chr.17: 43881449-43881449 on Build GRCh38
rs759721961 Chr.17: 43881390-43881390 on Build GRCh38
rs999036171 Chr.17: 43881225-43881225 on Build GRCh38
rs376136984 Chr.17: 43881194-43881194 on Build GRCh38
rs775269681 Chr.17: 43881335-43881335 on Build GRCh38
rs753007109 Chr.17: 43881407-43881407 on Build GRCh38
rs908017799 Chr.17: 43881250-43881250 on Build GRCh38
rs146465961 Chr.17: 43881441-43881441 on Build GRCh38
rs956878047 Chr.17: 43881036-43881036 on Build GRCh38
rs770095533 Chr.17: 43881259-43881259 on Build GRCh38
rs557642139 Chr.17: 43881048-43881048 on Build GRCh38
rs778532798 Chr.17: 43881170-43881170 on Build GRCh38
rs765375486 Chr.17: 43881392-43881392 on Build GRCh38
rs200915083 Chr.17: 43881147-43881147 on Build GRCh38
rs747981740 Chr.17: 43881182-43881182 on Build GRCh38
rs763349390 Chr.17: 43881080-43881080 on Build GRCh38
rs942064371 Chr.17: 43881151-43881151 on Build GRCh38
rs779543198 Chr.17: 43881438-43881438 on Build GRCh38
rs201844703 Chr.17: 43881408-43881408 on Build GRCh38
rs200286143 Chr.17: 43881443-43881443 on Build GRCh38
rs981209405 Chr.17: 43881263-43881263 on Build GRCh38
rs370891454 Chr.17: 43881256-43881256 on Build GRCh38
rs755251793 Chr.17: 43881166-43881166 on Build GRCh38
rs369615449 Chr.17: 43881043-43881043 on Build GRCh38
rs564636607 Chr.17: 43881079-43881079 on Build GRCh38
rs776247536 Chr.17: 43881208-43881208 on Build GRCh38
rs1055479544 Chr.17: 43881355-43881355 on Build GRCh38
rs751037540 Chr.17: 43881230-43881230 on Build GRCh38

View Full Product Details