Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32160377-32160878 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KPNA6
Entrez Gene ID
23633
Gene Name
karyopherin subunit alpha 6
Gene Aliases
IPOA7, KPNA7
UniGene
Hs.470588
Gene Chromosome Location
Chr.1: 32108043-32176568 on Build GRCh38
Transcripts
NM_012316.4 XM_005270711.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3488101 c.638C>T p.P213L Chr.1: 32160694-32160694 on Build GRCh38
5633243 c.583G>T p.A195S Chr.1: 32160624-32160624 on Build GRCh38
6251949 c.638G>T p.C213F Chr.1: 32160679-32160679 on Build GRCh38
5791191 c.652C>G p.P218A Chr.1: 32160693-32160693 on Build GRCh38
3664722 c.637C>G p.P213A Chr.1: 32160693-32160693 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs950139744 Chr.1: 32160604-32160604 on Build GRCh38
rs954436201 Chr.1: 32160419-32160419 on Build GRCh38
rs973157279 Chr.1: 32160532-32160532 on Build GRCh38
rs918646494 Chr.1: 32160558-32160558 on Build GRCh38
rs985770161 Chr.1: 32160486-32160486 on Build GRCh38
rs1057142341 Chr.1: 32160469-32160469 on Build GRCh38
rs762144365 Chr.1: 32160741-32160741 on Build GRCh38
rs533648374 Chr.1: 32160755-32160755 on Build GRCh38
rs773258459 Chr.1: 32160715-32160715 on Build GRCh38
rs373811592 Chr.1: 32160751-32160751 on Build GRCh38
rs938644810 Chr.1: 32160446-32160446 on Build GRCh38
rs74401724 Chr.1: 32160512-32160512 on Build GRCh38
rs545157853 Chr.1: 32160438-32160438 on Build GRCh38
rs149299526 Chr.1: 32160843-32160843 on Build GRCh38
rs772078065 Chr.1: 32160723-32160724 on Build GRCh38
rs1012159162 Chr.1: 32160489-32160489 on Build GRCh38
rs747022440 Chr.1: 32160733-32160737 on Build GRCh38
rs776001549 Chr.1: 32160532-32160532 on Build GRCh38
rs572673779 Chr.1: 32160472-32160472 on Build GRCh38
rs77488173 Chr.1: 32160835-32160835 on Build GRCh38
rs746578940 Chr.1: 32160620-32160620 on Build GRCh38
rs775643662 Chr.1: 32160728-32160728 on Build GRCh38
rs148103875 Chr.1: 32160530-32160530 on Build GRCh38
rs1044859892 Chr.1: 32160497-32160497 on Build GRCh38
rs747478809 Chr.1: 32160687-32160687 on Build GRCh38
rs993235205 Chr.1: 32160776-32160776 on Build GRCh38
rs780066570 Chr.1: 32160731-32160732 on Build GRCh38
rs764372340 Chr.1: 32160572-32160572 on Build GRCh38
rs757723333 Chr.1: 32160587-32160587 on Build GRCh38
rs751114633 Chr.1: 32160414-32160414 on Build GRCh38
rs769748870 Chr.1: 32160703-32160703 on Build GRCh38
rs1052311866 Chr.1: 32160841-32160841 on Build GRCh38
rs35495206 Chr.1: 32160425-32160426 on Build GRCh38
rs908645066 Chr.1: 32160428-32160428 on Build GRCh38
rs368776674 Chr.1: 32160748-32160748 on Build GRCh38
rs1030667900 Chr.1: 32160549-32160549 on Build GRCh38
rs370899853 Chr.1: 32160600-32160600 on Build GRCh38
rs560598408 Chr.1: 32160471-32160471 on Build GRCh38
rs1044935821 Chr.1: 32160820-32160820 on Build GRCh38
rs1019313672 Chr.1: 32160677-32160677 on Build GRCh38
rs997303834 Chr.1: 32160769-32160769 on Build GRCh38
rs376794884 Chr.1: 32160590-32160590 on Build GRCh38
rs781088395 Chr.1: 32160669-32160669 on Build GRCh38
rs768370964 Chr.1: 32160636-32160636 on Build GRCh38
rs190061892 Chr.1: 32160836-32160836 on Build GRCh38
rs962772529 Chr.1: 32160527-32160527 on Build GRCh38
rs532015184 Chr.1: 32160612-32160612 on Build GRCh38
rs1017158926 Chr.1: 32160847-32160847 on Build GRCh38
rs750981787 Chr.1: 32160606-32160606 on Build GRCh38
rs57514043 Chr.1: 32160733-32160733 on Build GRCh38
rs866382018 Chr.1: 32160453-32160453 on Build GRCh38
rs754212561 Chr.1: 32160583-32160583 on Build GRCh38
rs765540020 Chr.1: 32160563-32160563 on Build GRCh38
rs938813835 Chr.1: 32160774-32160774 on Build GRCh38
rs1047150217 Chr.1: 32160416-32160416 on Build GRCh38
rs561469397 Chr.1: 32160607-32160607 on Build GRCh38
rs866637173 Chr.1: 32160657-32160657 on Build GRCh38
rs562328248 Chr.1: 32160734-32160734 on Build GRCh38
rs892205595 Chr.1: 32160853-32160853 on Build GRCh38
rs937251273 Chr.1: 32160716-32160716 on Build GRCh38
rs138639071 Chr.1: 32160816-32160816 on Build GRCh38
rs150179311 Chr.1: 32160750-32160750 on Build GRCh38
rs756671249 Chr.1: 32160850-32160850 on Build GRCh38
rs533356795 Chr.1: 32160849-32160849 on Build GRCh38
rs1029834532 Chr.1: 32160797-32160797 on Build GRCh38
rs200622778 Chr.1: 32160560-32160560 on Build GRCh38
rs772040266 Chr.1: 32160723-32160723 on Build GRCh38

View Full Product Details