Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 62551958-62552506 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TLK2
Entrez Gene ID
11011
Gene Name
tousled like kinase 2
Gene Aliases
HsHPK, PKU-ALPHA
UniGene
Hs.445078
Gene Chromosome Location
Chr.17: 62477481-62615481 on Build GRCh38
Transcripts
NM_001284333.1 NM_001284363.1 NM_006852.3 XM_011524215.2 XM_011524216.2 XM_011524217.2 XM_011524218.2 XM_011524220.2 XM_011524221.2 XM_011524222.2 XM_011524223.2 XM_011524224.2 XM_011524225.2 XM_011524226.2 XM_011524227.2 XM_011524228.2 XM_011524229.2 XM_011524231.1 XM_017024044.1 XM_017024045.1 XM_017024046.1 XM_017024047.1 XM_017024048.1 XM_017024049.1 XM_017024050.1 XM_017024051.1 XM_017024052.1 XM_017024053.1 XM_017024054.1 XM_017024055.1 XM_017024056.1 XM_017024057.1 XM_017024058.1 XM_017024059.1 XM_017024060.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7402914 c.621G>A p.Q207Q Chr.17: 62552391-62552391 on Build GRCh38
6358612 c.627+2T>C p.? Chr.17: 62552399-62552399 on Build GRCh38
4776234 c.574G>A p.E192K Chr.17: 62552344-62552344 on Build GRCh38
1640767 c.593delA p.K199fs*16 Chr.17: 62552363-62552363 on Build GRCh38
7188845 c.627+1G>A p.? Chr.17: 62552398-62552398 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746120932 Chr.17: 62552328-62552328 on Build GRCh38
rs991688839 Chr.17: 62552177-62552177 on Build GRCh38
rs766062653 Chr.17: 62552271-62552271 on Build GRCh38
rs891072556 Chr.17: 62552433-62552433 on Build GRCh38
rs754416438 Chr.17: 62552440-62552440 on Build GRCh38
rs373218515 Chr.17: 62552278-62552278 on Build GRCh38
rs189481213 Chr.17: 62552109-62552109 on Build GRCh38
rs967875803 Chr.17: 62552083-62552083 on Build GRCh38
rs573051798 Chr.17: 62552056-62552056 on Build GRCh38
rs996777623 Chr.17: 62551991-62551991 on Build GRCh38
rs773953658 Chr.17: 62552420-62552420 on Build GRCh38
rs761055106 Chr.17: 62552436-62552436 on Build GRCh38
rs376620030 Chr.17: 62552330-62552330 on Build GRCh38
rs575563317 Chr.17: 62552186-62552186 on Build GRCh38
rs762592881 Chr.17: 62552268-62552268 on Build GRCh38
rs1008866903 Chr.17: 62552437-62552437 on Build GRCh38
rs374779927 Chr.17: 62552438-62552438 on Build GRCh38
rs371475757 Chr.17: 62552367-62552367 on Build GRCh38
rs779226351 Chr.17: 62552327-62552327 on Build GRCh38
rs765833346 Chr.17: 62552418-62552418 on Build GRCh38
rs751901297 Chr.17: 62552275-62552275 on Build GRCh38
rs565201710 Chr.17: 62552147-62552147 on Build GRCh38
rs1009350429 Chr.17: 62552047-62552047 on Build GRCh38
rs753079937 Chr.17: 62552291-62552291 on Build GRCh38
rs560932781 Chr.17: 62552297-62552297 on Build GRCh38
rs767956123 Chr.17: 62552290-62552290 on Build GRCh38
rs757369447 Chr.17: 62552449-62552449 on Build GRCh38
rs748026036 Chr.17: 62552338-62552338 on Build GRCh38
rs769841221 Chr.17: 62552363-62552363 on Build GRCh38
rs759231991 Chr.17: 62552427-62552427 on Build GRCh38
rs767759721 Chr.17: 62552428-62552428 on Build GRCh38
rs1029612081 Chr.17: 62552000-62552000 on Build GRCh38
rs890442800 Chr.17: 62552012-62552012 on Build GRCh38
rs543874657 Chr.17: 62552211-62552211 on Build GRCh38
rs374185016 Chr.17: 62552117-62552117 on Build GRCh38
rs764075068 Chr.17: 62552304-62552304 on Build GRCh38
rs949745560 Chr.17: 62552229-62552229 on Build GRCh38
rs917514627 Chr.17: 62552219-62552219 on Build GRCh38
rs958919348 Chr.17: 62552163-62552163 on Build GRCh38
rs750347124 Chr.17: 62552449-62552449 on Build GRCh38
rs545190162 Chr.17: 62552131-62552131 on Build GRCh38
rs1033161737 Chr.17: 62552126-62552126 on Build GRCh38
rs772839759 Chr.17: 62552263-62552263 on Build GRCh38
rs552937704 Chr.17: 62551990-62551990 on Build GRCh38
rs772680744 Chr.17: 62552366-62552366 on Build GRCh38
rs529801466 Chr.17: 62552462-62552462 on Build GRCh38
rs770451777 Chr.17: 62552412-62552412 on Build GRCh38
rs978863346 Chr.17: 62552121-62552121 on Build GRCh38
rs867639215 Chr.17: 62552411-62552411 on Build GRCh38
rs903245632 Chr.17: 62552478-62552478 on Build GRCh38
rs754009737 Chr.17: 62552316-62552316 on Build GRCh38
rs2032738 Chr.17: 62552042-62552042 on Build GRCh38
rs1020709441 Chr.17: 62552474-62552474 on Build GRCh38
rs769131929 Chr.17: 62552252-62552252 on Build GRCh38
rs757529236 Chr.17: 62552322-62552322 on Build GRCh38

View Full Product Details