Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 46938906-46939409 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GOSR2
Entrez Gene ID
9570
Gene Name
golgi SNAP receptor complex member 2
Gene Aliases
Bos1, EPM6, GS27
UniGene
Hs.463278
Gene Chromosome Location
Chr.17: 46923086-46975519 on Build GRCh38
Transcripts
NM_001321133.1 NM_001321134.1 NM_004287.4 NM_054022.3 XM_005257844.3 XM_006722190.3 XM_011525501.2 XM_011525502.2 XM_017025378.1 XM_017025379.1 XM_017025380.1 XM_017025381.1 XM_017025382.1 XM_017025383.1 XM_017025384.1 XM_017025385.1 XM_017025386.1 XM_017025387.1 XM_017025388.1 XM_017025389.1 XM_017025390.1 XM_017025391.1 XM_017025392.1 XM_017025393.1 XM_017025394.1 XM_017025395.1

SNP Detail

SNP ID Chromosome Location
rs201923580 Chr.17: 46939205-46939205 on Build GRCh38
rs1040226915 Chr.17: 46939106-46939106 on Build GRCh38
rs765279001 Chr.17: 46939177-46939177 on Build GRCh38
rs367735669 Chr.17: 46938992-46938992 on Build GRCh38
rs8078556 Chr.17: 46939061-46939061 on Build GRCh38
rs1019748144 Chr.17: 46939382-46939382 on Build GRCh38
rs765576738 Chr.17: 46939214-46939214 on Build GRCh38
rs1012679747 Chr.17: 46938993-46938993 on Build GRCh38
rs1031804398 Chr.17: 46939263-46939263 on Build GRCh38
rs1044181454 Chr.17: 46939230-46939230 on Build GRCh38
rs1023112805 Chr.17: 46938996-46938996 on Build GRCh38
rs78693564 Chr.17: 46939046-46939046 on Build GRCh38
rs775729401 Chr.17: 46939271-46939271 on Build GRCh38
rs919051435 Chr.17: 46939121-46939121 on Build GRCh38
rs139209716 Chr.17: 46939057-46939057 on Build GRCh38
rs1023551346 Chr.17: 46939129-46939129 on Build GRCh38
rs1048723073 Chr.17: 46938966-46938966 on Build GRCh38
rs990769151 Chr.17: 46939047-46939047 on Build GRCh38
rs991787122 Chr.17: 46939099-46939099 on Build GRCh38
rs887467904 Chr.17: 46939204-46939204 on Build GRCh38
rs139652892 Chr.17: 46939231-46939231 on Build GRCh38
rs985583638 Chr.17: 46939071-46939071 on Build GRCh38
rs886053079 Chr.17: 46939092-46939092 on Build GRCh38
rs965453562 Chr.17: 46939384-46939384 on Build GRCh38
rs891001127 Chr.17: 46939009-46939009 on Build GRCh38
rs886053080 Chr.17: 46939172-46939172 on Build GRCh38
rs754380153 Chr.17: 46939070-46939070 on Build GRCh38
rs761117888 Chr.17: 46939001-46939001 on Build GRCh38
rs985854411 Chr.17: 46939351-46939351 on Build GRCh38
rs924334643 Chr.17: 46939191-46939191 on Build GRCh38
rs143006022 Chr.17: 46939354-46939354 on Build GRCh38
rs867280023 Chr.17: 46939316-46939316 on Build GRCh38
rs368780514 Chr.17: 46938956-46938956 on Build GRCh38
rs537489434 Chr.17: 46939012-46939012 on Build GRCh38
rs1002984564 Chr.17: 46939019-46939019 on Build GRCh38
rs912384800 Chr.17: 46939371-46939371 on Build GRCh38
rs954097897 Chr.17: 46939318-46939318 on Build GRCh38
rs971561894 Chr.17: 46939172-46939172 on Build GRCh38
rs944554880 Chr.17: 46939094-46939094 on Build GRCh38
rs181200484 Chr.17: 46939287-46939287 on Build GRCh38
rs189095245 Chr.17: 46939088-46939088 on Build GRCh38
rs376212419 Chr.17: 46939056-46939057 on Build GRCh38
rs1022272650 Chr.17: 46939308-46939308 on Build GRCh38
rs955808922 Chr.17: 46939206-46939206 on Build GRCh38
rs755362352 Chr.17: 46939083-46939083 on Build GRCh38
rs1003035029 Chr.17: 46939246-46939246 on Build GRCh38
rs184197582 Chr.17: 46938961-46938961 on Build GRCh38
rs529000124 Chr.17: 46939035-46939035 on Build GRCh38
rs562694673 Chr.17: 46939289-46939289 on Build GRCh38
rs929244962 Chr.17: 46939147-46939147 on Build GRCh38

View Full Product Details