Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 50464408-50464906 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSF2
Entrez Gene ID
80221
Gene Name
acyl-CoA synthetase family member 2
Gene Aliases
ACSMW, AVYV493
UniGene
Hs.288959
Gene Chromosome Location
Chr.17: 50426158-50474845 on Build GRCh38
Transcripts
NM_001288968.1 NM_001288969.1 NM_001288970.1 NM_001288971.1 NM_001288972.1 NM_025149.5 XM_006722110.3 XM_011525294.1 NM_001267.2 XM_011524214.2

Gene Symbol
CHAD
Entrez Gene ID
1101
Gene Name
chondroadherin
Gene Aliases
SLRR4A
UniGene
Hs.97220
Gene Chromosome Location
Chr.17: 50464492-50469881 on Build GRCh38
Transcripts
NM_001288968.1 NM_001288969.1 NM_001288970.1 NM_001288971.1 NM_001288972.1 NM_025149.5 XM_006722110.3 XM_011525294.1 NM_001267.2 XM_011524214.2

SNP Detail

SNP ID Chromosome Location
rs1026070446 Chr.17: 50464772-50464773 on Build GRCh38
rs796126022 Chr.17: 50464777-50464778 on Build GRCh38
rs1022224274 Chr.17: 50464743-50464743 on Build GRCh38
rs1052255 Chr.17: 50464578-50464578 on Build GRCh38
rs559060672 Chr.17: 50464775-50464775 on Build GRCh38
rs1007803826 Chr.17: 50464856-50464856 on Build GRCh38
rs756976860 Chr.17: 50464493-50464493 on Build GRCh38
rs915565212 Chr.17: 50464774-50464774 on Build GRCh38
rs762545921 Chr.17: 50464518-50464518 on Build GRCh38
rs561431292 Chr.17: 50464606-50464606 on Build GRCh38
rs558652489 Chr.17: 50464528-50464528 on Build GRCh38
rs771153191 Chr.17: 50464451-50464451 on Build GRCh38
rs775319800 Chr.17: 50464508-50464508 on Build GRCh38
rs528886197 Chr.17: 50464699-50464699 on Build GRCh38
rs958370471 Chr.17: 50464747-50464747 on Build GRCh38
rs146704733 Chr.17: 50464469-50464469 on Build GRCh38
rs565678266 Chr.17: 50464770-50464771 on Build GRCh38
rs187050319 Chr.17: 50464573-50464573 on Build GRCh38
rs150309436 Chr.17: 50464641-50464641 on Build GRCh38
rs191492703 Chr.17: 50464708-50464708 on Build GRCh38
rs902057383 Chr.17: 50464860-50464860 on Build GRCh38
rs905466196 Chr.17: 50464815-50464815 on Build GRCh38
rs763678229 Chr.17: 50464664-50464664 on Build GRCh38
rs1013831362 Chr.17: 50464600-50464600 on Build GRCh38
rs762449295 Chr.17: 50464544-50464544 on Build GRCh38
rs781027982 Chr.17: 50464769-50464770 on Build GRCh38
rs749072009 Chr.17: 50464489-50464489 on Build GRCh38
rs912494488 Chr.17: 50464823-50464823 on Build GRCh38
rs144893851 Chr.17: 50464659-50464659 on Build GRCh38
rs566193091 Chr.17: 50464538-50464538 on Build GRCh38
rs113020804 Chr.17: 50464778-50464778 on Build GRCh38
rs889390014 Chr.17: 50464748-50464748 on Build GRCh38
rs368836827 Chr.17: 50464771-50464772 on Build GRCh38
rs547285186 Chr.17: 50464767-50464768 on Build GRCh38
rs570375504 Chr.17: 50464472-50464472 on Build GRCh38
rs60053197 Chr.17: 50464778-50464779 on Build GRCh38
rs995228917 Chr.17: 50464576-50464576 on Build GRCh38
rs934516171 Chr.17: 50464777-50464778 on Build GRCh38
rs78392850 Chr.17: 50464822-50464822 on Build GRCh38
rs77557245 Chr.17: 50464684-50464684 on Build GRCh38
rs548404227 Chr.17: 50464590-50464590 on Build GRCh38
rs1010914079 Chr.17: 50464702-50464702 on Build GRCh38
rs572925085 Chr.17: 50464777-50464777 on Build GRCh38
rs921071566 Chr.17: 50464468-50464468 on Build GRCh38
rs889107817 Chr.17: 50464853-50464853 on Build GRCh38
rs948649199 Chr.17: 50464775-50464776 on Build GRCh38
rs939897601 Chr.17: 50464858-50464858 on Build GRCh38
rs769547926 Chr.17: 50464456-50464456 on Build GRCh38
rs12449644 Chr.17: 50464583-50464583 on Build GRCh38
rs772073697 Chr.17: 50464589-50464589 on Build GRCh38
rs946779684 Chr.17: 50464827-50464827 on Build GRCh38
rs953595910 Chr.17: 50464464-50464464 on Build GRCh38
rs202209206 Chr.17: 50464774-50464775 on Build GRCh38
rs912253496 Chr.17: 50464479-50464479 on Build GRCh38
rs577577426 Chr.17: 50464771-50464771 on Build GRCh38
rs761691099 Chr.17: 50464831-50464831 on Build GRCh38
rs1032304374 Chr.17: 50464714-50464714 on Build GRCh38
rs140186676 Chr.17: 50464480-50464480 on Build GRCh38
rs79519301 Chr.17: 50464830-50464830 on Build GRCh38
rs61391073 Chr.17: 50464773-50464774 on Build GRCh38
rs981349920 Chr.17: 50464776-50464777 on Build GRCh38
rs539784859 Chr.17: 50464772-50464772 on Build GRCh38
rs755736467 Chr.17: 50464477-50464477 on Build GRCh38
rs1004068470 Chr.17: 50464660-50464660 on Build GRCh38
rs75281762 Chr.17: 50464779-50464779 on Build GRCh38

View Full Product Details