Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81250793-81251291 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC38A10
Entrez Gene ID
124565
Gene Name
solute carrier family 38 member 10
Gene Aliases
PP1744
UniGene
Hs.352240
Gene Chromosome Location
Chr.17: 81244811-81295433 on Build GRCh38
Transcripts
NM_001037984.2 NM_138570.3 XM_005257019.1 XM_011524288.1 XM_011524289.1 XM_011524290.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7341718 c.2282_2283insA p.H762fs*10 Chr.17: 81251275-81251276 on Build GRCh38
5387624 c.2275C>T p.P759S Chr.17: 81251283-81251283 on Build GRCh38
3388290 c.2290C>T p.R764W Chr.17: 81251268-81251268 on Build GRCh38
1741059 c.2279_2280insA p.F760fs*2 Chr.17: 81251278-81251279 on Build GRCh38
5414466 c.2332G>A p.A778T Chr.17: 81251226-81251226 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs190975573 Chr.17: 81250833-81250833 on Build GRCh38
rs779947917 Chr.17: 81250971-81250971 on Build GRCh38
rs748036710 Chr.17: 81251044-81251044 on Build GRCh38
rs140396168 Chr.17: 81251221-81251221 on Build GRCh38
rs116990218 Chr.17: 81251240-81251240 on Build GRCh38
rs745587399 Chr.17: 81250881-81250881 on Build GRCh38
rs944859808 Chr.17: 81251004-81251004 on Build GRCh38
rs187896866 Chr.17: 81251033-81251033 on Build GRCh38
rs868341831 Chr.17: 81251242-81251242 on Build GRCh38
rs578184166 Chr.17: 81251213-81251213 on Build GRCh38
rs751942651 Chr.17: 81251170-81251170 on Build GRCh38
rs781365446 Chr.17: 81251174-81251174 on Build GRCh38
rs983461132 Chr.17: 81251202-81251202 on Build GRCh38
rs908684314 Chr.17: 81250995-81250995 on Build GRCh38
rs556660475 Chr.17: 81251065-81251065 on Build GRCh38
rs149950205 Chr.17: 81250863-81250863 on Build GRCh38
rs536705041 Chr.17: 81250977-81250977 on Build GRCh38
rs770900028 Chr.17: 81251205-81251205 on Build GRCh38
rs386799869 Chr.17: 81251226-81251292 on Build GRCh38
rs746748268 Chr.17: 81250984-81250984 on Build GRCh38
rs920583263 Chr.17: 81250973-81250973 on Build GRCh38
rs926048329 Chr.17: 81251036-81251036 on Build GRCh38
rs759342652 Chr.17: 81251220-81251220 on Build GRCh38
rs775303765 Chr.17: 81251222-81251222 on Build GRCh38
rs368971915 Chr.17: 81251099-81251099 on Build GRCh38
rs763425404 Chr.17: 81251261-81251261 on Build GRCh38
rs113882474 Chr.17: 81251247-81251247 on Build GRCh38
rs745508972 Chr.17: 81251159-81251159 on Build GRCh38
rs557587675 Chr.17: 81251262-81251262 on Build GRCh38
rs766481389 Chr.17: 81251187-81251187 on Build GRCh38
rs911942730 Chr.17: 81251216-81251216 on Build GRCh38
rs565704741 Chr.17: 81251231-81251231 on Build GRCh38
rs372419572 Chr.17: 81251196-81251196 on Build GRCh38
rs569926923 Chr.17: 81251115-81251115 on Build GRCh38
rs374319971 Chr.17: 81251268-81251268 on Build GRCh38
rs970381470 Chr.17: 81250879-81250879 on Build GRCh38
rs757975420 Chr.17: 81250956-81250956 on Build GRCh38
rs994631578 Chr.17: 81250958-81250958 on Build GRCh38
rs998516428 Chr.17: 81251096-81251096 on Build GRCh38
rs755370875 Chr.17: 81251267-81251267 on Build GRCh38
rs934716151 Chr.17: 81251037-81251037 on Build GRCh38
rs977058924 Chr.17: 81251212-81251212 on Build GRCh38
rs561783472 Chr.17: 81250948-81250948 on Build GRCh38
rs1031406793 Chr.17: 81250832-81250832 on Build GRCh38
rs552068416 Chr.17: 81251135-81251135 on Build GRCh38
rs755244392 Chr.17: 81251173-81251173 on Build GRCh38
rs67972063 Chr.17: 81251226-81251226 on Build GRCh38
rs780346933 Chr.17: 81251164-81251164 on Build GRCh38
rs895613878 Chr.17: 81251055-81251065 on Build GRCh38
rs895560855 Chr.17: 81251108-81251108 on Build GRCh38
rs965619253 Chr.17: 81251012-81251012 on Build GRCh38
rs11557647 Chr.17: 81251035-81251035 on Build GRCh38
rs149029325 Chr.17: 81251194-81251194 on Build GRCh38
rs969970183 Chr.17: 81251162-81251162 on Build GRCh38
rs1022844851 Chr.17: 81250864-81250864 on Build GRCh38
rs776731292 Chr.17: 81251034-81251034 on Build GRCh38
rs976997639 Chr.17: 81251022-81251022 on Build GRCh38
rs950463188 Chr.17: 81251171-81251173 on Build GRCh38
rs113562695 Chr.17: 81251256-81251256 on Build GRCh38
rs973376969 Chr.17: 81250917-81250917 on Build GRCh38
rs776197772 Chr.17: 81251257-81251257 on Build GRCh38
rs182182292 Chr.17: 81250978-81250978 on Build GRCh38
rs145108422 Chr.17: 81251190-81251190 on Build GRCh38
rs904177941 Chr.17: 81251071-81251071 on Build GRCh38
rs563694726 Chr.17: 81251138-81251138 on Build GRCh38
rs946984256 Chr.17: 81251045-81251045 on Build GRCh38
rs66535907 Chr.17: 81251200-81251200 on Build GRCh38
rs533659893 Chr.17: 81251120-81251120 on Build GRCh38
rs921307279 Chr.17: 81251025-81251025 on Build GRCh38
rs774060952 Chr.17: 81251206-81251206 on Build GRCh38
rs911999383 Chr.17: 81250985-81250985 on Build GRCh38
rs1022632279 Chr.17: 81250919-81250919 on Build GRCh38
rs970126412 Chr.17: 81250922-81250922 on Build GRCh38
rs72860022 Chr.17: 81251185-81251185 on Build GRCh38
rs1052891340 Chr.17: 81251045-81251045 on Build GRCh38
rs1031442658 Chr.17: 81250825-81250825 on Build GRCh38

View Full Product Details