Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 40624101-40624599 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RIMS3
Entrez Gene ID
9783
Gene Name
regulating synaptic membrane exocytosis 3
Gene Aliases
NIM3, RIM3
UniGene
Hs.654808
Gene Chromosome Location
Chr.1: 40620680-40692104 on Build GRCh38
Transcripts
NM_014747.2 XM_011542479.2

SNP Detail

SNP ID Chromosome Location
rs890284037 Chr.1: 40624447-40624447 on Build GRCh38
rs565847268 Chr.1: 40624380-40624380 on Build GRCh38
rs114969131 Chr.1: 40624177-40624177 on Build GRCh38
rs112570698 Chr.1: 40624279-40624279 on Build GRCh38
rs897507148 Chr.1: 40624391-40624391 on Build GRCh38
rs992649311 Chr.1: 40624256-40624256 on Build GRCh38
rs567441918 Chr.1: 40624440-40624440 on Build GRCh38
rs938231410 Chr.1: 40624228-40624228 on Build GRCh38
rs898094547 Chr.1: 40624416-40624416 on Build GRCh38
rs890173961 Chr.1: 40624399-40624399 on Build GRCh38
rs995085444 Chr.1: 40624422-40624422 on Build GRCh38
rs1017865259 Chr.1: 40624466-40624466 on Build GRCh38
rs982207293 Chr.1: 40624202-40624202 on Build GRCh38
rs1050143084 Chr.1: 40624379-40624379 on Build GRCh38
rs7520333 Chr.1: 40624578-40624578 on Build GRCh38
rs994852889 Chr.1: 40624122-40624122 on Build GRCh38
rs775767756 Chr.1: 40624350-40624350 on Build GRCh38
rs761123890 Chr.1: 40624175-40624181 on Build GRCh38
rs532393040 Chr.1: 40624307-40624307 on Build GRCh38
rs535994111 Chr.1: 40624480-40624480 on Build GRCh38
rs905201943 Chr.1: 40624451-40624451 on Build GRCh38
rs747061392 Chr.1: 40624524-40624524 on Build GRCh38
rs991470800 Chr.1: 40624299-40624299 on Build GRCh38
rs993096332 Chr.1: 40624515-40624515 on Build GRCh38
rs755811716 Chr.1: 40624497-40624497 on Build GRCh38
rs141219643 Chr.1: 40624552-40624552 on Build GRCh38
rs1035531214 Chr.1: 40624487-40624487 on Build GRCh38
rs978642576 Chr.1: 40624554-40624554 on Build GRCh38
rs978424799 Chr.1: 40624301-40624301 on Build GRCh38
rs552220570 Chr.1: 40624308-40624308 on Build GRCh38
rs547619518 Chr.1: 40624410-40624410 on Build GRCh38
rs752520755 Chr.1: 40624411-40624411 on Build GRCh38
rs966730192 Chr.1: 40624529-40624529 on Build GRCh38
rs1007428170 Chr.1: 40624458-40624458 on Build GRCh38
rs766446222 Chr.1: 40624127-40624127 on Build GRCh38
rs945974282 Chr.1: 40624298-40624298 on Build GRCh38
rs772129003 Chr.1: 40624200-40624200 on Build GRCh38
rs966761418 Chr.1: 40624187-40624187 on Build GRCh38
rs969499926 Chr.1: 40624162-40624162 on Build GRCh38
rs1017562210 Chr.1: 40624125-40624125 on Build GRCh38
rs1035075215 Chr.1: 40624178-40624178 on Build GRCh38
rs751390790 Chr.1: 40624153-40624153 on Build GRCh38
rs188210579 Chr.1: 40624405-40624405 on Build GRCh38
rs929042794 Chr.1: 40624408-40624408 on Build GRCh38
rs11208546 Chr.1: 40624192-40624192 on Build GRCh38
rs1027638564 Chr.1: 40624150-40624150 on Build GRCh38
rs1002163902 Chr.1: 40624455-40624455 on Build GRCh38
rs1020099739 Chr.1: 40624525-40624525 on Build GRCh38
rs767279294 Chr.1: 40624302-40624302 on Build GRCh38
rs928105364 Chr.1: 40624217-40624217 on Build GRCh38
rs960146661 Chr.1: 40624204-40624204 on Build GRCh38

View Full Product Details