Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 79902728-79903233 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PQLC1
Entrez Gene ID
80148
Gene Name
PQ loop repeat containing 1
Gene Aliases
-
UniGene
Hs.288284
Gene Chromosome Location
Chr.18: 79902420-79951665 on Build GRCh38
Transcripts
NM_001146343.1 NM_001146345.1 NM_025078.4 XM_005266770.2 XM_011526188.1 XM_017025997.1 XM_017025998.1 XM_017026001.1 XM_017026003.1 XM_017026004.1 XM_017026005.1

SNP Detail

SNP ID Chromosome Location
rs561991074 Chr.18: 79902921-79902921 on Build GRCh38
rs567914004 Chr.18: 79903092-79903092 on Build GRCh38
rs916700359 Chr.18: 79903164-79903164 on Build GRCh38
rs111394122 Chr.18: 79903112-79903112 on Build GRCh38
rs569910146 Chr.18: 79902752-79902752 on Build GRCh38
rs944547040 Chr.18: 79902785-79902785 on Build GRCh38
rs571659851 Chr.18: 79903185-79903185 on Build GRCh38
rs553205673 Chr.18: 79902825-79902825 on Build GRCh38
rs1025791706 Chr.18: 79903196-79903196 on Build GRCh38
rs33945058 Chr.18: 79903071-79903072 on Build GRCh38
rs965754455 Chr.18: 79903063-79903063 on Build GRCh38
rs962921659 Chr.18: 79902875-79902875 on Build GRCh38
rs115495003 Chr.18: 79902881-79902881 on Build GRCh38
rs987317205 Chr.18: 79903108-79903108 on Build GRCh38
rs1045689218 Chr.18: 79903106-79903106 on Build GRCh38
rs935799199 Chr.18: 79903069-79903069 on Build GRCh38
rs895354770 Chr.18: 79903026-79903026 on Build GRCh38
rs868335853 Chr.18: 79903014-79903014 on Build GRCh38
rs879160946 Chr.18: 79902963-79902963 on Build GRCh38
rs1056247346 Chr.18: 79903145-79903145 on Build GRCh38
rs983112090 Chr.18: 79902886-79902886 on Build GRCh38
rs554705471 Chr.18: 79903010-79903010 on Build GRCh38
rs913152321 Chr.18: 79903104-79903104 on Build GRCh38
rs1055581664 Chr.18: 79903163-79903163 on Build GRCh38
rs367853563 Chr.18: 79903156-79903156 on Build GRCh38
rs537273482 Chr.18: 79903070-79903071 on Build GRCh38
rs1015018590 Chr.18: 79902942-79902942 on Build GRCh38
rs945798702 Chr.18: 79902847-79902847 on Build GRCh38
rs944477072 Chr.18: 79902914-79902914 on Build GRCh38
rs116944853 Chr.18: 79902751-79902751 on Build GRCh38
rs953273839 Chr.18: 79903099-79903099 on Build GRCh38
rs928990555 Chr.18: 79902756-79902756 on Build GRCh38
rs781641418 Chr.18: 79903118-79903118 on Build GRCh38
rs534716063 Chr.18: 79903142-79903142 on Build GRCh38
rs1001119816 Chr.18: 79902781-79902781 on Build GRCh38
rs1034958168 Chr.18: 79902864-79902864 on Build GRCh38
rs970783446 Chr.18: 79902790-79902790 on Build GRCh38
rs994954452 Chr.18: 79903084-79903089 on Build GRCh38
rs755242758 Chr.18: 79903105-79903105 on Build GRCh38
rs921818130 Chr.18: 79902957-79902957 on Build GRCh38
rs921706438 Chr.18: 79903065-79903065 on Build GRCh38
rs781357000 Chr.18: 79902903-79902903 on Build GRCh38
rs935868286 Chr.18: 79902925-79902925 on Build GRCh38
rs3047785 Chr.18: 79903076-79903077 on Build GRCh38
rs906798269 Chr.18: 79903193-79903193 on Build GRCh38
rs567581834 Chr.18: 79902794-79902794 on Build GRCh38
rs558025258 Chr.18: 79902850-79902850 on Build GRCh38
rs3744883 Chr.18: 79903046-79903046 on Build GRCh38
rs576288418 Chr.18: 79902855-79902855 on Build GRCh38
rs867138355 Chr.18: 79903049-79903049 on Build GRCh38
rs963220336 Chr.18: 79902944-79902944 on Build GRCh38
rs536566964 Chr.18: 79902920-79902920 on Build GRCh38
rs990018108 Chr.18: 79903070-79903070 on Build GRCh38
rs911814144 Chr.18: 79903111-79903111 on Build GRCh38
rs974704915 Chr.18: 79902876-79902876 on Build GRCh38
rs995628055 Chr.18: 79903189-79903189 on Build GRCh38
rs1010235034 Chr.18: 79902949-79902949 on Build GRCh38
rs3744884 Chr.18: 79902962-79902962 on Build GRCh38
rs537386904 Chr.18: 79902755-79902755 on Build GRCh38
rs116527960 Chr.18: 79902982-79902982 on Build GRCh38
rs945726611 Chr.18: 79902988-79902988 on Build GRCh38
rs1038854173 Chr.18: 79902917-79902917 on Build GRCh38
rs371187969 Chr.18: 79903031-79903031 on Build GRCh38
rs1000550890 Chr.18: 79902820-79902820 on Build GRCh38
rs147633148 Chr.18: 79902834-79902834 on Build GRCh38
rs973305999 Chr.18: 79902808-79902808 on Build GRCh38
rs1047428709 Chr.18: 79902759-79902759 on Build GRCh38
rs950352096 Chr.18: 79902884-79902884 on Build GRCh38
rs1033007174 Chr.18: 79902786-79902786 on Build GRCh38
rs539170434 Chr.18: 79903186-79903186 on Build GRCh38
rs991240107 Chr.18: 79902787-79902787 on Build GRCh38
rs971509026 Chr.18: 79902854-79902854 on Build GRCh38
rs534988778 Chr.18: 79902819-79902819 on Build GRCh38
rs931639412 Chr.18: 79902827-79902827 on Build GRCh38
rs764689733 Chr.18: 79902851-79902851 on Build GRCh38
rs974875232 Chr.18: 79903064-79903064 on Build GRCh38
rs907701512 Chr.18: 79902800-79902800 on Build GRCh38
rs963780415 Chr.18: 79903078-79903078 on Build GRCh38
rs911639029 Chr.18: 79902889-79902889 on Build GRCh38
rs941850253 Chr.18: 79902807-79902807 on Build GRCh38
rs748285234 Chr.18: 79903180-79903180 on Build GRCh38
rs911586547 Chr.18: 79903062-79903062 on Build GRCh38
rs116377797 Chr.18: 79903017-79903017 on Build GRCh38
rs891727492 Chr.18: 79902943-79902943 on Build GRCh38
rs549634029 Chr.18: 79902788-79902788 on Build GRCh38
rs573573861 Chr.18: 79902939-79902939 on Build GRCh38
rs1029242062 Chr.18: 79903086-79903086 on Build GRCh38
rs1041422603 Chr.18: 79903003-79903003 on Build GRCh38
rs112346830 Chr.18: 79903011-79903011 on Build GRCh38
rs532907568 Chr.18: 79902997-79902997 on Build GRCh38
rs374469059 Chr.18: 79902835-79902835 on Build GRCh38
rs911648936 Chr.18: 79902760-79902760 on Build GRCh38
rs142298803 Chr.18: 79903032-79903032 on Build GRCh38

View Full Product Details