Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 79918967-79919457 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PQLC1
Entrez Gene ID
80148
Gene Name
PQ loop repeat containing 1
Gene Aliases
-
UniGene
Hs.288284
Gene Chromosome Location
Chr.18: 79902420-79951665 on Build GRCh38
Transcripts
NM_001146343.1 NM_001146345.1 NM_025078.4 XM_005266770.2 XM_011526188.1 XM_017025997.1 XM_017025998.1 XM_017025999.1 XM_017026001.1 XM_017026003.1 XM_017026004.1 XM_017026005.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7112879 c.411C>G p.F137L Chr.18: 79919381-79919381 on Build GRCh38
4717342 c.567G>A p.Q189Q Chr.18: 79919225-79919225 on Build GRCh38
474064 c.438C>A p.Y146* Chr.18: 79919354-79919354 on Build GRCh38
6433592 c.415C>A p.Q139K Chr.18: 79919377-79919377 on Build GRCh38
6328864 c.563C>A p.P188H Chr.18: 79919229-79919229 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs977392528 Chr.18: 79919130-79919130 on Build GRCh38
rs1036409650 Chr.18: 79919021-79919021 on Build GRCh38
rs924413963 Chr.18: 79919405-79919405 on Build GRCh38
rs759689625 Chr.18: 79919134-79919134 on Build GRCh38
rs767377080 Chr.18: 79919309-79919309 on Build GRCh38
rs866012471 Chr.18: 79919201-79919201 on Build GRCh38
rs1000199129 Chr.18: 79919104-79919104 on Build GRCh38
rs760659634 Chr.18: 79919315-79919315 on Build GRCh38
rs888840002 Chr.18: 79919317-79919317 on Build GRCh38
rs746417508 Chr.18: 79919287-79919287 on Build GRCh38
rs372250799 Chr.18: 79919330-79919330 on Build GRCh38
rs770487880 Chr.18: 79919331-79919331 on Build GRCh38
rs867346057 Chr.18: 79919024-79919024 on Build GRCh38
rs910067113 Chr.18: 79919236-79919236 on Build GRCh38
rs781186428 Chr.18: 79919173-79919173 on Build GRCh38
rs534242382 Chr.18: 79919059-79919059 on Build GRCh38
rs190400986 Chr.18: 79919061-79919061 on Build GRCh38
rs140411049 Chr.18: 79919321-79919321 on Build GRCh38
rs771345678 Chr.18: 79919279-79919279 on Build GRCh38
rs897524797 Chr.18: 79919058-79919058 on Build GRCh38
rs180963171 Chr.18: 79919001-79919001 on Build GRCh38
rs750078700 Chr.18: 79919077-79919077 on Build GRCh38
rs200664421 Chr.18: 79919259-79919259 on Build GRCh38
rs188473343 Chr.18: 79919106-79919106 on Build GRCh38
rs1004489986 Chr.18: 79919015-79919015 on Build GRCh38
rs768553253 Chr.18: 79919298-79919298 on Build GRCh38
rs752985716 Chr.18: 79919150-79919150 on Build GRCh38
rs948901046 Chr.18: 79918999-79918999 on Build GRCh38
rs199805905 Chr.18: 79919338-79919338 on Build GRCh38
rs760068053 Chr.18: 79919217-79919217 on Build GRCh38
rs187873662 Chr.18: 79919342-79919342 on Build GRCh38
rs770741798 Chr.18: 79919401-79919401 on Build GRCh38
rs372882177 Chr.18: 79919433-79919433 on Build GRCh38
rs777206274 Chr.18: 79919214-79919214 on Build GRCh38
rs971690969 Chr.18: 79919022-79919022 on Build GRCh38
rs746711547 Chr.18: 79919107-79919107 on Build GRCh38
rs574218585 Chr.18: 79919014-79919014 on Build GRCh38
rs969233813 Chr.18: 79918987-79918987 on Build GRCh38
rs776569036 Chr.18: 79919352-79919352 on Build GRCh38
rs575102840 Chr.18: 79919392-79919392 on Build GRCh38
rs751045128 Chr.18: 79919135-79919137 on Build GRCh38
rs747182588 Chr.18: 79919399-79919399 on Build GRCh38
rs575064804 Chr.18: 79919136-79919136 on Build GRCh38
rs759563401 Chr.18: 79919408-79919408 on Build GRCh38
rs755636023 Chr.18: 79919177-79919177 on Build GRCh38
rs758127377 Chr.18: 79919360-79919360 on Build GRCh38
rs367755489 Chr.18: 79919291-79919291 on Build GRCh38
rs150969347 Chr.18: 79919192-79919192 on Build GRCh38
rs150340109 Chr.18: 79919322-79919322 on Build GRCh38
rs773053503 Chr.18: 79919348-79919348 on Build GRCh38
rs780887814 Chr.18: 79919354-79919354 on Build GRCh38
rs753327216 Chr.18: 79919272-79919272 on Build GRCh38
rs570486466 Chr.18: 79919354-79919354 on Build GRCh38
rs376461831 Chr.18: 79919167-79919167 on Build GRCh38
rs761893159 Chr.18: 79919305-79919305 on Build GRCh38
rs757953021 Chr.18: 79919328-79919328 on Build GRCh38
rs772165178 Chr.18: 79919347-79919347 on Build GRCh38
rs34425104 Chr.18: 79919327-79919327 on Build GRCh38
rs183043398 Chr.18: 79919072-79919072 on Build GRCh38
rs373259554 Chr.18: 79919345-79919345 on Build GRCh38
rs201846863 Chr.18: 79919190-79919190 on Build GRCh38
rs556201315 Chr.18: 79919370-79919370 on Build GRCh38
rs758653160 Chr.18: 79919162-79919162 on Build GRCh38
rs3744879 Chr.18: 79919006-79919006 on Build GRCh38
rs1041649206 Chr.18: 79919093-79919093 on Build GRCh38
rs114428753 Chr.18: 79919249-79919249 on Build GRCh38
rs563166504 Chr.18: 79919109-79919109 on Build GRCh38
rs542569199 Chr.18: 79919168-79919168 on Build GRCh38
rs764019325 Chr.18: 79919163-79919163 on Build GRCh38
rs780520150 Chr.18: 79919372-79919372 on Build GRCh38
rs369096451 Chr.18: 79919404-79919404 on Build GRCh38
rs969012060 Chr.18: 79919128-79919128 on Build GRCh38
rs781368119 Chr.18: 79919282-79919282 on Build GRCh38
rs750192280 Chr.18: 79919310-79919310 on Build GRCh38
rs528372880 Chr.18: 79919218-79919218 on Build GRCh38
rs754685293 Chr.18: 79919273-79919273 on Build GRCh38
rs780710194 Chr.18: 79919334-79919334 on Build GRCh38
rs771949291 Chr.18: 79919209-79919209 on Build GRCh38
rs894643594 Chr.18: 79918986-79918986 on Build GRCh38
rs763315416 Chr.18: 79919227-79919227 on Build GRCh38
rs773500920 Chr.18: 79919196-79919196 on Build GRCh38
rs138426075 Chr.18: 79919396-79919396 on Build GRCh38
rs776374864 Chr.18: 79919407-79919407 on Build GRCh38
rs767160813 Chr.18: 79919248-79919248 on Build GRCh38
rs114615170 Chr.18: 79919361-79919361 on Build GRCh38
rs185687241 Chr.18: 79919027-79919027 on Build GRCh38
rs745726958 Chr.18: 79919176-79919176 on Build GRCh38
rs756006525 Chr.18: 79919254-79919254 on Build GRCh38
rs755410098 Chr.18: 79919378-79919378 on Build GRCh38
rs747696074 Chr.18: 79919276-79919276 on Build GRCh38
rs779490691 Chr.18: 79919188-79919188 on Build GRCh38
rs766099297 Chr.18: 79919351-79919351 on Build GRCh38
rs774648165 Chr.18: 79919299-79919299 on Build GRCh38
rs149523150 Chr.18: 79919339-79919339 on Build GRCh38
rs768269773 Chr.18: 79919195-79919195 on Build GRCh38
rs114579767 Chr.18: 79919290-79919290 on Build GRCh38
rs765072600 Chr.18: 79919414-79919414 on Build GRCh38
rs766293899 Chr.18: 79919318-79919318 on Build GRCh38
rs375190137 Chr.18: 79919356-79919356 on Build GRCh38
rs564712147 Chr.18: 79919240-79919240 on Build GRCh38
rs763119703 Chr.18: 79919297-79919297 on Build GRCh38
rs751189042 Chr.18: 79919366-79919366 on Build GRCh38
rs191651723 Chr.18: 79919326-79919326 on Build GRCh38
rs556606952 Chr.18: 79919096-79919096 on Build GRCh38
rs760461464 Chr.18: 79919349-79919349 on Build GRCh38
rs140775989 Chr.18: 79919228-79919228 on Build GRCh38
rs748167009 Chr.18: 79919395-79919395 on Build GRCh38
rs775679883 Chr.18: 79919224-79919224 on Build GRCh38
rs376654152 Chr.18: 79919209-79919209 on Build GRCh38
rs201323361 Chr.18: 79919376-79919376 on Build GRCh38
rs866610393 Chr.18: 79919359-79919359 on Build GRCh38
rs369594354 Chr.18: 79919344-79919344 on Build GRCh38
rs61735018 Chr.18: 79919274-79919274 on Build GRCh38
rs942875519 Chr.18: 79919078-79919078 on Build GRCh38
rs751376316 Chr.18: 79919237-79919237 on Build GRCh38
rs759347267 Chr.18: 79919353-79919353 on Build GRCh38
rs747389517 Chr.18: 79919199-79919199 on Build GRCh38

View Full Product Details