Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 2076627-2077124 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MOB3A
Entrez Gene ID
126308
Gene Name
MOB kinase activator 3A
Gene Aliases
MOB1C, MOBKL2A, moblak
UniGene
Hs.744938
Gene Chromosome Location
Chr.19: 2071036-2096502 on Build GRCh38
Transcripts
NM_130807.2 XM_011527683.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3531256 c.444C>T p.F148F Chr.19: 2076991-2076991 on Build GRCh38
993562 c.579C>T p.F193F Chr.19: 2076856-2076856 on Build GRCh38
1680485 c.438G>C p.K146N Chr.19: 2076997-2076997 on Build GRCh38
3937960 c.452C>T p.T151M Chr.19: 2076983-2076983 on Build GRCh38
6714077 c.565C>T p.H189Y Chr.19: 2076870-2076870 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs953911936 Chr.19: 2076759-2076759 on Build GRCh38
rs994718390 Chr.19: 2077016-2077016 on Build GRCh38
rs772437903 Chr.19: 2076951-2076951 on Build GRCh38
rs770222312 Chr.19: 2076808-2076808 on Build GRCh38
rs770536200 Chr.19: 2077035-2077035 on Build GRCh38
rs772773904 Chr.19: 2076910-2076910 on Build GRCh38
rs576174515 Chr.19: 2076998-2076998 on Build GRCh38
rs778043036 Chr.19: 2076939-2076939 on Build GRCh38
rs989755124 Chr.19: 2076776-2076776 on Build GRCh38
rs879546370 Chr.19: 2076922-2076922 on Build GRCh38
rs776272693 Chr.19: 2076876-2076878 on Build GRCh38
rs141042584 Chr.19: 2076975-2076975 on Build GRCh38
rs906652385 Chr.19: 2076674-2076674 on Build GRCh38
rs756166447 Chr.19: 2076982-2076982 on Build GRCh38
rs747581677 Chr.19: 2076955-2076955 on Build GRCh38
rs976863022 Chr.19: 2076856-2076856 on Build GRCh38
rs370786420 Chr.19: 2076918-2076918 on Build GRCh38
rs537075452 Chr.19: 2076662-2076662 on Build GRCh38
rs774070272 Chr.19: 2076966-2076966 on Build GRCh38
rs748034778 Chr.19: 2076852-2076852 on Build GRCh38
rs752351072 Chr.19: 2076951-2076960 on Build GRCh38
rs759307055 Chr.19: 2077040-2077040 on Build GRCh38
rs1014788931 Chr.19: 2076717-2076720 on Build GRCh38
rs748547038 Chr.19: 2077021-2077021 on Build GRCh38
rs371597769 Chr.19: 2076855-2076855 on Build GRCh38
rs751133450 Chr.19: 2076914-2076914 on Build GRCh38
rs201748512 Chr.19: 2076789-2076789 on Build GRCh38
rs775980541 Chr.19: 2076898-2076898 on Build GRCh38
rs749543358 Chr.19: 2076868-2076868 on Build GRCh38
rs764661801 Chr.19: 2076883-2076884 on Build GRCh38
rs757233836 Chr.19: 2076945-2076945 on Build GRCh38
rs375974312 Chr.19: 2076988-2076988 on Build GRCh38
rs756119804 Chr.19: 2076926-2076926 on Build GRCh38
rs773731864 Chr.19: 2077038-2077038 on Build GRCh38
rs576809612 Chr.19: 2076946-2076946 on Build GRCh38
rs781472894 Chr.19: 2076769-2076769 on Build GRCh38
rs777988945 Chr.19: 2076864-2076864 on Build GRCh38
rs534131418 Chr.19: 2076917-2076917 on Build GRCh38
rs774625797 Chr.19: 2076884-2076884 on Build GRCh38
rs151181290 Chr.19: 2076962-2076962 on Build GRCh38
rs770604419 Chr.19: 2076978-2076978 on Build GRCh38
rs575702897 Chr.19: 2077010-2077010 on Build GRCh38
rs768500243 Chr.19: 2076742-2076742 on Build GRCh38
rs776954278 Chr.19: 2076958-2076958 on Build GRCh38
rs762563833 Chr.19: 2076911-2076911 on Build GRCh38
rs144377379 Chr.19: 2076940-2076940 on Build GRCh38
rs998560047 Chr.19: 2076977-2076977 on Build GRCh38
rs761342670 Chr.19: 2076807-2076807 on Build GRCh38
rs1013465339 Chr.19: 2076649-2076649 on Build GRCh38
rs753825901 Chr.19: 2077059-2077059 on Build GRCh38
rs750088021 Chr.19: 2076819-2076819 on Build GRCh38
rs771832617 Chr.19: 2077041-2077041 on Build GRCh38
rs753150560 Chr.19: 2076805-2076805 on Build GRCh38
rs748261328 Chr.19: 2076773-2076773 on Build GRCh38
rs773889547 Chr.19: 2076983-2076983 on Build GRCh38
rs779754513 Chr.19: 2076823-2076823 on Build GRCh38
rs1003366730 Chr.19: 2076712-2076712 on Build GRCh38
rs763182036 Chr.19: 2076778-2076778 on Build GRCh38
rs764782777 Chr.19: 2076906-2076906 on Build GRCh38
rs758053844 Chr.19: 2076820-2076820 on Build GRCh38
rs751349284 Chr.19: 2076834-2076834 on Build GRCh38
rs144786970 Chr.19: 2077019-2077019 on Build GRCh38
rs765742498 Chr.19: 2076964-2076964 on Build GRCh38
rs557526411 Chr.19: 2076933-2076933 on Build GRCh38
rs948081891 Chr.19: 2076661-2076661 on Build GRCh38
rs897718585 Chr.19: 2076730-2076730 on Build GRCh38
rs536451789 Chr.19: 2076760-2076760 on Build GRCh38
rs1004283234 Chr.19: 2076694-2076694 on Build GRCh38
rs781472413 Chr.19: 2076718-2076718 on Build GRCh38
rs747397350 Chr.19: 2077013-2077013 on Build GRCh38
rs568928398 Chr.19: 2076692-2076692 on Build GRCh38
rs775819086 Chr.19: 2076952-2076952 on Build GRCh38
rs965737297 Chr.19: 2077071-2077071 on Build GRCh38
rs746010262 Chr.19: 2076949-2076949 on Build GRCh38
rs771149865 Chr.19: 2076882-2076882 on Build GRCh38
rs759031172 Chr.19: 2076882-2076883 on Build GRCh38
rs758613979 Chr.19: 2077007-2077007 on Build GRCh38
rs763726785 Chr.19: 2077049-2077049 on Build GRCh38
rs1015633683 Chr.19: 2076696-2076696 on Build GRCh38
rs373826686 Chr.19: 2076889-2076889 on Build GRCh38
rs760573173 Chr.19: 2077046-2077046 on Build GRCh38
rs752742337 Chr.19: 2076924-2076924 on Build GRCh38
rs1028620121 Chr.19: 2076735-2076735 on Build GRCh38
rs1027146952 Chr.19: 2076671-2076671 on Build GRCh38
rs376132264 Chr.19: 2076788-2076788 on Build GRCh38
rs769955422 Chr.19: 2076777-2076777 on Build GRCh38
rs764660429 Chr.19: 2076812-2076812 on Build GRCh38
rs111825801 Chr.19: 2077097-2077097 on Build GRCh38
rs894643118 Chr.19: 2076648-2076648 on Build GRCh38
rs939436083 Chr.19: 2076973-2076973 on Build GRCh38
rs775231308 Chr.19: 2077043-2077043 on Build GRCh38
rs759168837 Chr.19: 2077006-2077006 on Build GRCh38
rs965345201 Chr.19: 2076716-2076716 on Build GRCh38
rs746262774 Chr.19: 2076888-2076888 on Build GRCh38
rs781311202 Chr.19: 2076848-2076848 on Build GRCh38
rs761111804 Chr.19: 2076900-2076900 on Build GRCh38
rs142102227 Chr.19: 2076912-2076912 on Build GRCh38
rs953159655 Chr.19: 2076770-2076770 on Build GRCh38
rs768608116 Chr.19: 2076762-2076762 on Build GRCh38
rs771261352 Chr.19: 2076787-2076787 on Build GRCh38
rs888570032 Chr.19: 2076691-2076691 on Build GRCh38
rs147932341 Chr.19: 2077095-2077095 on Build GRCh38
rs754681414 Chr.19: 2076835-2076835 on Build GRCh38
rs767985564 Chr.19: 2076790-2076790 on Build GRCh38
rs543937620 Chr.19: 2076971-2076971 on Build GRCh38

View Full Product Details