Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 2291376-2291867 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINGO3
Entrez Gene ID
645191
Gene Name
leucine rich repeat and Ig domain containing 3
Gene Aliases
LERN2, LRRN6B
UniGene
Hs.371587
Gene Chromosome Location
Chr.19: 2289775-2308157 on Build GRCh38
Transcripts
NM_001101391.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5533835 c.362T>A p.V121D Chr.19: 2291415-2291415 on Build GRCh38
6701446 c.370C>T p.R124C Chr.19: 2291407-2291407 on Build GRCh38
4698108 c.358G>A p.G120R Chr.19: 2291419-2291419 on Build GRCh38
2748409 c.272C>T p.A91V Chr.19: 2291505-2291505 on Build GRCh38
6046462 c.273G>A p.A91A Chr.19: 2291504-2291504 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374349778 Chr.19: 2291426-2291426 on Build GRCh38
rs755322420 Chr.19: 2291488-2291488 on Build GRCh38
rs763049878 Chr.19: 2291633-2291633 on Build GRCh38
rs769981219 Chr.19: 2291618-2291618 on Build GRCh38
rs1052522363 Chr.19: 2291753-2291753 on Build GRCh38
rs368475120 Chr.19: 2291555-2291555 on Build GRCh38
rs199967398 Chr.19: 2291407-2291407 on Build GRCh38
rs942732213 Chr.19: 2291537-2291537 on Build GRCh38
rs888671902 Chr.19: 2291536-2291536 on Build GRCh38
rs927752205 Chr.19: 2291491-2291491 on Build GRCh38
rs747976349 Chr.19: 2291598-2291598 on Build GRCh38
rs768443305 Chr.19: 2291513-2291513 on Build GRCh38
rs780451895 Chr.19: 2291400-2291400 on Build GRCh38
rs745406491 Chr.19: 2291501-2291501 on Build GRCh38
rs765510095 Chr.19: 2291524-2291524 on Build GRCh38
rs774866588 Chr.19: 2291715-2291715 on Build GRCh38
rs768208968 Chr.19: 2291796-2291796 on Build GRCh38
rs529083265 Chr.19: 2291578-2291578 on Build GRCh38
rs984208851 Chr.19: 2291435-2291435 on Build GRCh38
rs758941206 Chr.19: 2291476-2291476 on Build GRCh38
rs942950279 Chr.19: 2291717-2291717 on Build GRCh38
rs750045332 Chr.19: 2291838-2291838 on Build GRCh38
rs760434360 Chr.19: 2291449-2291449 on Build GRCh38
rs761812750 Chr.19: 2291467-2291467 on Build GRCh38
rs933953673 Chr.19: 2291742-2291742 on Build GRCh38
rs1041063267 Chr.19: 2291716-2291717 on Build GRCh38
rs753085846 Chr.19: 2291811-2291811 on Build GRCh38
rs752039603 Chr.19: 2291543-2291543 on Build GRCh38
rs762008973 Chr.19: 2291519-2291519 on Build GRCh38
rs1002113483 Chr.19: 2291801-2291801 on Build GRCh38
rs764324261 Chr.19: 2291461-2291461 on Build GRCh38
rs533205768 Chr.19: 2291798-2291798 on Build GRCh38
rs1034629773 Chr.19: 2291660-2291660 on Build GRCh38
rs984721002 Chr.19: 2291847-2291847 on Build GRCh38
rs376005631 Chr.19: 2291525-2291525 on Build GRCh38
rs201895707 Chr.19: 2291433-2291433 on Build GRCh38
rs559382174 Chr.19: 2291726-2291726 on Build GRCh38
rs971807173 Chr.19: 2291695-2291695 on Build GRCh38
rs753173891 Chr.19: 2291554-2291554 on Build GRCh38
rs748370587 Chr.19: 2291827-2291827 on Build GRCh38
rs547609531 Chr.19: 2291716-2291716 on Build GRCh38
rs1011188941 Chr.19: 2291762-2291762 on Build GRCh38
rs777530966 Chr.19: 2291489-2291489 on Build GRCh38
rs1039758475 Chr.19: 2291720-2291720 on Build GRCh38
rs775141893 Chr.19: 2291461-2291462 on Build GRCh38
rs918967520 Chr.19: 2291696-2291696 on Build GRCh38
rs989815570 Chr.19: 2291458-2291458 on Build GRCh38
rs760111112 Chr.19: 2291768-2291768 on Build GRCh38
rs1002315599 Chr.19: 2291646-2291646 on Build GRCh38
rs748956241 Chr.19: 2291497-2291497 on Build GRCh38
rs370489824 Chr.19: 2291423-2291423 on Build GRCh38
rs866177237 Chr.19: 2291544-2291544 on Build GRCh38
rs564458944 Chr.19: 2291520-2291520 on Build GRCh38
rs757865660 Chr.19: 2291843-2291843 on Build GRCh38
rs372562029 Chr.19: 2291574-2291574 on Build GRCh38
rs1022186992 Chr.19: 2291469-2291469 on Build GRCh38
rs775309301 Chr.19: 2291511-2291511 on Build GRCh38
rs745329479 Chr.19: 2291558-2291558 on Build GRCh38
rs562191489 Chr.19: 2291573-2291573 on Build GRCh38
rs543901400 Chr.19: 2291561-2291561 on Build GRCh38
rs768010341 Chr.19: 2291553-2291553 on Build GRCh38
rs960471979 Chr.19: 2291667-2291667 on Build GRCh38
rs776775013 Chr.19: 2291518-2291518 on Build GRCh38
rs759534500 Chr.19: 2291429-2291429 on Build GRCh38
rs892515795 Chr.19: 2291757-2291757 on Build GRCh38
rs901253071 Chr.19: 2291396-2291396 on Build GRCh38
rs766376561 Chr.19: 2291657-2291657 on Build GRCh38
rs952382769 Chr.19: 2291408-2291408 on Build GRCh38
rs769458369 Chr.19: 2291406-2291406 on Build GRCh38
rs773331053 Chr.19: 2291626-2291626 on Build GRCh38
rs1022639920 Chr.19: 2291606-2291606 on Build GRCh38
rs1049124319 Chr.19: 2291536-2291537 on Build GRCh38
rs200778841 Chr.19: 2291500-2291500 on Build GRCh38
rs747400509 Chr.19: 2291404-2291404 on Build GRCh38
rs771957087 Chr.19: 2291472-2291472 on Build GRCh38
rs1034877227 Chr.19: 2291813-2291813 on Build GRCh38
rs756565329 Chr.19: 2291829-2291829 on Build GRCh38
rs1043562833 Chr.19: 2291765-2291766 on Build GRCh38
rs372031496 Chr.19: 2291534-2291534 on Build GRCh38
rs766862011 Chr.19: 2291541-2291541 on Build GRCh38
rs951828613 Chr.19: 2291844-2291844 on Build GRCh38
rs930138234 Chr.19: 2291504-2291504 on Build GRCh38
rs969323843 Chr.19: 2291625-2291625 on Build GRCh38
rs780465898 Chr.19: 2291483-2291483 on Build GRCh38
rs1026055446 Chr.19: 2291841-2291841 on Build GRCh38
rs765311639 Chr.19: 2291470-2291470 on Build GRCh38
rs751932548 Chr.19: 2291486-2291486 on Build GRCh38
rs764940971 Chr.19: 2291834-2291834 on Build GRCh38
rs778754172 Chr.19: 2291557-2291557 on Build GRCh38
rs776405786 Chr.19: 2291596-2291596 on Build GRCh38
rs998059803 Chr.19: 2291420-2291420 on Build GRCh38
rs867533018 Chr.19: 2291527-2291527 on Build GRCh38
rs748760022 Chr.19: 2291418-2291418 on Build GRCh38
rs762991714 Chr.19: 2291526-2291526 on Build GRCh38
rs760476245 Chr.19: 2291512-2291512 on Build GRCh38
rs755306858 Chr.19: 2291552-2291552 on Build GRCh38
rs201142184 Chr.19: 2291502-2291502 on Build GRCh38

View Full Product Details