Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3225401-3225895 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELF5
Entrez Gene ID
60680
Gene Name
CUGBP, Elav-like family member 5
Gene Aliases
BRUNOL-5, BRUNOL5, CELF-5
UniGene
Hs.655747
Gene Chromosome Location
Chr.19: 3224676-3297076 on Build GRCh38
Transcripts
NM_001172673.1 NM_021938.3 NR_033342.1 XM_006722832.1 XM_006722833.1 XM_006722837.3

SNP Detail

SNP ID Chromosome Location
rs540347625 Chr.19: 3225699-3225699 on Build GRCh38
rs73517111 Chr.19: 3225697-3225697 on Build GRCh38
rs894117251 Chr.19: 3225707-3225707 on Build GRCh38
rs986809756 Chr.19: 3225838-3225838 on Build GRCh38
rs941475470 Chr.19: 3225650-3225650 on Build GRCh38
rs541069021 Chr.19: 3225784-3225784 on Build GRCh38
rs1045266742 Chr.19: 3225773-3225773 on Build GRCh38
rs982961344 Chr.19: 3225776-3225776 on Build GRCh38
rs561227174 Chr.19: 3225786-3225786 on Build GRCh38
rs575487155 Chr.19: 3225657-3225657 on Build GRCh38
rs1036630452 Chr.19: 3225479-3225479 on Build GRCh38
rs564948169 Chr.19: 3225728-3225728 on Build GRCh38
rs312056 Chr.19: 3225683-3225683 on Build GRCh38
rs936732048 Chr.19: 3225782-3225782 on Build GRCh38
rs925808749 Chr.19: 3225866-3225866 on Build GRCh38
rs1017557685 Chr.19: 3225791-3225791 on Build GRCh38
rs892452240 Chr.19: 3225788-3225788 on Build GRCh38
rs949533253 Chr.19: 3225507-3225507 on Build GRCh38
rs983357135 Chr.19: 3225598-3225598 on Build GRCh38
rs1052066393 Chr.19: 3225663-3225663 on Build GRCh38
rs1009802467 Chr.19: 3225669-3225669 on Build GRCh38
rs1024472204 Chr.19: 3225742-3225742 on Build GRCh38
rs945868924 Chr.19: 3225676-3225676 on Build GRCh38
rs993070287 Chr.19: 3225532-3225532 on Build GRCh38
rs917781665 Chr.19: 3225706-3225706 on Build GRCh38
rs984423929 Chr.19: 3225438-3225438 on Build GRCh38
rs945996002 Chr.19: 3225856-3225856 on Build GRCh38
rs975435812 Chr.19: 3225651-3225651 on Build GRCh38
rs1033964136 Chr.19: 3225704-3225704 on Build GRCh38
rs933942341 Chr.19: 3225833-3225833 on Build GRCh38
rs1024526124 Chr.19: 3225535-3225535 on Build GRCh38
rs1013893550 Chr.19: 3225721-3225721 on Build GRCh38
rs372139427 Chr.19: 3225596-3225596 on Build GRCh38
rs984474126 Chr.19: 3225692-3225692 on Build GRCh38
rs1010275391 Chr.19: 3225790-3225790 on Build GRCh38
rs890383169 Chr.19: 3225673-3225673 on Build GRCh38
rs911806285 Chr.19: 3225854-3225854 on Build GRCh38
rs942576633 Chr.19: 3225454-3225454 on Build GRCh38
rs542504943 Chr.19: 3225670-3225670 on Build GRCh38
rs1022059514 Chr.19: 3225772-3225772 on Build GRCh38
rs921288303 Chr.19: 3225656-3225656 on Build GRCh38
rs917746473 Chr.19: 3225480-3225480 on Build GRCh38
rs951724928 Chr.19: 3225760-3225760 on Build GRCh38
rs997843841 Chr.19: 3225589-3225589 on Build GRCh38
rs965573499 Chr.19: 3225671-3225671 on Build GRCh38
rs949250650 Chr.19: 3225722-3225722 on Build GRCh38
rs553974820 Chr.19: 3225575-3225575 on Build GRCh38
rs1011049761 Chr.19: 3225714-3225714 on Build GRCh38
rs909934452 Chr.19: 3225646-3225646 on Build GRCh38
rs768952703 Chr.19: 3225826-3225826 on Build GRCh38
rs940564866 Chr.19: 3225476-3225476 on Build GRCh38
rs951555262 Chr.19: 3225582-3225582 on Build GRCh38
rs969280816 Chr.19: 3225778-3225778 on Build GRCh38
rs962807250 Chr.19: 3225810-3225810 on Build GRCh38
rs1005962634 Chr.19: 3225873-3225873 on Build GRCh38
rs1039623446 Chr.19: 3225475-3225475 on Build GRCh38
rs59268131 Chr.19: 3225816-3225816 on Build GRCh38
rs931640783 Chr.19: 3225425-3225425 on Build GRCh38
rs909142504 Chr.19: 3225465-3225465 on Build GRCh38
rs1041535587 Chr.19: 3225861-3225861 on Build GRCh38

View Full Product Details