Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 4028004-4028522 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIAS4
Entrez Gene ID
51588
Gene Name
protein inhibitor of activated STAT 4
Gene Aliases
PIAS-gamma, PIASY, Piasg, ZMIZ6
UniGene
Hs.105779
Gene Chromosome Location
Chr.19: 4007598-4039386 on Build GRCh38
Transcripts
NM_015897.3 XM_011528060.2 XM_017026868.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6334885 c.582-10G>T p.? Chr.19: 4028500-4028500 on Build GRCh38
6214719 c.565G>A p.V189M Chr.19: 4028171-4028171 on Build GRCh38
1290946 c.573C>T p.V191V Chr.19: 4028179-4028179 on Build GRCh38
6524423 c.554G>A p.G185E Chr.19: 4028160-4028160 on Build GRCh38
996418 c.552C>A p.P184P Chr.19: 4028158-4028158 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs535221827 Chr.19: 4028249-4028249 on Build GRCh38
rs907357177 Chr.19: 4028451-4028451 on Build GRCh38
rs754263952 Chr.19: 4028473-4028473 on Build GRCh38
rs928676584 Chr.19: 4028246-4028246 on Build GRCh38
rs4520954 Chr.19: 4028069-4028069 on Build GRCh38
rs767450165 Chr.19: 4028482-4028482 on Build GRCh38
rs762074960 Chr.19: 4028200-4028200 on Build GRCh38
rs767505230 Chr.19: 4028208-4028208 on Build GRCh38
rs773581976 Chr.19: 4028468-4028468 on Build GRCh38
rs972386281 Chr.19: 4028332-4028332 on Build GRCh38
rs918380535 Chr.19: 4028339-4028339 on Build GRCh38
rs766303153 Chr.19: 4028104-4028104 on Build GRCh38
rs751397524 Chr.19: 4028206-4028206 on Build GRCh38
rs879901172 Chr.19: 4028376-4028376 on Build GRCh38
rs1042350063 Chr.19: 4028379-4028379 on Build GRCh38
rs912348183 Chr.19: 4028404-4028404 on Build GRCh38
rs1054355565 Chr.19: 4028443-4028443 on Build GRCh38
rs549375430 Chr.19: 4028091-4028091 on Build GRCh38
rs759338110 Chr.19: 4028471-4028471 on Build GRCh38
rs543625038 Chr.19: 4028422-4028422 on Build GRCh38
rs9941481 Chr.19: 4028414-4028414 on Build GRCh38
rs759462573 Chr.19: 4028207-4028207 on Build GRCh38
rs985300269 Chr.19: 4028390-4028390 on Build GRCh38
rs1054741544 Chr.19: 4028371-4028371 on Build GRCh38
rs949900529 Chr.19: 4028076-4028076 on Build GRCh38
rs919787538 Chr.19: 4028039-4028039 on Build GRCh38
rs896885387 Chr.19: 4028478-4028478 on Build GRCh38
rs937293302 Chr.19: 4028369-4028369 on Build GRCh38
rs768162611 Chr.19: 4028222-4028222 on Build GRCh38
rs924597280 Chr.19: 4028102-4028102 on Build GRCh38
rs771151053 Chr.19: 4028156-4028156 on Build GRCh38
rs779225437 Chr.19: 4028159-4028159 on Build GRCh38
rs2289864 Chr.19: 4028077-4028077 on Build GRCh38
rs889712484 Chr.19: 4028483-4028483 on Build GRCh38
rs976558516 Chr.19: 4028416-4028416 on Build GRCh38
rs902633299 Chr.19: 4028388-4028388 on Build GRCh38
rs961019466 Chr.19: 4028375-4028375 on Build GRCh38
rs199857186 Chr.19: 4028234-4028234 on Build GRCh38
rs145626990 Chr.19: 4028179-4028179 on Build GRCh38
rs906953479 Chr.19: 4028297-4028297 on Build GRCh38
rs889871686 Chr.19: 4028486-4028486 on Build GRCh38
rs981575667 Chr.19: 4028450-4028450 on Build GRCh38
rs763976804 Chr.19: 4028497-4028497 on Build GRCh38
rs375572427 Chr.19: 4028205-4028205 on Build GRCh38
rs201647511 Chr.19: 4028170-4028170 on Build GRCh38
rs1007148325 Chr.19: 4028405-4028405 on Build GRCh38
rs986090364 Chr.19: 4028262-4028262 on Build GRCh38
rs563785400 Chr.19: 4028075-4028075 on Build GRCh38
rs571958540 Chr.19: 4028349-4028349 on Build GRCh38
rs927092456 Chr.19: 4028356-4028356 on Build GRCh38
rs957590538 Chr.19: 4028228-4028228 on Build GRCh38
rs113938534 Chr.19: 4028355-4028355 on Build GRCh38
rs747577365 Chr.19: 4028237-4028237 on Build GRCh38
rs999391280 Chr.19: 4028209-4028209 on Build GRCh38
rs200774683 Chr.19: 4028167-4028167 on Build GRCh38
rs748244572 Chr.19: 4028461-4028461 on Build GRCh38
rs1044607682 Chr.19: 4028447-4028447 on Build GRCh38
rs557168552 Chr.19: 4028204-4028204 on Build GRCh38
rs879181025 Chr.19: 4028026-4028026 on Build GRCh38
rs529401627 Chr.19: 4028438-4028438 on Build GRCh38
rs1030230140 Chr.19: 4028242-4028242 on Build GRCh38
rs777223960 Chr.19: 4028142-4028142 on Build GRCh38
rs953841933 Chr.19: 4028068-4028068 on Build GRCh38
rs958749823 Chr.19: 4028455-4028455 on Build GRCh38
rs749000055 Chr.19: 4028154-4028154 on Build GRCh38
rs937243256 Chr.19: 4028094-4028094 on Build GRCh38
rs1026922562 Chr.19: 4028378-4028378 on Build GRCh38
rs556711233 Chr.19: 4028060-4028060 on Build GRCh38
rs199596840 Chr.19: 4028125-4028125 on Build GRCh38
rs914832783 Chr.19: 4028444-4028444 on Build GRCh38
rs532989068 Chr.19: 4028059-4028059 on Build GRCh38
rs755663449 Chr.19: 4028127-4028127 on Build GRCh38
rs775241310 Chr.19: 4028494-4028494 on Build GRCh38
rs760099200 Chr.19: 4028301-4028301 on Build GRCh38
rs760636057 Chr.19: 4028496-4028496 on Build GRCh38
rs770129941 Chr.19: 4028463-4028463 on Build GRCh38
rs944756876 Chr.19: 4028081-4028081 on Build GRCh38
rs751452387 Chr.19: 4028111-4028111 on Build GRCh38
rs557785309 Chr.19: 4028372-4028372 on Build GRCh38
rs866963871 Chr.19: 4028377-4028377 on Build GRCh38
rs754936743 Chr.19: 4028112-4028112 on Build GRCh38
rs962956241 Chr.19: 4028418-4028418 on Build GRCh38
rs948933987 Chr.19: 4028446-4028446 on Build GRCh38
rs990349121 Chr.19: 4028229-4028229 on Build GRCh38
rs953916198 Chr.19: 4028389-4028389 on Build GRCh38
rs995185914 Chr.19: 4028481-4028481 on Build GRCh38
rs1015294223 Chr.19: 4028365-4028365 on Build GRCh38
rs1044924188 Chr.19: 4028293-4028293 on Build GRCh38
rs571784826 Chr.19: 4028171-4028171 on Build GRCh38
rs755539511 Chr.19: 4028218-4028218 on Build GRCh38
rs370495454 Chr.19: 4028236-4028236 on Build GRCh38
rs753335912 Chr.19: 4028232-4028232 on Build GRCh38
rs746867423 Chr.19: 4028118-4028118 on Build GRCh38
rs998387397 Chr.19: 4028238-4028238 on Build GRCh38
rs776744081 Chr.19: 4028460-4028460 on Build GRCh38
rs746214299 Chr.19: 4028235-4028235 on Build GRCh38
rs776029917 Chr.19: 4028173-4028173 on Build GRCh38
rs72976957 Chr.19: 4028292-4028292 on Build GRCh38
rs769094916 Chr.19: 4028459-4028459 on Build GRCh38
rs12460406 Chr.19: 4028342-4028342 on Build GRCh38
rs566124258 Chr.19: 4028095-4028095 on Build GRCh38
rs905806156 Chr.19: 4028092-4028092 on Build GRCh38
rs981583033 Chr.19: 4028241-4028241 on Build GRCh38
rs374877228 Chr.19: 4028472-4028472 on Build GRCh38
rs563500568 Chr.19: 4028437-4028437 on Build GRCh38
rs202213110 Chr.19: 4028491-4028491 on Build GRCh38
rs954332264 Chr.19: 4028243-4028243 on Build GRCh38
rs776671450 Chr.19: 4028182-4028182 on Build GRCh38
rs1013051237 Chr.19: 4028259-4028259 on Build GRCh38

View Full Product Details