Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 4327209-4327714 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STAP2
Entrez Gene ID
55620
Gene Name
signal transducing adaptor family member 2
Gene Aliases
BKS
UniGene
Hs.194385
Gene Chromosome Location
Chr.19: 4324043-4338877 on Build GRCh38
Transcripts
NM_001013841.1 NM_017720.2 XM_011528123.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5855995 c.660+1G>A p.? Chr.19: 4327315-4327315 on Build GRCh38
6852676 c.643A>T p.I215F Chr.19: 4327333-4327333 on Build GRCh38
7222507 c.592A>G p.T198A Chr.19: 4327384-4327384 on Build GRCh38
7177011 c.591-1G>A p.? Chr.19: 4327386-4327386 on Build GRCh38
4856367 c.640G>A p.V214M Chr.19: 4327336-4327336 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766658608 Chr.19: 4327296-4327296 on Build GRCh38
rs34389341 Chr.19: 4327317-4327318 on Build GRCh38
rs542998520 Chr.19: 4327450-4327450 on Build GRCh38
rs928001038 Chr.19: 4327444-4327444 on Build GRCh38
rs373701857 Chr.19: 4327346-4327346 on Build GRCh38
rs1008858384 Chr.19: 4327563-4327563 on Build GRCh38
rs888146258 Chr.19: 4327691-4327691 on Build GRCh38
rs201322610 Chr.19: 4327384-4327384 on Build GRCh38
rs996537354 Chr.19: 4327261-4327261 on Build GRCh38
rs141244385 Chr.19: 4327334-4327334 on Build GRCh38
rs761279669 Chr.19: 4327386-4327386 on Build GRCh38
rs957169836 Chr.19: 4327610-4327610 on Build GRCh38
rs781039850 Chr.19: 4327343-4327343 on Build GRCh38
rs549890185 Chr.19: 4327575-4327575 on Build GRCh38
rs367842237 Chr.19: 4327391-4327391 on Build GRCh38
rs577690188 Chr.19: 4327383-4327383 on Build GRCh38
rs370264236 Chr.19: 4327328-4327328 on Build GRCh38
rs748861370 Chr.19: 4327421-4327421 on Build GRCh38
rs888934577 Chr.19: 4327542-4327542 on Build GRCh38
rs771947497 Chr.19: 4327272-4327272 on Build GRCh38
rs759732198 Chr.19: 4327303-4327304 on Build GRCh38
rs559975671 Chr.19: 4327452-4327452 on Build GRCh38
rs7343166 Chr.19: 4327239-4327239 on Build GRCh38
rs573688112 Chr.19: 4327424-4327424 on Build GRCh38
rs752792664 Chr.19: 4327315-4327315 on Build GRCh38
rs749823271 Chr.19: 4327574-4327574 on Build GRCh38
rs59388432 Chr.19: 4327422-4327422 on Build GRCh38
rs998495439 Chr.19: 4327571-4327571 on Build GRCh38
rs759936336 Chr.19: 4327381-4327381 on Build GRCh38
rs768682330 Chr.19: 4327340-4327340 on Build GRCh38
rs746861123 Chr.19: 4327336-4327336 on Build GRCh38
rs777900233 Chr.19: 4327237-4327237 on Build GRCh38
rs769146053 Chr.19: 4327353-4327353 on Build GRCh38
rs777131821 Chr.19: 4327420-4327420 on Build GRCh38
rs762621476 Chr.19: 4327295-4327295 on Build GRCh38
rs757558185 Chr.19: 4327322-4327322 on Build GRCh38
rs764013581 Chr.19: 4327387-4327387 on Build GRCh38
rs151322217 Chr.19: 4327361-4327361 on Build GRCh38
rs376996833 Chr.19: 4327265-4327265 on Build GRCh38
rs774712029 Chr.19: 4327587-4327587 on Build GRCh38
rs373836602 Chr.19: 4327232-4327232 on Build GRCh38
rs781071181 Chr.19: 4327409-4327409 on Build GRCh38
rs969518630 Chr.19: 4327685-4327685 on Build GRCh38
rs573371663 Chr.19: 4327460-4327460 on Build GRCh38
rs1046677360 Chr.19: 4327320-4327321 on Build GRCh38
rs200429772 Chr.19: 4327410-4327410 on Build GRCh38
rs989586218 Chr.19: 4327382-4327382 on Build GRCh38
rs765623298 Chr.19: 4327403-4327403 on Build GRCh38
rs776561071 Chr.19: 4327239-4327241 on Build GRCh38
rs564173215 Chr.19: 4327566-4327566 on Build GRCh38
rs774690523 Chr.19: 4327379-4327379 on Build GRCh38
rs780027297 Chr.19: 4327280-4327280 on Build GRCh38
rs768738306 Chr.19: 4327586-4327586 on Build GRCh38
rs916785707 Chr.19: 4327618-4327618 on Build GRCh38
rs769464271 Chr.19: 4327292-4327292 on Build GRCh38
rs776193640 Chr.19: 4327289-4327289 on Build GRCh38
rs563317053 Chr.19: 4327576-4327576 on Build GRCh38
rs746013225 Chr.19: 4327598-4327598 on Build GRCh38
rs757541521 Chr.19: 4327393-4327393 on Build GRCh38
rs765240563 Chr.19: 4327324-4327324 on Build GRCh38
rs768729713 Chr.19: 4327288-4327288 on Build GRCh38
rs143252691 Chr.19: 4327337-4327337 on Build GRCh38
rs1025077380 Chr.19: 4327485-4327485 on Build GRCh38
rs772691080 Chr.19: 4327293-4327293 on Build GRCh38
rs528857075 Chr.19: 4327592-4327592 on Build GRCh38
rs991178435 Chr.19: 4327613-4327613 on Build GRCh38
rs760020038 Chr.19: 4327309-4327309 on Build GRCh38
rs899223802 Chr.19: 4327504-4327504 on Build GRCh38
rs763188541 Chr.19: 4327405-4327405 on Build GRCh38
rs756357376 Chr.19: 4327236-4327236 on Build GRCh38
rs995092469 Chr.19: 4327517-4327517 on Build GRCh38
rs767978843 Chr.19: 4327313-4327313 on Build GRCh38
rs777934276 Chr.19: 4327316-4327316 on Build GRCh38
rs775725781 Chr.19: 4327403-4327406 on Build GRCh38
rs557988260 Chr.19: 4327360-4327360 on Build GRCh38
rs1018647505 Chr.19: 4327247-4327247 on Build GRCh38
rs555197098 Chr.19: 4327276-4327276 on Build GRCh38
rs932354280 Chr.19: 4327666-4327666 on Build GRCh38
rs771495477 Chr.19: 4327583-4327583 on Build GRCh38
rs922382161 Chr.19: 4327665-4327665 on Build GRCh38
rs369462863 Chr.19: 4327635-4327635 on Build GRCh38
rs751931964 Chr.19: 4327307-4327307 on Build GRCh38
rs549039536 Chr.19: 4327595-4327595 on Build GRCh38
rs967458345 Chr.19: 4327569-4327569 on Build GRCh38
rs1050279408 Chr.19: 4327518-4327518 on Build GRCh38
rs545554665 Chr.19: 4327540-4327540 on Build GRCh38
rs756087903 Chr.19: 4327412-4327412 on Build GRCh38
rs370588876 Chr.19: 4327402-4327402 on Build GRCh38
rs533017054 Chr.19: 4327568-4327568 on Build GRCh38
rs746877530 Chr.19: 4327283-4327283 on Build GRCh38
rs565933265 Chr.19: 4327633-4327633 on Build GRCh38
rs945581857 Chr.19: 4327646-4327646 on Build GRCh38
rs989726755 Chr.19: 4327631-4327631 on Build GRCh38
rs770625272 Chr.19: 4327362-4327362 on Build GRCh38

View Full Product Details