Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 4542560-4543064 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEMA6B
Entrez Gene ID
10501
Gene Name
semaphorin 6B
Gene Aliases
SEM-SEMA-Y, SEMA-VIB, SEMAN, semaZ
UniGene
Hs.465642
Gene Chromosome Location
Chr.19: 4542588-4581484 on Build GRCh38
Transcripts
NM_032108.3 XM_011527639.2 XM_011527640.2

SNP Detail

SNP ID Chromosome Location
rs960561930 Chr.19: 4542989-4542989 on Build GRCh38
rs1011957668 Chr.19: 4542631-4542631 on Build GRCh38
rs904441615 Chr.19: 4542927-4542927 on Build GRCh38
rs78237691 Chr.19: 4543036-4543036 on Build GRCh38
rs1041766433 Chr.19: 4542924-4542924 on Build GRCh38
rs983460534 Chr.19: 4542981-4542981 on Build GRCh38
rs1056127412 Chr.19: 4542945-4542945 on Build GRCh38
rs1000153266 Chr.19: 4542935-4542935 on Build GRCh38
rs894772009 Chr.19: 4542630-4542630 on Build GRCh38
rs752649533 Chr.19: 4542903-4542904 on Build GRCh38
rs982427013 Chr.19: 4542845-4542845 on Build GRCh38
rs757919197 Chr.19: 4542789-4542789 on Build GRCh38
rs886337046 Chr.19: 4542603-4542603 on Build GRCh38
rs770384038 Chr.19: 4542872-4542872 on Build GRCh38
rs983519337 Chr.19: 4542823-4542823 on Build GRCh38
rs779599144 Chr.19: 4542799-4542799 on Build GRCh38
rs1052189503 Chr.19: 4542591-4542591 on Build GRCh38
rs764952814 Chr.19: 4543011-4543011 on Build GRCh38
rs947687228 Chr.19: 4542888-4542888 on Build GRCh38
rs571595498 Chr.19: 4542620-4542620 on Build GRCh38
rs890425840 Chr.19: 4542607-4542607 on Build GRCh38
rs1000345721 Chr.19: 4542673-4542673 on Build GRCh38
rs964741001 Chr.19: 4542773-4542773 on Build GRCh38
rs748572597 Chr.19: 4542854-4542854 on Build GRCh38
rs553439373 Chr.19: 4542906-4542906 on Build GRCh38
rs920667355 Chr.19: 4542791-4542791 on Build GRCh38
rs963589547 Chr.19: 4543034-4543034 on Build GRCh38
rs1014522963 Chr.19: 4542955-4542955 on Build GRCh38
rs1022045486 Chr.19: 4542648-4542649 on Build GRCh38
rs746497059 Chr.19: 4542800-4542800 on Build GRCh38
rs1027589186 Chr.19: 4542967-4542967 on Build GRCh38
rs768395524 Chr.19: 4542811-4542811 on Build GRCh38
rs752822566 Chr.19: 4542992-4542992 on Build GRCh38
rs564914054 Chr.19: 4542928-4542928 on Build GRCh38
rs903632668 Chr.19: 4542652-4542652 on Build GRCh38
rs573221572 Chr.19: 4542910-4542910 on Build GRCh38
rs753714536 Chr.19: 4542713-4542713 on Build GRCh38
rs370559194 Chr.19: 4542857-4542857 on Build GRCh38
rs1031178252 Chr.19: 4542906-4542907 on Build GRCh38
rs374468538 Chr.19: 4542969-4542969 on Build GRCh38
rs916481763 Chr.19: 4543012-4543012 on Build GRCh38
rs866636165 Chr.19: 4542904-4542904 on Build GRCh38
rs375724234 Chr.19: 4542751-4542751 on Build GRCh38
rs911996832 Chr.19: 4542912-4542912 on Build GRCh38
rs397759133 Chr.19: 4542912-4542913 on Build GRCh38
rs761599854 Chr.19: 4543005-4543005 on Build GRCh38
rs778850977 Chr.19: 4542767-4542767 on Build GRCh38
rs573875723 Chr.19: 4542787-4542787 on Build GRCh38
rs747243893 Chr.19: 4542905-4542905 on Build GRCh38
rs62115199 Chr.19: 4542776-4542776 on Build GRCh38
rs973977385 Chr.19: 4542892-4542892 on Build GRCh38
rs149072027 Chr.19: 4542904-4542905 on Build GRCh38
rs748814540 Chr.19: 4542756-4542756 on Build GRCh38
rs907991736 Chr.19: 4542983-4542983 on Build GRCh38
rs1047584323 Chr.19: 4542584-4542584 on Build GRCh38
rs531142114 Chr.19: 4542966-4542966 on Build GRCh38
rs780690434 Chr.19: 4542816-4542816 on Build GRCh38
rs1056792187 Chr.19: 4542625-4542625 on Build GRCh38
rs894271488 Chr.19: 4542946-4542946 on Build GRCh38
rs143507855 Chr.19: 4542703-4542703 on Build GRCh38
rs542408969 Chr.19: 4542887-4542887 on Build GRCh38
rs774525347 Chr.19: 4542918-4542918 on Build GRCh38
rs1020187628 Chr.19: 4542660-4542660 on Build GRCh38
rs370376709 Chr.19: 4542911-4542911 on Build GRCh38
rs74173010 Chr.19: 4542783-4542784 on Build GRCh38
rs1031818202 Chr.19: 4542682-4542682 on Build GRCh38
rs958693703 Chr.19: 4542758-4542758 on Build GRCh38
rs1034228183 Chr.19: 4542757-4542757 on Build GRCh38
rs1000014149 Chr.19: 4542755-4542755 on Build GRCh38
rs756997393 Chr.19: 4542717-4542717 on Build GRCh38
rs987709273 Chr.19: 4542750-4542750 on Build GRCh38
rs770246647 Chr.19: 4542905-4542905 on Build GRCh38
rs992558270 Chr.19: 4542790-4542790 on Build GRCh38
rs567329709 Chr.19: 4542876-4542876 on Build GRCh38
rs879654306 Chr.19: 4542724-4542724 on Build GRCh38
rs557232502 Chr.19: 4542723-4542723 on Build GRCh38
rs750206933 Chr.19: 4543015-4543015 on Build GRCh38
rs377321891 Chr.19: 4542644-4542644 on Build GRCh38

View Full Product Details