Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 6468604-6469033 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENND1C
Entrez Gene ID
79958
Gene Name
DENN domain containing 1C
Gene Aliases
FAM31C
UniGene
Hs.236449
Gene Chromosome Location
Chr.19: 6467204-6481808 on Build GRCh38
Transcripts
NM_001290331.1 NM_024898.3 XM_006722905.2 XM_006722906.3 XM_011528318.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5857018 c.1525G>A p.A509T Chr.19: 6468914-6468914 on Build GRCh38
5580797 c.1430G>A p.R477K Chr.19: 6468931-6468931 on Build GRCh38
4676973 c.1487G>A p.R496H Chr.19: 6468874-6468874 on Build GRCh38
1397377 c.1514delG p.G505fs*3 Chr.19: 6468925-6468925 on Build GRCh38
6642276 c.1515C>G p.G505G Chr.19: 6468924-6468924 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs865787522 Chr.19: 6468888-6468888 on Build GRCh38
rs977510866 Chr.19: 6469006-6469006 on Build GRCh38
rs767511942 Chr.19: 6468838-6468838 on Build GRCh38
rs913774136 Chr.19: 6468733-6468733 on Build GRCh38
rs548905025 Chr.19: 6468632-6468632 on Build GRCh38
rs766394009 Chr.19: 6468886-6468886 on Build GRCh38
rs201868759 Chr.19: 6468974-6468974 on Build GRCh38
rs754805887 Chr.19: 6468895-6468895 on Build GRCh38
rs369454469 Chr.19: 6468663-6468663 on Build GRCh38
rs760807759 Chr.19: 6468944-6468944 on Build GRCh38
rs78367457 Chr.19: 6468829-6468829 on Build GRCh38
rs1000418310 Chr.19: 6468643-6468643 on Build GRCh38
rs535243417 Chr.19: 6468883-6468883 on Build GRCh38
rs760047759 Chr.19: 6468858-6468858 on Build GRCh38
rs780887724 Chr.19: 6468830-6468830 on Build GRCh38
rs764127124 Chr.19: 6468647-6468647 on Build GRCh38
rs765268365 Chr.19: 6468967-6468967 on Build GRCh38
rs139920554 Chr.19: 6468654-6468654 on Build GRCh38
rs1028190915 Chr.19: 6468985-6468985 on Build GRCh38
rs751547084 Chr.19: 6468968-6468968 on Build GRCh38
rs767965564 Chr.19: 6468861-6468861 on Build GRCh38
rs139457098 Chr.19: 6469001-6469001 on Build GRCh38
rs911962111 Chr.19: 6468880-6468880 on Build GRCh38
rs752967042 Chr.19: 6468689-6468689 on Build GRCh38
rs373068344 Chr.19: 6468716-6468716 on Build GRCh38
rs980525260 Chr.19: 6468659-6468659 on Build GRCh38
rs368056243 Chr.19: 6468926-6468926 on Build GRCh38
rs773631698 Chr.19: 6468641-6468641 on Build GRCh38
rs775749909 Chr.19: 6468924-6468925 on Build GRCh38
rs113599909 Chr.19: 6468948-6468948 on Build GRCh38
rs866837181 Chr.19: 6468623-6468623 on Build GRCh38
rs374543472 Chr.19: 6468911-6468911 on Build GRCh38
rs958341478 Chr.19: 6468760-6468760 on Build GRCh38
rs753884918 Chr.19: 6468882-6468882 on Build GRCh38
rs376952718 Chr.19: 6468875-6468875 on Build GRCh38
rs758311467 Chr.19: 6468812-6468812 on Build GRCh38
rs554182712 Chr.19: 6468958-6468958 on Build GRCh38
rs113149947 Chr.19: 6468837-6468837 on Build GRCh38
rs200632898 Chr.19: 6468835-6468835 on Build GRCh38
rs373180917 Chr.19: 6468668-6468668 on Build GRCh38
rs572041247 Chr.19: 6468932-6468932 on Build GRCh38
rs751885294 Chr.19: 6468656-6468656 on Build GRCh38
rs201009705 Chr.19: 6468627-6468627 on Build GRCh38
rs759853453 Chr.19: 6468662-6468662 on Build GRCh38
rs755726574 Chr.19: 6468904-6468904 on Build GRCh38
rs747820435 Chr.19: 6468900-6468900 on Build GRCh38
rs763494134 Chr.19: 6468642-6468642 on Build GRCh38
rs774988018 Chr.19: 6468845-6468845 on Build GRCh38
rs756305428 Chr.19: 6468690-6468690 on Build GRCh38
rs943917458 Chr.19: 6468752-6468754 on Build GRCh38
rs751493772 Chr.19: 6468887-6468887 on Build GRCh38
rs370958111 Chr.19: 6468833-6468833 on Build GRCh38
rs755849805 Chr.19: 6468991-6468991 on Build GRCh38
rs35001260 Chr.19: 6468896-6468896 on Build GRCh38
rs887851450 Chr.19: 6468860-6468860 on Build GRCh38
rs1024052163 Chr.19: 6468782-6468782 on Build GRCh38
rs769236013 Chr.19: 6468834-6468834 on Build GRCh38
rs770019826 Chr.19: 6468919-6468922 on Build GRCh38
rs533786331 Chr.19: 6468670-6468670 on Build GRCh38
rs547405926 Chr.19: 6468775-6468775 on Build GRCh38
rs375852555 Chr.19: 6468824-6468824 on Build GRCh38
rs987924650 Chr.19: 6468731-6468731 on Build GRCh38
rs983022632 Chr.19: 6468790-6468790 on Build GRCh38
rs931972381 Chr.19: 6468847-6468847 on Build GRCh38
rs746745810 Chr.19: 6468826-6468826 on Build GRCh38
rs1000878555 Chr.19: 6468890-6468890 on Build GRCh38
rs769341468 Chr.19: 6468633-6468633 on Build GRCh38
rs772591704 Chr.19: 6468931-6468931 on Build GRCh38
rs971294004 Chr.19: 6468785-6468785 on Build GRCh38
rs771413408 Chr.19: 6468841-6468841 on Build GRCh38
rs779715574 Chr.19: 6468816-6468816 on Build GRCh38
rs185066181 Chr.19: 6468999-6468999 on Build GRCh38
rs957779780 Chr.19: 6468936-6468936 on Build GRCh38
rs745871835 Chr.19: 6468839-6468839 on Build GRCh38
rs760906473 Chr.19: 6468874-6468874 on Build GRCh38
rs746358321 Chr.19: 6468930-6468930 on Build GRCh38
rs750144847 Chr.19: 6468866-6468866 on Build GRCh38
rs776791284 Chr.19: 6468689-6468690 on Build GRCh38
rs750353525 Chr.19: 6468695-6468695 on Build GRCh38
rs752431328 Chr.19: 6468986-6468986 on Build GRCh38
rs775041449 Chr.19: 6468929-6468929 on Build GRCh38
rs777891035 Chr.19: 6468694-6468694 on Build GRCh38
rs1042012931 Chr.19: 6468629-6468629 on Build GRCh38
rs944800881 Chr.19: 6469005-6469005 on Build GRCh38
rs199536511 Chr.19: 6468871-6468871 on Build GRCh38
rs911862860 Chr.19: 6469003-6469003 on Build GRCh38
rs777162523 Chr.19: 6468925-6468925 on Build GRCh38
rs762017737 Chr.19: 6468965-6468965 on Build GRCh38

View Full Product Details