Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12389915-12390463 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF799
Entrez Gene ID
90576
Gene Name
zinc finger protein 799
Gene Aliases
HIT-40, ZNF842
UniGene
Hs.546637
Gene Chromosome Location
Chr.19: 12390014-12401274 on Build GRCh38
Transcripts
NM_001080821.2 NM_001322497.1 NM_001322498.1 XM_017027452.1

SNP Detail

SNP ID Chromosome Location
rs547074774 Chr.19: 12390221-12390221 on Build GRCh38
rs368382295 Chr.19: 12390024-12390024 on Build GRCh38
rs546110669 Chr.19: 12390336-12390336 on Build GRCh38
rs186332341 Chr.19: 12389986-12389986 on Build GRCh38
rs190938698 Chr.19: 12390047-12390047 on Build GRCh38
rs1015595095 Chr.19: 12390393-12390393 on Build GRCh38
rs898667703 Chr.19: 12390398-12390398 on Build GRCh38
rs201936838 Chr.19: 12390280-12390280 on Build GRCh38
rs969790771 Chr.19: 12390238-12390238 on Build GRCh38
rs557511388 Chr.19: 12390009-12390009 on Build GRCh38
rs1004716024 Chr.19: 12390390-12390390 on Build GRCh38
rs543438120 Chr.19: 12390201-12390201 on Build GRCh38
rs995631402 Chr.19: 12390400-12390400 on Build GRCh38
rs185267882 Chr.19: 12390210-12390210 on Build GRCh38
rs934544209 Chr.19: 12390013-12390013 on Build GRCh38
rs1029782591 Chr.19: 12390074-12390074 on Build GRCh38
rs1134428 Chr.19: 12390126-12390126 on Build GRCh38
rs566306408 Chr.19: 12390407-12390407 on Build GRCh38
rs374519272 Chr.19: 12390433-12390433 on Build GRCh38
rs893081776 Chr.19: 12390020-12390020 on Build GRCh38
rs529489045 Chr.19: 12390289-12390289 on Build GRCh38
rs1052833359 Chr.19: 12390279-12390279 on Build GRCh38
rs996885947 Chr.19: 12390346-12390346 on Build GRCh38
rs397839868 Chr.19: 12390057-12390057 on Build GRCh38
rs974275107 Chr.19: 12389978-12389978 on Build GRCh38
rs948357758 Chr.19: 12390005-12390005 on Build GRCh38
rs934389086 Chr.19: 12390277-12390277 on Build GRCh38
rs371544329 Chr.19: 12390352-12390352 on Build GRCh38
rs1023208637 Chr.19: 12390403-12390403 on Build GRCh38
rs558433918 Chr.19: 12390364-12390364 on Build GRCh38
rs761114549 Chr.19: 12390208-12390208 on Build GRCh38
rs555083048 Chr.19: 12390021-12390021 on Build GRCh38
rs916126989 Chr.19: 12390169-12390169 on Build GRCh38
rs568592164 Chr.19: 12390246-12390246 on Build GRCh38
rs879650353 Chr.19: 12390314-12390314 on Build GRCh38
rs533109128 Chr.19: 12390124-12390124 on Build GRCh38
rs962821030 Chr.19: 12390132-12390132 on Build GRCh38
rs572892117 Chr.19: 12390012-12390012 on Build GRCh38
rs569525141 Chr.19: 12390342-12390342 on Build GRCh38
rs915593480 Chr.19: 12389982-12389982 on Build GRCh38
rs955628535 Chr.19: 12390103-12390104 on Build GRCh38
rs1011517273 Chr.19: 12390022-12390022 on Build GRCh38
rs201019469 Chr.19: 12390267-12390267 on Build GRCh38
rs532337597 Chr.19: 12390316-12390316 on Build GRCh38
rs573466052 Chr.19: 12390054-12390054 on Build GRCh38
rs562419467 Chr.19: 12390115-12390115 on Build GRCh38
rs2910433 Chr.19: 12390014-12390014 on Build GRCh38
rs563325908 Chr.19: 12390302-12390302 on Build GRCh38
rs706546 Chr.19: 12390244-12390244 on Build GRCh38
rs574832887 Chr.19: 12390121-12390121 on Build GRCh38
rs569601879 Chr.19: 12389979-12389979 on Build GRCh38
rs997252323 Chr.19: 12390071-12390071 on Build GRCh38
rs1004406489 Chr.19: 12390122-12390122 on Build GRCh38
rs180986673 Chr.19: 12390160-12390160 on Build GRCh38

View Full Product Details