Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 43404289-43404789 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SZT2
Entrez Gene ID
23334
Gene Name
seizure threshold 2 homolog (mouse)
Gene Aliases
C1orf84, EIEE18, KIAA0467, SZT2A, SZT2B
UniGene
Hs.643560
Gene Chromosome Location
Chr.1: 43389884-43454247 on Build GRCh38
Transcripts
NM_015284.3 XM_005270686.3 XM_006710501.3 XM_011541106.2 XM_017000819.1 XM_017000820.1 XM_017000821.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7227237 c.357T>C p.V119V Chr.1: 43404415-43404415 on Build GRCh38
269437 c.384C>T p.G128G Chr.1: 43404442-43404442 on Build GRCh38
7392871 c.362A>G p.H121R Chr.1: 43404420-43404420 on Build GRCh38
1738473 c.461A>T p.Q154L Chr.1: 43404513-43404513 on Build GRCh38
681304 c.436G>T p.E146* Chr.1: 43404494-43404494 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767516322 Chr.1: 43404436-43404436 on Build GRCh38
rs12036439 Chr.1: 43404329-43404329 on Build GRCh38
rs1015908246 Chr.1: 43404675-43404675 on Build GRCh38
rs1032229756 Chr.1: 43404628-43404628 on Build GRCh38
rs946042976 Chr.1: 43404757-43404757 on Build GRCh38
rs758311269 Chr.1: 43404462-43404462 on Build GRCh38
rs766399416 Chr.1: 43404551-43404551 on Build GRCh38
rs777574536 Chr.1: 43404350-43404350 on Build GRCh38
rs973368578 Chr.1: 43404690-43404690 on Build GRCh38
rs771488309 Chr.1: 43404374-43404374 on Build GRCh38
rs952708206 Chr.1: 43404700-43404700 on Build GRCh38
rs746223184 Chr.1: 43404715-43404715 on Build GRCh38
rs755751265 Chr.1: 43404582-43404582 on Build GRCh38
rs369633184 Chr.1: 43404741-43404741 on Build GRCh38
rs746392852 Chr.1: 43404520-43404520 on Build GRCh38
rs780787955 Chr.1: 43404576-43404576 on Build GRCh38
rs932679777 Chr.1: 43404762-43404762 on Build GRCh38
rs940116821 Chr.1: 43404725-43404725 on Build GRCh38
rs777000292 Chr.1: 43404680-43404680 on Build GRCh38
rs566220593 Chr.1: 43404378-43404378 on Build GRCh38
rs148786528 Chr.1: 43404507-43404507 on Build GRCh38
rs1009283713 Chr.1: 43404646-43404646 on Build GRCh38
rs570637110 Chr.1: 43404702-43404702 on Build GRCh38
rs907156500 Chr.1: 43404767-43404767 on Build GRCh38
rs1024719412 Chr.1: 43404662-43404662 on Build GRCh38
rs41312024 Chr.1: 43404400-43404400 on Build GRCh38
rs984347505 Chr.1: 43404455-43404455 on Build GRCh38
rs757232559 Chr.1: 43404355-43404355 on Build GRCh38
rs529198330 Chr.1: 43404415-43404415 on Build GRCh38
rs754537006 Chr.1: 43404565-43404565 on Build GRCh38
rs751178797 Chr.1: 43404561-43404561 on Build GRCh38
rs987024516 Chr.1: 43404697-43404700 on Build GRCh38
rs746219999 Chr.1: 43404596-43404596 on Build GRCh38
rs908636957 Chr.1: 43404457-43404457 on Build GRCh38
rs750195768 Chr.1: 43404541-43404541 on Build GRCh38
rs764964124 Chr.1: 43404537-43404537 on Build GRCh38
rs760925618 Chr.1: 43404443-43404443 on Build GRCh38
rs932647223 Chr.1: 43404556-43404556 on Build GRCh38
rs745451951 Chr.1: 43404366-43404366 on Build GRCh38
rs918728621 Chr.1: 43404368-43404368 on Build GRCh38
rs762038104 Chr.1: 43404534-43404534 on Build GRCh38
rs267598616 Chr.1: 43404511-43404511 on Build GRCh38
rs918855854 Chr.1: 43404693-43404693 on Build GRCh38
rs746489525 Chr.1: 43404487-43404487 on Build GRCh38
rs970418841 Chr.1: 43404666-43404666 on Build GRCh38
rs914552611 Chr.1: 43404747-43404747 on Build GRCh38
rs34426631 Chr.1: 43404431-43404432 on Build GRCh38
rs780984579 Chr.1: 43404501-43404501 on Build GRCh38
rs147519875 Chr.1: 43404420-43404420 on Build GRCh38
rs750341206 Chr.1: 43404458-43404458 on Build GRCh38
rs771367855 Chr.1: 43404674-43404674 on Build GRCh38
rs772600300 Chr.1: 43404523-43404523 on Build GRCh38
rs74069985 Chr.1: 43404723-43404723 on Build GRCh38
rs1032117227 Chr.1: 43404359-43404359 on Build GRCh38
rs776845092 Chr.1: 43404531-43404531 on Build GRCh38
rs976840318 Chr.1: 43404748-43404748 on Build GRCh38
rs955686163 Chr.1: 43404697-43404698 on Build GRCh38
rs375255354 Chr.1: 43404453-43404453 on Build GRCh38
rs756280392 Chr.1: 43404339-43404339 on Build GRCh38
rs531444734 Chr.1: 43404585-43404585 on Build GRCh38
rs865796643 Chr.1: 43404482-43404482 on Build GRCh38
rs572497343 Chr.1: 43404430-43404430 on Build GRCh38
rs764368395 Chr.1: 43404530-43404530 on Build GRCh38
rs779465575 Chr.1: 43404479-43404479 on Build GRCh38
rs747821314 Chr.1: 43404580-43404580 on Build GRCh38
rs540255054 Chr.1: 43404336-43404336 on Build GRCh38
rs551147929 Chr.1: 43404641-43404641 on Build GRCh38
rs753597423 Chr.1: 43404353-43404353 on Build GRCh38
rs1000399958 Chr.1: 43404616-43404616 on Build GRCh38
rs775594426 Chr.1: 43404527-43404527 on Build GRCh38
rs371591286 Chr.1: 43404433-43404433 on Build GRCh38
rs369520119 Chr.1: 43404514-43404514 on Build GRCh38
rs150074610 Chr.1: 43404431-43404431 on Build GRCh38
rs774721892 Chr.1: 43404421-43404421 on Build GRCh38
rs373503507 Chr.1: 43404759-43404759 on Build GRCh38
rs187131803 Chr.1: 43404442-43404442 on Build GRCh38
rs562799762 Chr.1: 43404444-43404444 on Build GRCh38
rs771210352 Chr.1: 43404513-43404513 on Build GRCh38
rs779563217 Chr.1: 43404390-43404390 on Build GRCh38
rs754729230 Chr.1: 43404497-43404497 on Build GRCh38
rs778704314 Chr.1: 43404356-43404356 on Build GRCh38
rs546985681 Chr.1: 43404763-43404763 on Build GRCh38
rs980715879 Chr.1: 43404432-43404432 on Build GRCh38
rs779184973 Chr.1: 43404584-43404584 on Build GRCh38
rs773753945 Chr.1: 43404409-43404409 on Build GRCh38
rs893044157 Chr.1: 43404456-43404456 on Build GRCh38
rs758310746 Chr.1: 43404548-43404548 on Build GRCh38

View Full Product Details